Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95037067-95037567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C8
Entrez Gene ID
1558
Gene Name
cytochrome P450 family 2 subfamily C member 8
Gene Aliases
CPC8, CYPIIC8, MP-12/MP-20
UniGene
Hs.709188
Gene Chromosome Location
Chr.10: 95036772-95069512 on Build GRCh38
Transcripts
NM_000770.3 NM_001198853.1 NM_001198854.1 NM_001198855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4442677 c.1091A>G p.N364S Chr.10: 95037204-95037204 on Build GRCh38
5608205 c.1464C>T p.I488I Chr.10: 95037137-95037137 on Build GRCh38
5758170 c.1018C>G p.R340G Chr.10: 95037277-95037277 on Build GRCh38
4929917 c.1083T>C p.D361D Chr.10: 95037212-95037212 on Build GRCh38
7306035 c.1001C>A p.A334E Chr.10: 95037294-95037294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764390443 Chr.10: 95037151-95037151 on Build GRCh38
rs1003204878 Chr.10: 95037096-95037096 on Build GRCh38
rs1038722024 Chr.10: 95037445-95037445 on Build GRCh38
rs200819252 Chr.10: 95037103-95037103 on Build GRCh38
rs766345387 Chr.10: 95037155-95037155 on Build GRCh38
rs140481138 Chr.10: 95037135-95037135 on Build GRCh38
rs771530542 Chr.10: 95037487-95037487 on Build GRCh38
rs762359438 Chr.10: 95037120-95037120 on Build GRCh38
rs748974036 Chr.10: 95037289-95037291 on Build GRCh38
rs993994200 Chr.10: 95037250-95037250 on Build GRCh38
rs868584215 Chr.10: 95037304-95037304 on Build GRCh38
rs779958772 Chr.10: 95037278-95037278 on Build GRCh38
rs760584366 Chr.10: 95037137-95037137 on Build GRCh38
rs1001618379 Chr.10: 95037508-95037509 on Build GRCh38
rs753802612 Chr.10: 95037195-95037195 on Build GRCh38
rs778978318 Chr.10: 95037156-95037156 on Build GRCh38
rs772969257 Chr.10: 95037113-95037113 on Build GRCh38
rs746564693 Chr.10: 95037473-95037473 on Build GRCh38
rs934131841 Chr.10: 95037411-95037411 on Build GRCh38
rs534467359 Chr.10: 95037491-95037491 on Build GRCh38
rs775683799 Chr.10: 95037524-95037527 on Build GRCh38
rs755161565 Chr.10: 95037295-95037295 on Build GRCh38
rs549598593 Chr.10: 95037242-95037242 on Build GRCh38
rs758345299 Chr.10: 95037171-95037171 on Build GRCh38
rs1015871520 Chr.10: 95037185-95037185 on Build GRCh38
rs965100375 Chr.10: 95037520-95037520 on Build GRCh38
rs757657903 Chr.10: 95037154-95037154 on Build GRCh38
rs762150516 Chr.10: 95037272-95037272 on Build GRCh38
rs868818549 Chr.10: 95037177-95037177 on Build GRCh38
rs781321835 Chr.10: 95037198-95037198 on Build GRCh38
rs939114328 Chr.10: 95037388-95037388 on Build GRCh38
rs769860444 Chr.10: 95037216-95037216 on Build GRCh38
rs1000911956 Chr.10: 95037216-95037217 on Build GRCh38
rs754717779 Chr.10: 95037216-95037216 on Build GRCh38
rs1040210990 Chr.10: 95037109-95037109 on Build GRCh38
rs747936556 Chr.10: 95037200-95037200 on Build GRCh38
rs756623714 Chr.10: 95037143-95037143 on Build GRCh38
rs1037602486 Chr.10: 95037470-95037470 on Build GRCh38
rs989486134 Chr.10: 95037537-95037537 on Build GRCh38
rs955161249 Chr.10: 95037498-95037498 on Build GRCh38
rs1051582714 Chr.10: 95037149-95037149 on Build GRCh38
rs1042533369 Chr.10: 95037345-95037345 on Build GRCh38
rs11572177 Chr.10: 95037513-95037513 on Build GRCh38
rs866496314 Chr.10: 95037294-95037294 on Build GRCh38
rs1016828529 Chr.10: 95037180-95037180 on Build GRCh38
rs772732311 Chr.10: 95037239-95037239 on Build GRCh38
rs754390283 Chr.10: 95037268-95037268 on Build GRCh38
rs551964175 Chr.10: 95037387-95037387 on Build GRCh38
rs3832694 Chr.10: 95037217-95037219 on Build GRCh38
rs894823642 Chr.10: 95037407-95037407 on Build GRCh38
rs186730985 Chr.10: 95037317-95037317 on Build GRCh38
rs376016142 Chr.10: 95037160-95037160 on Build GRCh38
rs61757318 Chr.10: 95037188-95037188 on Build GRCh38
rs142997544 Chr.10: 95037254-95037254 on Build GRCh38
rs769405632 Chr.10: 95037311-95037311 on Build GRCh38
rs764554009 Chr.10: 95037257-95037257 on Build GRCh38
rs770644728 Chr.10: 95037336-95037336 on Build GRCh38
rs1029509049 Chr.10: 95037493-95037493 on Build GRCh38
rs529725725 Chr.10: 95037187-95037187 on Build GRCh38
rs1017668308 Chr.10: 95037515-95037515 on Build GRCh38
rs1058932 Chr.10: 95037104-95037104 on Build GRCh38
rs928099015 Chr.10: 95037382-95037382 on Build GRCh38
rs748167187 Chr.10: 95037303-95037303 on Build GRCh38
rs769773346 Chr.10: 95037205-95037205 on Build GRCh38
rs959849063 Chr.10: 95037257-95037257 on Build GRCh38
rs11572178 Chr.10: 95037335-95037335 on Build GRCh38
rs138495387 Chr.10: 95037276-95037276 on Build GRCh38
rs866725748 Chr.10: 95037172-95037172 on Build GRCh38
rs145903792 Chr.10: 95037251-95037251 on Build GRCh38
rs773915472 Chr.10: 95037354-95037354 on Build GRCh38
rs780198250 Chr.10: 95037184-95037184 on Build GRCh38
rs10882519 Chr.10: 95037401-95037401 on Build GRCh38
rs537219541 Chr.10: 95037529-95037529 on Build GRCh38
rs748839363 Chr.10: 95037328-95037328 on Build GRCh38
rs369600584 Chr.10: 95037274-95037274 on Build GRCh38
rs770284698 Chr.10: 95037247-95037247 on Build GRCh38
rs371808080 Chr.10: 95037112-95037112 on Build GRCh38
rs1005200109 Chr.10: 95037432-95037432 on Build GRCh38
rs372999683 Chr.10: 95037288-95037288 on Build GRCh38
rs940585220 Chr.10: 95037464-95037464 on Build GRCh38
rs777320933 Chr.10: 95037452-95037452 on Build GRCh38

View Full Product Details