Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95606583-95607109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH18A1
Entrez Gene ID
5832
Gene Name
aldehyde dehydrogenase 18 family member A1
Gene Aliases
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
UniGene
Hs.500645
Gene Chromosome Location
Chr.10: 95605929-95656810 on Build GRCh38
Transcripts
NM_001017423.1 NM_001323412.1 NM_001323413.1 NM_001323414.1 NM_001323415.1 NM_001323416.1 NM_001323417.1 NM_001323418.1 NM_001323419.1 NM_002860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135074 c.2208A>C p.G736G Chr.10: 95606942-95606942 on Build GRCh38
278768 c.2347C>A p.L783I Chr.10: 95606803-95606803 on Build GRCh38
3808079 c.2248G>T p.G750* Chr.10: 95606902-95606902 on Build GRCh38
7131427 c.2246G>T p.R749L Chr.10: 95606904-95606904 on Build GRCh38
4440252 c.2249delG p.G750fs*3 Chr.10: 95606901-95606901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765661446 Chr.10: 95606840-95606840 on Build GRCh38
rs752649171 Chr.10: 95606977-95606977 on Build GRCh38
rs7099284 Chr.10: 95606989-95606989 on Build GRCh38
rs1037188823 Chr.10: 95606976-95606976 on Build GRCh38
rs374232219 Chr.10: 95606927-95606927 on Build GRCh38
rs765567477 Chr.10: 95606925-95606925 on Build GRCh38
rs751744859 Chr.10: 95606957-95606957 on Build GRCh38
rs537043237 Chr.10: 95606856-95606856 on Build GRCh38
rs496426 Chr.10: 95607077-95607077 on Build GRCh38
rs747072972 Chr.10: 95606867-95606867 on Build GRCh38
rs772994890 Chr.10: 95606780-95606780 on Build GRCh38
rs781195861 Chr.10: 95606969-95606969 on Build GRCh38
rs774047299 Chr.10: 95606805-95606805 on Build GRCh38
rs867708267 Chr.10: 95606777-95606777 on Build GRCh38
rs560951967 Chr.10: 95606732-95606732 on Build GRCh38
rs1047715712 Chr.10: 95606822-95606822 on Build GRCh38
rs960367311 Chr.10: 95606689-95606689 on Build GRCh38
rs144542561 Chr.10: 95606995-95606995 on Build GRCh38
rs548288287 Chr.10: 95606744-95606744 on Build GRCh38
rs76824727 Chr.10: 95606622-95606622 on Build GRCh38
rs748925635 Chr.10: 95606904-95606904 on Build GRCh38
rs747918055 Chr.10: 95606767-95606767 on Build GRCh38
rs121434583 Chr.10: 95606800-95606800 on Build GRCh38
rs762271422 Chr.10: 95606919-95606919 on Build GRCh38
rs563231416 Chr.10: 95606645-95606645 on Build GRCh38
rs770655744 Chr.10: 95606905-95606905 on Build GRCh38
rs552533320 Chr.10: 95607069-95607069 on Build GRCh38
rs1016708525 Chr.10: 95606619-95606619 on Build GRCh38
rs764336997 Chr.10: 95606721-95606721 on Build GRCh38
rs763340722 Chr.10: 95606930-95606930 on Build GRCh38
rs368872834 Chr.10: 95606734-95606734 on Build GRCh38
rs779053250 Chr.10: 95606730-95606730 on Build GRCh38
rs201876082 Chr.10: 95606754-95606754 on Build GRCh38
rs748094627 Chr.10: 95606892-95606892 on Build GRCh38
rs553385013 Chr.10: 95606821-95606821 on Build GRCh38
rs572568700 Chr.10: 95606997-95606997 on Build GRCh38
rs377506019 Chr.10: 95606789-95606789 on Build GRCh38
rs765966176 Chr.10: 95606799-95606799 on Build GRCh38
rs114393346 Chr.10: 95606706-95606706 on Build GRCh38
rs759016790 Chr.10: 95606909-95606909 on Build GRCh38
rs148601288 Chr.10: 95606918-95606918 on Build GRCh38
rs11592809 Chr.10: 95607060-95607060 on Build GRCh38
rs997892959 Chr.10: 95606688-95606688 on Build GRCh38
rs369153920 Chr.10: 95606842-95606842 on Build GRCh38
rs769481001 Chr.10: 95606778-95606778 on Build GRCh38
rs149309642 Chr.10: 95606946-95606946 on Build GRCh38
rs755019667 Chr.10: 95606872-95606872 on Build GRCh38
rs758671182 Chr.10: 95606846-95606846 on Build GRCh38
rs753243343 Chr.10: 95606719-95606719 on Build GRCh38
rs766054778 Chr.10: 95606714-95606714 on Build GRCh38
rs565757058 Chr.10: 95606724-95606724 on Build GRCh38
rs979620706 Chr.10: 95606741-95606741 on Build GRCh38
rs781126562 Chr.10: 95606874-95606874 on Build GRCh38
rs776453485 Chr.10: 95606765-95606765 on Build GRCh38
rs773903524 Chr.10: 95606908-95606908 on Build GRCh38
rs896221238 Chr.10: 95606970-95606970 on Build GRCh38
rs1002367939 Chr.10: 95606933-95606933 on Build GRCh38
rs757587417 Chr.10: 95606726-95606726 on Build GRCh38
rs1049396120 Chr.10: 95607055-95607055 on Build GRCh38
rs754301597 Chr.10: 95606820-95606820 on Build GRCh38
rs1001935271 Chr.10: 95606687-95606687 on Build GRCh38
rs543458427 Chr.10: 95606627-95606627 on Build GRCh38
rs763942723 Chr.10: 95606616-95606616 on Build GRCh38
rs1053905 Chr.10: 95606607-95606607 on Build GRCh38
rs184457864 Chr.10: 95606720-95606720 on Build GRCh38
rs762484852 Chr.10: 95606713-95606713 on Build GRCh38
rs773180798 Chr.10: 95606901-95606901 on Build GRCh38
rs769707204 Chr.10: 95606897-95606897 on Build GRCh38
rs748453966 Chr.10: 95607000-95607000 on Build GRCh38
rs143874727 Chr.10: 95606815-95606815 on Build GRCh38

View Full Product Details