Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95756560-95757069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD1
Entrez Gene ID
953
Gene Name
ectonucleoside triphosphate diphosphohydrolase 1
Gene Aliases
ATPDase, CD39, NTPDase-1, SPG64
UniGene
Hs.576612
Gene Chromosome Location
Chr.10: 95710901-95877266 on Build GRCh38
Transcripts
NM_001098175.1 NM_001164178.1 NM_001164179.1 NM_001164181.1 NM_001164182.1 NM_001164183.1 NM_001312654.1 NM_001320916.1 NM_001776.5 XM_011540370.2 XM_011540371.2 XM_011540372.2 XM_011540373.2 XM_011540374.2 XM_011540376.2 XM_011540377.2 XM_017016958.1 XM_017016959.1 XM_017016960.1 XM_017016961.1 XM_017016962.1 XM_017016963.1 XM_017016964.1 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001098175.1 NM_001164178.1 NM_001164179.1 NM_001164181.1 NM_001164182.1 NM_001164183.1 NM_001312654.1 NM_001320916.1 NM_001776.5 XM_011540370.2 XM_011540371.2 XM_011540372.2 XM_011540373.2 XM_011540374.2 XM_011540376.2 XM_011540377.2 XM_017016958.1 XM_017016959.1 XM_017016960.1 XM_017016961.1 XM_017016962.1 XM_017016963.1 XM_017016964.1 NR_038444.1

SNP Detail

SNP ID Chromosome Location
rs959226549 Chr.10: 95756940-95756940 on Build GRCh38
rs12262943 Chr.10: 95756687-95756687 on Build GRCh38
rs541124533 Chr.10: 95756718-95756718 on Build GRCh38
rs556934166 Chr.10: 95756616-95756616 on Build GRCh38
rs1018799242 Chr.10: 95756737-95756737 on Build GRCh38
rs959401438 Chr.10: 95756661-95756661 on Build GRCh38
rs1022256749 Chr.10: 95756681-95756681 on Build GRCh38
rs750254614 Chr.10: 95756895-95756895 on Build GRCh38
rs919534623 Chr.10: 95756711-95756711 on Build GRCh38
rs757286622 Chr.10: 95756755-95756755 on Build GRCh38
rs986414789 Chr.10: 95756691-95756691 on Build GRCh38
rs62625035 Chr.10: 95756644-95756644 on Build GRCh38
rs889206186 Chr.10: 95756591-95756591 on Build GRCh38
rs934809012 Chr.10: 95756922-95756922 on Build GRCh38
rs1003423584 Chr.10: 95756899-95756899 on Build GRCh38
rs765457803 Chr.10: 95756653-95756653 on Build GRCh38
rs376507394 Chr.10: 95756634-95756634 on Build GRCh38
rs910698695 Chr.10: 95756739-95756739 on Build GRCh38
rs558686142 Chr.10: 95756650-95756650 on Build GRCh38
rs961101160 Chr.10: 95756999-95756999 on Build GRCh38
rs915542764 Chr.10: 95756843-95756844 on Build GRCh38
rs766733646 Chr.10: 95756607-95756607 on Build GRCh38
rs930350478 Chr.10: 95756756-95756756 on Build GRCh38
rs984412566 Chr.10: 95756843-95756843 on Build GRCh38
rs563720538 Chr.10: 95756987-95756987 on Build GRCh38
rs74671500 Chr.10: 95756855-95756855 on Build GRCh38
rs1007489817 Chr.10: 95757046-95757046 on Build GRCh38
rs753031329 Chr.10: 95756811-95756825 on Build GRCh38
rs552506389 Chr.10: 95757011-95757011 on Build GRCh38
rs543417252 Chr.10: 95756983-95756983 on Build GRCh38
rs983805775 Chr.10: 95756671-95756671 on Build GRCh38
rs10748643 Chr.10: 95757007-95757007 on Build GRCh38
rs906450974 Chr.10: 95756897-95756897 on Build GRCh38
rs181951626 Chr.10: 95756647-95756647 on Build GRCh38
rs574761274 Chr.10: 95756884-95756884 on Build GRCh38
rs1009560258 Chr.10: 95756625-95756625 on Build GRCh38
rs909622468 Chr.10: 95756682-95756682 on Build GRCh38
rs943448869 Chr.10: 95756801-95756801 on Build GRCh38
rs1011635369 Chr.10: 95757017-95757017 on Build GRCh38
rs987714137 Chr.10: 95757039-95757039 on Build GRCh38
rs1053903126 Chr.10: 95756849-95756849 on Build GRCh38
rs1052364851 Chr.10: 95756738-95756738 on Build GRCh38
rs1021314396 Chr.10: 95756640-95756640 on Build GRCh38
rs377670918 Chr.10: 95756694-95756694 on Build GRCh38
rs893785583 Chr.10: 95756619-95756619 on Build GRCh38
rs933551717 Chr.10: 95756736-95756736 on Build GRCh38
rs761067947 Chr.10: 95756604-95756604 on Build GRCh38
rs771067775 Chr.10: 95757033-95757033 on Build GRCh38
rs1009610750 Chr.10: 95756857-95756857 on Build GRCh38
rs779791620 Chr.10: 95757010-95757010 on Build GRCh38
rs1005322643 Chr.10: 95756986-95756986 on Build GRCh38
rs1033942612 Chr.10: 95756903-95756903 on Build GRCh38
rs918988597 Chr.10: 95756750-95756750 on Build GRCh38

View Full Product Details