Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97224072-97224620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP19
Entrez Gene ID
84986
Gene Name
Rho GTPase activating protein 19
Gene Aliases
-
UniGene
Hs.80305
Gene Chromosome Location
Chr.10: 97222173-97292673 on Build GRCh38
Transcripts
NM_001204300.1 NM_001256423.1 NM_032900.5 NR_037909.1

Gene Symbol
ARHGAP19-SLIT1
Entrez Gene ID
100533184
Gene Name
ARHGAP19-SLIT1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80305
Gene Chromosome Location
Chr.10: 97153042-97292673 on Build GRCh38
Transcripts
NM_001204300.1 NM_001256423.1 NM_032900.5 NR_037909.1

SNP Detail

SNP ID Chromosome Location
rs948611165 Chr.10: 97224458-97224458 on Build GRCh38
rs988352471 Chr.10: 97224118-97224118 on Build GRCh38
rs371019075 Chr.10: 97224518-97224518 on Build GRCh38
rs1048057274 Chr.10: 97224554-97224554 on Build GRCh38
rs553723917 Chr.10: 97224559-97224559 on Build GRCh38
rs1002644237 Chr.10: 97224396-97224396 on Build GRCh38
rs946410438 Chr.10: 97224165-97224165 on Build GRCh38
rs988078870 Chr.10: 97224517-97224517 on Build GRCh38
rs571904538 Chr.10: 97224570-97224570 on Build GRCh38
rs566255524 Chr.10: 97224209-97224209 on Build GRCh38
rs954901377 Chr.10: 97224369-97224369 on Build GRCh38
rs374836395 Chr.10: 97224103-97224103 on Build GRCh38
rs201234130 Chr.10: 97224299-97224299 on Build GRCh38
rs12255419 Chr.10: 97224555-97224555 on Build GRCh38
rs960885171 Chr.10: 97224440-97224440 on Build GRCh38
rs1017013966 Chr.10: 97224335-97224336 on Build GRCh38
rs975034426 Chr.10: 97224295-97224295 on Build GRCh38
rs984449563 Chr.10: 97224224-97224224 on Build GRCh38
rs564209506 Chr.10: 97224254-97224254 on Build GRCh38
rs10748691 Chr.10: 97224198-97224198 on Build GRCh38
rs936795934 Chr.10: 97224384-97224384 on Build GRCh38
rs573894622 Chr.10: 97224285-97224285 on Build GRCh38
rs979184262 Chr.10: 97224171-97224171 on Build GRCh38
rs374327335 Chr.10: 97224417-97224417 on Build GRCh38
rs887282565 Chr.10: 97224302-97224302 on Build GRCh38
rs923408253 Chr.10: 97224330-97224330 on Build GRCh38
rs980711557 Chr.10: 97224100-97224100 on Build GRCh38
rs924040943 Chr.10: 97224375-97224375 on Build GRCh38
rs1005584188 Chr.10: 97224321-97224321 on Build GRCh38
rs916610728 Chr.10: 97224431-97224431 on Build GRCh38
rs950331996 Chr.10: 97224211-97224211 on Build GRCh38
rs910329783 Chr.10: 97224592-97224592 on Build GRCh38
rs1015978917 Chr.10: 97224232-97224232 on Build GRCh38
rs753319357 Chr.10: 97224111-97224111 on Build GRCh38
rs1006001767 Chr.10: 97224590-97224590 on Build GRCh38
rs1003012512 Chr.10: 97224121-97224121 on Build GRCh38
rs188626583 Chr.10: 97224558-97224558 on Build GRCh38
rs1047177324 Chr.10: 97224265-97224265 on Build GRCh38
rs752464588 Chr.10: 97224480-97224480 on Build GRCh38
rs905183273 Chr.10: 97224336-97224336 on Build GRCh38
rs1044338639 Chr.10: 97224470-97224470 on Build GRCh38
rs760310686 Chr.10: 97224514-97224514 on Build GRCh38
rs910213678 Chr.10: 97224197-97224197 on Build GRCh38
rs913755724 Chr.10: 97224141-97224141 on Build GRCh38
rs12255418 Chr.10: 97224540-97224540 on Build GRCh38
rs978547999 Chr.10: 97224374-97224374 on Build GRCh38
rs375921518 Chr.10: 97224105-97224105 on Build GRCh38
rs570323060 Chr.10: 97224510-97224510 on Build GRCh38

View Full Product Details