Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97319586-97320101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRAT1
Entrez Gene ID
10023
Gene Name
frequently rearranged in advanced T-cell lymphomas 1
Gene Aliases
-
UniGene
Hs.126057
Gene Chromosome Location
Chr.10: 97319265-97321915 on Build GRCh38
Transcripts
NM_005479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6361236 c.452G>A p.G151E Chr.10: 97319905-97319905 on Build GRCh38
4769816 c.476G>T p.G159V Chr.10: 97319929-97319929 on Build GRCh38
6262775 c.160G>T p.D54Y Chr.10: 97319613-97319613 on Build GRCh38
6518157 c.543C>A p.R181R Chr.10: 97319996-97319996 on Build GRCh38
6457016 c.142G>A p.G48S Chr.10: 97319595-97319595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557452732 Chr.10: 97319698-97319698 on Build GRCh38
rs991163797 Chr.10: 97319703-97319703 on Build GRCh38
rs761302783 Chr.10: 97319910-97319910 on Build GRCh38
rs572941311 Chr.10: 97319934-97319934 on Build GRCh38
rs868228044 Chr.10: 97320031-97320031 on Build GRCh38
rs949793145 Chr.10: 97319714-97319714 on Build GRCh38
rs554985083 Chr.10: 97319979-97319979 on Build GRCh38
rs571328245 Chr.10: 97319632-97319632 on Build GRCh38
rs762452955 Chr.10: 97319741-97319741 on Build GRCh38
rs747856495 Chr.10: 97320047-97320047 on Build GRCh38
rs113340568 Chr.10: 97319673-97319673 on Build GRCh38
rs758451050 Chr.10: 97320014-97320014 on Build GRCh38
rs112575281 Chr.10: 97319637-97319637 on Build GRCh38
rs926894366 Chr.10: 97319953-97319953 on Build GRCh38
rs765330063 Chr.10: 97320010-97320010 on Build GRCh38
rs746912110 Chr.10: 97320035-97320035 on Build GRCh38
rs932653467 Chr.10: 97320050-97320050 on Build GRCh38
rs961969391 Chr.10: 97319669-97319669 on Build GRCh38
rs538845227 Chr.10: 97319650-97319650 on Build GRCh38
rs750539686 Chr.10: 97320012-97320012 on Build GRCh38
rs186819363 Chr.10: 97319801-97319801 on Build GRCh38
rs754106585 Chr.10: 97319994-97319994 on Build GRCh38
rs932064245 Chr.10: 97319782-97319782 on Build GRCh38
rs897753571 Chr.10: 97319894-97319894 on Build GRCh38
rs958564653 Chr.10: 97319971-97319971 on Build GRCh38
rs916988203 Chr.10: 97319712-97319712 on Build GRCh38
rs867305174 Chr.10: 97320063-97320063 on Build GRCh38
rs996241089 Chr.10: 97319644-97319644 on Build GRCh38
rs1008892943 Chr.10: 97319659-97319659 on Build GRCh38
rs549349375 Chr.10: 97319956-97319956 on Build GRCh38
rs761169300 Chr.10: 97319991-97319991 on Build GRCh38
rs3740516 Chr.10: 97319647-97319647 on Build GRCh38
rs879162450 Chr.10: 97320038-97320038 on Build GRCh38
rs764299948 Chr.10: 97319992-97319992 on Build GRCh38
rs890419595 Chr.10: 97319649-97319649 on Build GRCh38
rs769357845 Chr.10: 97319925-97319925 on Build GRCh38
rs776407463 Chr.10: 97319817-97319817 on Build GRCh38
rs1053978668 Chr.10: 97319775-97319775 on Build GRCh38
rs973657926 Chr.10: 97319677-97319677 on Build GRCh38
rs1053947947 Chr.10: 97319618-97319618 on Build GRCh38
rs375567122 Chr.10: 97319975-97319975 on Build GRCh38
rs748046530 Chr.10: 97320038-97320040 on Build GRCh38
rs1050535210 Chr.10: 97319792-97319792 on Build GRCh38
rs757374076 Chr.10: 97319999-97319999 on Build GRCh38
rs924214843 Chr.10: 97319737-97319737 on Build GRCh38
rs1029001684 Chr.10: 97319645-97319645 on Build GRCh38
rs780024440 Chr.10: 97320030-97320030 on Build GRCh38
rs754803073 Chr.10: 97320039-97320039 on Build GRCh38
rs992345502 Chr.10: 97319976-97319976 on Build GRCh38
rs908610043 Chr.10: 97319978-97319978 on Build GRCh38
rs769374955 Chr.10: 97320071-97320071 on Build GRCh38
rs776131347 Chr.10: 97319987-97319987 on Build GRCh38
rs924279857 Chr.10: 97320072-97320072 on Build GRCh38
rs569368961 Chr.10: 97319722-97319722 on Build GRCh38
rs536781114 Chr.10: 97319959-97319959 on Build GRCh38
rs779932909 Chr.10: 97319697-97319697 on Build GRCh38
rs1015296160 Chr.10: 97319667-97319667 on Build GRCh38
rs772812314 Chr.10: 97319958-97319958 on Build GRCh38
rs1027420801 Chr.10: 97319682-97319682 on Build GRCh38
rs1014896917 Chr.10: 97319887-97319887 on Build GRCh38
rs562965940 Chr.10: 97320060-97320060 on Build GRCh38
rs935571194 Chr.10: 97319765-97319765 on Build GRCh38
rs376349131 Chr.10: 97320046-97320046 on Build GRCh38
rs1009337160 Chr.10: 97319812-97319812 on Build GRCh38
rs1050311595 Chr.10: 97320054-97320054 on Build GRCh38
rs768320093 Chr.10: 97319816-97319816 on Build GRCh38

View Full Product Details