Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97321063-97321571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRAT1
Entrez Gene ID
10023
Gene Name
frequently rearranged in advanced T-cell lymphomas 1
Gene Aliases
-
UniGene
Hs.126057
Gene Chromosome Location
Chr.10: 97319265-97321915 on Build GRCh38
Transcripts
NM_005479.3

SNP Detail

SNP ID Chromosome Location
rs1028000559 Chr.10: 97321220-97321220 on Build GRCh38
rs1007054172 Chr.10: 97321530-97321530 on Build GRCh38
rs889934381 Chr.10: 97321244-97321244 on Build GRCh38
rs889487703 Chr.10: 97321223-97321223 on Build GRCh38
rs552536377 Chr.10: 97321239-97321239 on Build GRCh38
rs538127991 Chr.10: 97321295-97321295 on Build GRCh38
rs371880589 Chr.10: 97321230-97321230 on Build GRCh38
rs770075501 Chr.10: 97321481-97321481 on Build GRCh38
rs897403263 Chr.10: 97321228-97321228 on Build GRCh38
rs1023031566 Chr.10: 97321477-97321477 on Build GRCh38
rs1036681888 Chr.10: 97321439-97321439 on Build GRCh38
rs111329810 Chr.10: 97321234-97321234 on Build GRCh38
rs990622228 Chr.10: 97321421-97321421 on Build GRCh38
rs548660817 Chr.10: 97321180-97321180 on Build GRCh38
rs1334892 Chr.10: 97321224-97321224 on Build GRCh38
rs979214574 Chr.10: 97321117-97321117 on Build GRCh38
rs914374628 Chr.10: 97321314-97321314 on Build GRCh38
rs937861963 Chr.10: 97321149-97321149 on Build GRCh38
rs375818457 Chr.10: 97321275-97321275 on Build GRCh38
rs1052959769 Chr.10: 97321298-97321298 on Build GRCh38
rs1054780668 Chr.10: 97321208-97321208 on Build GRCh38
rs1019674355 Chr.10: 97321263-97321263 on Build GRCh38
rs965878785 Chr.10: 97321266-97321266 on Build GRCh38
rs534078365 Chr.10: 97321238-97321238 on Build GRCh38
rs970535281 Chr.10: 97321482-97321482 on Build GRCh38
rs896229390 Chr.10: 97321213-97321213 on Build GRCh38
rs988760900 Chr.10: 97321529-97321529 on Build GRCh38
rs981831863 Chr.10: 97321284-97321284 on Build GRCh38
rs981479955 Chr.10: 97321484-97321484 on Build GRCh38
rs535664450 Chr.10: 97321366-97321366 on Build GRCh38
rs1006343498 Chr.10: 97321464-97321464 on Build GRCh38
rs930984948 Chr.10: 97321480-97321480 on Build GRCh38
rs1007065624 Chr.10: 97321247-97321247 on Build GRCh38
rs540154788 Chr.10: 97321357-97321357 on Build GRCh38
rs564872851 Chr.10: 97321509-97321509 on Build GRCh38
rs1020725882 Chr.10: 97321337-97321337 on Build GRCh38
rs955989777 Chr.10: 97321513-97321513 on Build GRCh38
rs966680700 Chr.10: 97321231-97321231 on Build GRCh38
rs984808682 Chr.10: 97321240-97321240 on Build GRCh38
rs925149316 Chr.10: 97321125-97321125 on Build GRCh38
rs1011290704 Chr.10: 97321199-97321199 on Build GRCh38
rs892905141 Chr.10: 97321142-97321142 on Build GRCh38
rs928599992 Chr.10: 97321294-97321294 on Build GRCh38
rs539985917 Chr.10: 97321437-97321437 on Build GRCh38
rs947196404 Chr.10: 97321322-97321322 on Build GRCh38
rs1334891 Chr.10: 97321114-97321114 on Build GRCh38
rs11189129 Chr.10: 97321360-97321360 on Build GRCh38
rs1020239946 Chr.10: 97321545-97321545 on Build GRCh38
rs550919413 Chr.10: 97321100-97321100 on Build GRCh38
rs995787410 Chr.10: 97321237-97321237 on Build GRCh38
rs949047058 Chr.10: 97321221-97321221 on Build GRCh38
rs751552664 Chr.10: 97321291-97321291 on Build GRCh38
rs937809461 Chr.10: 97321386-97321386 on Build GRCh38
rs1019215732 Chr.10: 97321229-97321229 on Build GRCh38
rs376436649 Chr.10: 97321207-97321207 on Build GRCh38
rs370215134 Chr.10: 97321445-97321445 on Build GRCh38
rs529930770 Chr.10: 97321129-97321129 on Build GRCh38
rs1041079635 Chr.10: 97321390-97321390 on Build GRCh38
rs915989533 Chr.10: 97321258-97321258 on Build GRCh38
rs576512169 Chr.10: 97321406-97321406 on Build GRCh38
rs538457585 Chr.10: 97321426-97321426 on Build GRCh38
rs554322667 Chr.10: 97321379-97321379 on Build GRCh38
rs928603148 Chr.10: 97321508-97321508 on Build GRCh38
rs1048637889 Chr.10: 97321502-97321502 on Build GRCh38
rs902109208 Chr.10: 97321415-97321415 on Build GRCh38

View Full Product Details