Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97437158-97437670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC1
Entrez Gene ID
51013
Gene Name
exosome component 1
Gene Aliases
CGI-108, CSL4, Csl4p, SKI4, Ski4p, p13
UniGene
Hs.632089
Gene Chromosome Location
Chr.10: 97435909-97446065 on Build GRCh38
Transcripts
NM_001318362.1 NM_001318363.1 NM_001318364.1 NM_001318365.1 NM_001318366.1 NM_016046.4 NR_134586.1 NR_134587.1 XM_011539847.2 XM_017016304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
922021 c.468C>A p.A156A Chr.10: 97437204-97437204 on Build GRCh38
3442421 c.469C>T p.H157Y Chr.10: 97437203-97437203 on Build GRCh38
922022 c.447C>T p.N149N Chr.10: 97437225-97437225 on Build GRCh38
6657270 c.472A>G p.S158G Chr.10: 97437200-97437200 on Build GRCh38
4399307 c.402C>T p.S134S Chr.10: 97437270-97437270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757023510 Chr.10: 97437238-97437238 on Build GRCh38
rs141692186 Chr.10: 97437395-97437395 on Build GRCh38
rs754445779 Chr.10: 97437264-97437264 on Build GRCh38
rs373789172 Chr.10: 97437645-97437645 on Build GRCh38
rs774453483 Chr.10: 97437220-97437220 on Build GRCh38
rs775749388 Chr.10: 97437314-97437314 on Build GRCh38
rs937323832 Chr.10: 97437386-97437386 on Build GRCh38
rs748377518 Chr.10: 97437604-97437604 on Build GRCh38
rs927378117 Chr.10: 97437515-97437515 on Build GRCh38
rs3763672 Chr.10: 97437516-97437516 on Build GRCh38
rs780339590 Chr.10: 97437541-97437541 on Build GRCh38
rs542138788 Chr.10: 97437505-97437505 on Build GRCh38
rs148789012 Chr.10: 97437233-97437233 on Build GRCh38
rs1006556671 Chr.10: 97437460-97437460 on Build GRCh38
rs185609086 Chr.10: 97437391-97437391 on Build GRCh38
rs376945457 Chr.10: 97437631-97437631 on Build GRCh38
rs896912055 Chr.10: 97437418-97437418 on Build GRCh38
rs201847471 Chr.10: 97437508-97437508 on Build GRCh38
rs752740510 Chr.10: 97437412-97437412 on Build GRCh38
rs760283317 Chr.10: 97437250-97437250 on Build GRCh38
rs750550144 Chr.10: 97437504-97437504 on Build GRCh38
rs111826501 Chr.10: 97437292-97437292 on Build GRCh38
rs35614097 Chr.10: 97437229-97437229 on Build GRCh38
rs759740619 Chr.10: 97437225-97437225 on Build GRCh38
rs913369139 Chr.10: 97437476-97437476 on Build GRCh38
rs558802679 Chr.10: 97437200-97437200 on Build GRCh38
rs193222710 Chr.10: 97437368-97437368 on Build GRCh38
rs1026717916 Chr.10: 97437440-97437440 on Build GRCh38
rs1048208708 Chr.10: 97437607-97437607 on Build GRCh38
rs758335780 Chr.10: 97437450-97437450 on Build GRCh38
rs776721161 Chr.10: 97437321-97437321 on Build GRCh38
rs201899078 Chr.10: 97437245-97437245 on Build GRCh38
rs546554549 Chr.10: 97437567-97437567 on Build GRCh38
rs754265523 Chr.10: 97437322-97437322 on Build GRCh38
rs371679381 Chr.10: 97437291-97437291 on Build GRCh38
rs879117300 Chr.10: 97437356-97437356 on Build GRCh38
rs868589966 Chr.10: 97437472-97437472 on Build GRCh38
rs187225803 Chr.10: 97437405-97437405 on Build GRCh38
rs377237874 Chr.10: 97437234-97437234 on Build GRCh38
rs746131503 Chr.10: 97437295-97437295 on Build GRCh38
rs1002642834 Chr.10: 97437358-97437358 on Build GRCh38
rs545030395 Chr.10: 97437310-97437310 on Build GRCh38
rs560441413 Chr.10: 97437509-97437509 on Build GRCh38
rs1020849527 Chr.10: 97437407-97437407 on Build GRCh38
rs147866756 Chr.10: 97437326-97437326 on Build GRCh38
rs1020295600 Chr.10: 97437462-97437462 on Build GRCh38
rs758057645 Chr.10: 97437259-97437259 on Build GRCh38
rs200628663 Chr.10: 97437411-97437411 on Build GRCh38
rs767557166 Chr.10: 97437227-97437227 on Build GRCh38
rs968842209 Chr.10: 97437491-97437491 on Build GRCh38
rs761467590 Chr.10: 97437273-97437274 on Build GRCh38
rs926722035 Chr.10: 97437572-97437572 on Build GRCh38
rs781171413 Chr.10: 97437473-97437473 on Build GRCh38
rs1017801330 Chr.10: 97437420-97437420 on Build GRCh38
rs147061936 Chr.10: 97437421-97437421 on Build GRCh38
rs1019255210 Chr.10: 97437641-97437641 on Build GRCh38
rs779588843 Chr.10: 97437260-97437260 on Build GRCh38
rs767060799 Chr.10: 97437313-97437313 on Build GRCh38
rs1037344345 Chr.10: 97437568-97437568 on Build GRCh38
rs941542916 Chr.10: 97437557-97437557 on Build GRCh38
rs1000675822 Chr.10: 97437415-97437415 on Build GRCh38
rs746494191 Chr.10: 97437261-97437261 on Build GRCh38
rs905245208 Chr.10: 97437356-97437356 on Build GRCh38
rs771537397 Chr.10: 97437222-97437222 on Build GRCh38
rs78713835 Chr.10: 97437221-97437221 on Build GRCh38
rs775694035 Chr.10: 97437230-97437230 on Build GRCh38
rs771280526 Chr.10: 97437202-97437202 on Build GRCh38
rs201183350 Chr.10: 97437231-97437231 on Build GRCh38
rs577629564 Chr.10: 97437529-97437529 on Build GRCh38
rs554768056 Chr.10: 97437387-97437387 on Build GRCh38
rs75087139 Chr.10: 97437224-97437224 on Build GRCh38
rs764219543 Chr.10: 97437320-97437320 on Build GRCh38
rs143980729 Chr.10: 97437251-97437251 on Build GRCh38
rs889508094 Chr.10: 97437630-97437630 on Build GRCh38
rs184496031 Chr.10: 97437606-97437606 on Build GRCh38
rs749740686 Chr.10: 97437289-97437289 on Build GRCh38
rs749685694 Chr.10: 97437186-97437186 on Build GRCh38
rs367708076 Chr.10: 97437279-97437279 on Build GRCh38
rs3763673 Chr.10: 97437435-97437435 on Build GRCh38
rs574758684 Chr.10: 97437466-97437466 on Build GRCh38
rs372708165 Chr.10: 97437406-97437406 on Build GRCh38
rs374028079 Chr.10: 97437644-97437644 on Build GRCh38
rs757772009 Chr.10: 97437546-97437546 on Build GRCh38
rs368736902 Chr.10: 97437595-97437595 on Build GRCh38
rs761743572 Chr.10: 97437322-97437322 on Build GRCh38

View Full Product Details