Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97589993-97590501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf62
Entrez Gene ID
414157
Gene Name
chromosome 10 open reading frame 62
Gene Aliases
bA548K23.1
UniGene
Hs.662302
Gene Chromosome Location
Chr.10: 97589693-97590934 on Build GRCh38
Transcripts
NM_001009997.2 NM_001134670.1 NM_138413.3

Gene Symbol
HOGA1
Entrez Gene ID
112817
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
C10orf65, DHDPS2, DHDPSL, HP3, NPL2
UniGene
Hs.180346
Gene Chromosome Location
Chr.10: 97584345-97612802 on Build GRCh38
Transcripts
NM_001009997.2 NM_001134670.1 NM_138413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6414837 c.490A>T p.N164Y Chr.10: 97590387-97590387 on Build GRCh38
6395131 c.572C>A p.A191D Chr.10: 97590469-97590469 on Build GRCh38
3808133 c.345A>T p.K115N Chr.10: 97590242-97590242 on Build GRCh38
3868223 c.353C>T p.S118F Chr.10: 97590250-97590250 on Build GRCh38
6461460 c.383C>T p.A128V Chr.10: 97590280-97590280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748010979 Chr.10: 97590285-97590285 on Build GRCh38
rs755053963 Chr.10: 97590333-97590333 on Build GRCh38
rs571127035 Chr.10: 97590097-97590097 on Build GRCh38
rs543502447 Chr.10: 97590025-97590025 on Build GRCh38
rs761616246 Chr.10: 97590173-97590173 on Build GRCh38
rs562397606 Chr.10: 97590088-97590088 on Build GRCh38
rs140956245 Chr.10: 97590026-97590026 on Build GRCh38
rs757729928 Chr.10: 97590257-97590257 on Build GRCh38
rs780159949 Chr.10: 97590329-97590329 on Build GRCh38
rs978114324 Chr.10: 97590433-97590433 on Build GRCh38
rs753260665 Chr.10: 97590133-97590133 on Build GRCh38
rs770821820 Chr.10: 97590159-97590159 on Build GRCh38
rs150921567 Chr.10: 97590317-97590317 on Build GRCh38
rs999178434 Chr.10: 97590258-97590258 on Build GRCh38
rs548435079 Chr.10: 97590379-97590379 on Build GRCh38
rs747034275 Chr.10: 97590272-97590272 on Build GRCh38
rs750889147 Chr.10: 97590394-97590394 on Build GRCh38
rs566800470 Chr.10: 97590071-97590071 on Build GRCh38
rs558944969 Chr.10: 97590093-97590093 on Build GRCh38
rs548541319 Chr.10: 97590055-97590055 on Build GRCh38
rs201791108 Chr.10: 97590132-97590132 on Build GRCh38
rs781135280 Chr.10: 97590276-97590276 on Build GRCh38
rs773513735 Chr.10: 97590023-97590023 on Build GRCh38
rs368753194 Chr.10: 97590436-97590436 on Build GRCh38
rs200831134 Chr.10: 97590070-97590070 on Build GRCh38
rs775184722 Chr.10: 97590457-97590457 on Build GRCh38
rs781188795 Chr.10: 97590335-97590335 on Build GRCh38
rs776403631 Chr.10: 97590113-97590113 on Build GRCh38
rs774441181 Chr.10: 97590381-97590381 on Build GRCh38
rs199811878 Chr.10: 97590348-97590348 on Build GRCh38
rs757009497 Chr.10: 97590443-97590443 on Build GRCh38
rs370827332 Chr.10: 97590059-97590059 on Build GRCh38
rs375555828 Chr.10: 97590361-97590361 on Build GRCh38
rs113991462 Chr.10: 97590407-97590407 on Build GRCh38
rs773530578 Chr.10: 97590118-97590118 on Build GRCh38
rs770460210 Chr.10: 97590305-97590305 on Build GRCh38
rs759613912 Chr.10: 97590032-97590032 on Build GRCh38
rs12769613 Chr.10: 97590399-97590399 on Build GRCh38
rs766696367 Chr.10: 97590402-97590402 on Build GRCh38
rs770003996 Chr.10: 97590019-97590019 on Build GRCh38
rs370594513 Chr.10: 97590158-97590158 on Build GRCh38
rs550553605 Chr.10: 97590067-97590067 on Build GRCh38
rs371888091 Chr.10: 97590308-97590308 on Build GRCh38
rs777941183 Chr.10: 97590077-97590077 on Build GRCh38
rs201569136 Chr.10: 97590144-97590144 on Build GRCh38
rs753536393 Chr.10: 97590052-97590052 on Build GRCh38
rs1012895434 Chr.10: 97590307-97590307 on Build GRCh38
rs575393946 Chr.10: 97590136-97590136 on Build GRCh38
rs556810078 Chr.10: 97590134-97590134 on Build GRCh38
rs748519578 Chr.10: 97590018-97590018 on Build GRCh38
rs768442880 Chr.10: 97590168-97590168 on Build GRCh38
rs765971580 Chr.10: 97590208-97590208 on Build GRCh38
rs759164296 Chr.10: 97590365-97590365 on Build GRCh38
rs377684189 Chr.10: 97590044-97590044 on Build GRCh38
rs756094416 Chr.10: 97590417-97590417 on Build GRCh38
rs768239290 Chr.10: 97590466-97590466 on Build GRCh38
rs761489569 Chr.10: 97590115-97590115 on Build GRCh38
rs760465262 Chr.10: 97590104-97590104 on Build GRCh38
rs777584853 Chr.10: 97590344-97590344 on Build GRCh38
rs76383275 Chr.10: 97590346-97590346 on Build GRCh38
rs747031797 Chr.10: 97590163-97590163 on Build GRCh38
rs201473199 Chr.10: 97590143-97590143 on Build GRCh38
rs754404049 Chr.10: 97590312-97590312 on Build GRCh38
rs7093840 Chr.10: 97590260-97590260 on Build GRCh38
rs367679104 Chr.10: 97590137-97590137 on Build GRCh38
rs140364165 Chr.10: 97590428-97590428 on Build GRCh38
rs767649994 Chr.10: 97590410-97590410 on Build GRCh38
rs991528323 Chr.10: 97590064-97590064 on Build GRCh38
rs765813392 Chr.10: 97590383-97590383 on Build GRCh38
rs749580947 Chr.10: 97590078-97590078 on Build GRCh38
rs376893298 Chr.10: 97590099-97590099 on Build GRCh38
rs775131314 Chr.10: 97590374-97590374 on Build GRCh38
rs762455396 Chr.10: 97590192-97590192 on Build GRCh38
rs942609041 Chr.10: 97590110-97590110 on Build GRCh38
rs565007098 Chr.10: 97590266-97590266 on Build GRCh38
rs772977970 Chr.10: 97590288-97590288 on Build GRCh38
rs998788384 Chr.10: 97590035-97590035 on Build GRCh38
rs769587708 Chr.10: 97590287-97590287 on Build GRCh38
rs371440500 Chr.10: 97590107-97590107 on Build GRCh38
rs910224589 Chr.10: 97590445-97590445 on Build GRCh38
rs755178647 Chr.10: 97590408-97590408 on Build GRCh38
rs911089990 Chr.10: 97590105-97590105 on Build GRCh38
rs144811504 Chr.10: 97590027-97590027 on Build GRCh38
rs769717721 Chr.10: 97590340-97590340 on Build GRCh38
rs763759398 Chr.10: 97590048-97590048 on Build GRCh38
rs764976061 Chr.10: 97590176-97590176 on Build GRCh38
rs142514598 Chr.10: 97590313-97590313 on Build GRCh38
rs1025401328 Chr.10: 97590074-97590074 on Build GRCh38
rs538412135 Chr.10: 97590106-97590106 on Build GRCh38
rs758539967 Chr.10: 97590261-97590261 on Build GRCh38
rs754349047 Chr.10: 97590254-97590254 on Build GRCh38
rs772854255 Chr.10: 97590183-97590183 on Build GRCh38
rs776456872 Chr.10: 97590169-97590169 on Build GRCh38
rs751742777 Chr.10: 97590406-97590406 on Build GRCh38
rs368370225 Chr.10: 97590243-97590243 on Build GRCh38
rs750719654 Chr.10: 97590325-97590325 on Build GRCh38
rs762732537 Chr.10: 97590298-97590298 on Build GRCh38
rs771849333 Chr.10: 97590454-97590454 on Build GRCh38
rs373892055 Chr.10: 97590204-97590204 on Build GRCh38
rs7093643 Chr.10: 97590371-97590371 on Build GRCh38
rs751762663 Chr.10: 97590332-97590332 on Build GRCh38
rs775259178 Chr.10: 97590354-97590354 on Build GRCh38
rs145450971 Chr.10: 97590453-97590453 on Build GRCh38
rs768730543 Chr.10: 97590273-97590273 on Build GRCh38
rs146752978 Chr.10: 97590411-97590411 on Build GRCh38
rs764677438 Chr.10: 97590248-97590248 on Build GRCh38
rs748141701 Chr.10: 97590339-97590339 on Build GRCh38
rs373701009 Chr.10: 97590042-97590042 on Build GRCh38
rs755523964 Chr.10: 97590060-97590060 on Build GRCh38
rs773907690 Chr.10: 97590359-97590359 on Build GRCh38
rs750026258 Chr.10: 97590121-97590121 on Build GRCh38
rs746536146 Chr.10: 97590459-97590459 on Build GRCh38
rs1012482351 Chr.10: 97590302-97590302 on Build GRCh38
rs148949699 Chr.10: 97590045-97590045 on Build GRCh38
rs771276698 Chr.10: 97590079-97590079 on Build GRCh38
rs758692283 Chr.10: 97590326-97590326 on Build GRCh38
rs147809962 Chr.10: 97590238-97590238 on Build GRCh38

View Full Product Details