Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99705147-99705657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX15
Entrez Gene ID
1355
Gene Name
COX15, cytochrome c oxidase assembly homolog
Gene Aliases
CEMCOX2
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99694793-99732667 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

SNP Detail

SNP ID Chromosome Location
rs909925899 Chr.10: 99705624-99705624 on Build GRCh38
rs11190250 Chr.10: 99705282-99705282 on Build GRCh38
rs879307176 Chr.10: 99705550-99705550 on Build GRCh38
rs557452909 Chr.10: 99705278-99705278 on Build GRCh38
rs543041869 Chr.10: 99705288-99705288 on Build GRCh38
rs1039278844 Chr.10: 99705427-99705427 on Build GRCh38
rs1048931275 Chr.10: 99705195-99705195 on Build GRCh38
rs575131236 Chr.10: 99705524-99705524 on Build GRCh38
rs918414120 Chr.10: 99705192-99705192 on Build GRCh38
rs955690890 Chr.10: 99705219-99705219 on Build GRCh38
rs903337859 Chr.10: 99705291-99705291 on Build GRCh38
rs1032482977 Chr.10: 99705321-99705321 on Build GRCh38
rs1052159911 Chr.10: 99705352-99705352 on Build GRCh38
rs139309659 Chr.10: 99705397-99705397 on Build GRCh38
rs774137998 Chr.10: 99705237-99705237 on Build GRCh38
rs981974552 Chr.10: 99705462-99705462 on Build GRCh38
rs970403449 Chr.10: 99705442-99705442 on Build GRCh38
rs1023248093 Chr.10: 99705423-99705423 on Build GRCh38
rs775606623 Chr.10: 99705202-99705202 on Build GRCh38
rs553307594 Chr.10: 99705216-99705216 on Build GRCh38
rs918518831 Chr.10: 99705481-99705481 on Build GRCh38
rs943589049 Chr.10: 99705416-99705416 on Build GRCh38
rs904712148 Chr.10: 99705178-99705178 on Build GRCh38
rs929321491 Chr.10: 99705325-99705325 on Build GRCh38
rs764304984 Chr.10: 99705220-99705220 on Build GRCh38
rs753464136 Chr.10: 99705459-99705461 on Build GRCh38
rs970347396 Chr.10: 99705254-99705254 on Build GRCh38
rs578102765 Chr.10: 99705230-99705230 on Build GRCh38
rs114548202 Chr.10: 99705186-99705186 on Build GRCh38
rs185423638 Chr.10: 99705183-99705183 on Build GRCh38
rs545523868 Chr.10: 99705238-99705238 on Build GRCh38
rs35229927 Chr.10: 99705227-99705228 on Build GRCh38
rs941200071 Chr.10: 99705296-99705296 on Build GRCh38
rs983975011 Chr.10: 99705574-99705574 on Build GRCh38
rs942658569 Chr.10: 99705234-99705234 on Build GRCh38
rs957813458 Chr.10: 99705334-99705334 on Build GRCh38
rs929847339 Chr.10: 99705193-99705193 on Build GRCh38
rs1000382040 Chr.10: 99705317-99705317 on Build GRCh38
rs949309968 Chr.10: 99705173-99705173 on Build GRCh38
rs763158246 Chr.10: 99705203-99705203 on Build GRCh38
rs919375258 Chr.10: 99705319-99705319 on Build GRCh38
rs200161344 Chr.10: 99705437-99705437 on Build GRCh38

View Full Product Details