Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99707802-99708351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX15
Entrez Gene ID
1355
Gene Name
COX15, cytochrome c oxidase assembly homolog
Gene Aliases
CEMCOX2
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99694793-99732667 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

Gene Symbol
ENTPD7
Entrez Gene ID
57089
Gene Name
ectonucleoside triphosphate diphosphohydrolase 7
Gene Aliases
LALP1
UniGene
Hs.28326
Gene Chromosome Location
Chr.10: 99659506-99711245 on Build GRCh38
Transcripts
NM_001320974.1 XM_006717634.3 XM_017015715.1 NM_020354.3

SNP Detail

SNP ID Chromosome Location
rs989105762 Chr.10: 99708298-99708298 on Build GRCh38
rs207471307 Chr.10: 99708152-99708152 on Build GRCh38
rs1028035121 Chr.10: 99708165-99708165 on Build GRCh38
rs958902754 Chr.10: 99708143-99708143 on Build GRCh38
rs1010297661 Chr.10: 99707866-99707866 on Build GRCh38
rs1035646100 Chr.10: 99707951-99707951 on Build GRCh38
rs750792416 Chr.10: 99708181-99708181 on Build GRCh38
rs886480115 Chr.10: 99707969-99707969 on Build GRCh38
rs548739978 Chr.10: 99708275-99708275 on Build GRCh38
rs141799782 Chr.10: 99708318-99708318 on Build GRCh38
rs180982477 Chr.10: 99707887-99707887 on Build GRCh38
rs557887408 Chr.10: 99708193-99708193 on Build GRCh38
rs566885209 Chr.10: 99708294-99708294 on Build GRCh38
rs1003319452 Chr.10: 99707979-99707979 on Build GRCh38
rs951647688 Chr.10: 99708259-99708259 on Build GRCh38
rs756556486 Chr.10: 99708217-99708217 on Build GRCh38
rs796569875 Chr.10: 99707877-99707877 on Build GRCh38
rs905961443 Chr.10: 99707831-99707831 on Build GRCh38
rs532095456 Chr.10: 99708008-99708008 on Build GRCh38
rs147917263 Chr.10: 99708083-99708083 on Build GRCh38
rs892313953 Chr.10: 99707829-99707829 on Build GRCh38
rs947183944 Chr.10: 99708326-99708326 on Build GRCh38
rs953782354 Chr.10: 99708188-99708188 on Build GRCh38
rs974923532 Chr.10: 99708161-99708161 on Build GRCh38
rs912198716 Chr.10: 99708196-99708196 on Build GRCh38
rs564893685 Chr.10: 99707914-99707914 on Build GRCh38
rs1005526724 Chr.10: 99707976-99707976 on Build GRCh38
rs544927520 Chr.10: 99708039-99708039 on Build GRCh38
rs1002796420 Chr.10: 99707864-99707864 on Build GRCh38
rs907188880 Chr.10: 99707955-99707955 on Build GRCh38
rs780560892 Chr.10: 99708268-99708268 on Build GRCh38
rs572881890 Chr.10: 99707836-99707836 on Build GRCh38

View Full Product Details