Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99883913-99884409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMBP
Entrez Gene ID
23268
Gene Name
dynamin binding protein
Gene Aliases
ARHGEF36, TUBA
UniGene
Hs.500771
Gene Chromosome Location
Chr.10: 99875571-100009953 on Build GRCh38
Transcripts
NM_001318326.1 NM_001318327.1 NM_015221.3 XM_006717735.2 XM_011539559.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3433476 c.3926C>T p.P1309L Chr.10: 99884154-99884154 on Build GRCh38
4815555 c.4026A>C p.K1342N Chr.10: 99884054-99884054 on Build GRCh38
3433478 c.3908C>T p.S1303F Chr.10: 99884172-99884172 on Build GRCh38
3433474 c.3962A>G p.N1321S Chr.10: 99884118-99884118 on Build GRCh38
6045501 c.3894T>C p.A1298A Chr.10: 99884114-99884114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752318460 Chr.10: 99884154-99884154 on Build GRCh38
rs757689446 Chr.10: 99884016-99884016 on Build GRCh38
rs763249912 Chr.10: 99883994-99883994 on Build GRCh38
rs761260077 Chr.10: 99884251-99884251 on Build GRCh38
rs772510161 Chr.10: 99884239-99884239 on Build GRCh38
rs749432511 Chr.10: 99884211-99884211 on Build GRCh38
rs1004247800 Chr.10: 99884279-99884279 on Build GRCh38
rs952683143 Chr.10: 99884192-99884192 on Build GRCh38
rs770969792 Chr.10: 99883964-99883964 on Build GRCh38
rs764295904 Chr.10: 99884012-99884012 on Build GRCh38
rs376611193 Chr.10: 99884031-99884031 on Build GRCh38
rs774596635 Chr.10: 99884224-99884224 on Build GRCh38
rs373446562 Chr.10: 99883981-99883981 on Build GRCh38
rs974248899 Chr.10: 99883985-99883985 on Build GRCh38
rs533025422 Chr.10: 99883946-99883946 on Build GRCh38
rs376842527 Chr.10: 99884314-99884317 on Build GRCh38
rs200888442 Chr.10: 99883986-99883986 on Build GRCh38
rs769805859 Chr.10: 99884101-99884101 on Build GRCh38
rs866750696 Chr.10: 99884152-99884152 on Build GRCh38
rs944404890 Chr.10: 99884110-99884110 on Build GRCh38
rs781065611 Chr.10: 99884073-99884073 on Build GRCh38
rs954223156 Chr.10: 99884378-99884378 on Build GRCh38
rs200569120 Chr.10: 99884193-99884193 on Build GRCh38
rs1016255238 Chr.10: 99884291-99884291 on Build GRCh38
rs753371257 Chr.10: 99884028-99884028 on Build GRCh38
rs752023993 Chr.10: 99884015-99884015 on Build GRCh38
rs199990030 Chr.10: 99884060-99884060 on Build GRCh38
rs562309760 Chr.10: 99884112-99884112 on Build GRCh38
rs777426720 Chr.10: 99884058-99884058 on Build GRCh38
rs767050958 Chr.10: 99884183-99884185 on Build GRCh38
rs116046885 Chr.10: 99884118-99884118 on Build GRCh38
rs139354128 Chr.10: 99884168-99884168 on Build GRCh38
rs377597240 Chr.10: 99884128-99884128 on Build GRCh38
rs986406401 Chr.10: 99884313-99884313 on Build GRCh38
rs768115422 Chr.10: 99884023-99884023 on Build GRCh38
rs923302663 Chr.10: 99884302-99884302 on Build GRCh38
rs531400512 Chr.10: 99883967-99883967 on Build GRCh38
rs1028413178 Chr.10: 99884365-99884365 on Build GRCh38
rs138512003 Chr.10: 99884032-99884032 on Build GRCh38
rs146259512 Chr.10: 99884134-99884134 on Build GRCh38
rs758075996 Chr.10: 99884042-99884042 on Build GRCh38
rs373053661 Chr.10: 99884094-99884094 on Build GRCh38
rs147539253 Chr.10: 99884308-99884311 on Build GRCh38
rs551198081 Chr.10: 99883976-99883976 on Build GRCh38
rs1015821363 Chr.10: 99884270-99884270 on Build GRCh38
rs745738512 Chr.10: 99884179-99884179 on Build GRCh38
rs761280830 Chr.10: 99884138-99884138 on Build GRCh38
rs370997978 Chr.10: 99884223-99884223 on Build GRCh38
rs749911833 Chr.10: 99884255-99884256 on Build GRCh38
rs751569037 Chr.10: 99883979-99883980 on Build GRCh38
rs953090797 Chr.10: 99884046-99884046 on Build GRCh38
rs770740557 Chr.10: 99884068-99884068 on Build GRCh38
rs547356672 Chr.10: 99884231-99884231 on Build GRCh38
rs962969212 Chr.10: 99884305-99884305 on Build GRCh38
rs757135742 Chr.10: 99884107-99884108 on Build GRCh38
rs995715695 Chr.10: 99884327-99884327 on Build GRCh38
rs896461478 Chr.10: 99884278-99884278 on Build GRCh38
rs773720873 Chr.10: 99884250-99884250 on Build GRCh38
rs745828314 Chr.10: 99884093-99884093 on Build GRCh38
rs534628616 Chr.10: 99884153-99884153 on Build GRCh38
rs750916384 Chr.10: 99884137-99884137 on Build GRCh38
rs768726918 Chr.10: 99884216-99884216 on Build GRCh38
rs747749677 Chr.10: 99884038-99884038 on Build GRCh38
rs985818626 Chr.10: 99884384-99884384 on Build GRCh38
rs940744450 Chr.10: 99883941-99883941 on Build GRCh38
rs1015871760 Chr.10: 99883957-99883957 on Build GRCh38
rs373600848 Chr.10: 99884201-99884201 on Build GRCh38
rs780977934 Chr.10: 99884178-99884178 on Build GRCh38
rs767051488 Chr.10: 99884257-99884257 on Build GRCh38
rs370574450 Chr.10: 99884176-99884176 on Build GRCh38
rs567734686 Chr.10: 99884104-99884104 on Build GRCh38
rs112408258 Chr.10: 99883966-99883967 on Build GRCh38
rs777532425 Chr.10: 99884167-99884167 on Build GRCh38
rs757975644 Chr.10: 99884162-99884162 on Build GRCh38
rs963193047 Chr.10: 99883983-99883983 on Build GRCh38
rs763077330 Chr.10: 99884115-99884115 on Build GRCh38
rs776681167 Chr.10: 99883969-99883969 on Build GRCh38

View Full Product Details