Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100075777-100076297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPN1
Entrez Gene ID
1369
Gene Name
carboxypeptidase N subunit 1
Gene Aliases
CPN, SCPN
UniGene
Hs.2246
Gene Chromosome Location
Chr.10: 100042308-100081885 on Build GRCh38
Transcripts
NM_001308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011007 c.393C>A p.P131P Chr.10: 100075938-100075938 on Build GRCh38
273635 c.338G>A p.R113H Chr.10: 100075993-100075993 on Build GRCh38
6400554 c.379C>T p.P127S Chr.10: 100075952-100075952 on Build GRCh38
41006 c.363G>A p.T121T Chr.10: 100075968-100075968 on Build GRCh38
6406719 c.303G>T p.E101D Chr.10: 100076028-100076028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940408104 Chr.10: 100075994-100075994 on Build GRCh38
rs371676125 Chr.10: 100076128-100076128 on Build GRCh38
rs374761550 Chr.10: 100075961-100075961 on Build GRCh38
rs573175112 Chr.10: 100075867-100075867 on Build GRCh38
rs752074540 Chr.10: 100075911-100075911 on Build GRCh38
rs778548653 Chr.10: 100075941-100075941 on Build GRCh38
rs771919204 Chr.10: 100075950-100075950 on Build GRCh38
rs890289859 Chr.10: 100075820-100075820 on Build GRCh38
rs779479935 Chr.10: 100075928-100075928 on Build GRCh38
rs74565668 Chr.10: 100076243-100076243 on Build GRCh38
rs901216241 Chr.10: 100075936-100075936 on Build GRCh38
rs764687495 Chr.10: 100075888-100075888 on Build GRCh38
rs761119505 Chr.10: 100076156-100076156 on Build GRCh38
rs185903225 Chr.10: 100076273-100076273 on Build GRCh38
rs748852657 Chr.10: 100075929-100075929 on Build GRCh38
rs959375559 Chr.10: 100076033-100076033 on Build GRCh38
rs781206816 Chr.10: 100076019-100076019 on Build GRCh38
rs752085352 Chr.10: 100075984-100075984 on Build GRCh38
rs368820009 Chr.10: 100076027-100076027 on Build GRCh38
rs901210916 Chr.10: 100076134-100076134 on Build GRCh38
rs758849544 Chr.10: 100075887-100075887 on Build GRCh38
rs775824980 Chr.10: 100076032-100076032 on Build GRCh38
rs780989150 Chr.10: 100076118-100076118 on Build GRCh38
rs754558434 Chr.10: 100075937-100075937 on Build GRCh38
rs540311121 Chr.10: 100075967-100075967 on Build GRCh38
rs752224544 Chr.10: 100076066-100076066 on Build GRCh38
rs772524811 Chr.10: 100076154-100076154 on Build GRCh38
rs61754515 Chr.10: 100076007-100076007 on Build GRCh38
rs548844812 Chr.10: 100076197-100076197 on Build GRCh38
rs75664608 Chr.10: 100076246-100076246 on Build GRCh38
rs780234239 Chr.10: 100076131-100076131 on Build GRCh38
rs768694390 Chr.10: 100076142-100076142 on Build GRCh38
rs551887166 Chr.10: 100076272-100076272 on Build GRCh38
rs181050548 Chr.10: 100076195-100076195 on Build GRCh38
rs76156167 Chr.10: 100076029-100076029 on Build GRCh38
rs770920497 Chr.10: 100075966-100075966 on Build GRCh38
rs770069005 Chr.10: 100076031-100076031 on Build GRCh38
rs763408059 Chr.10: 100075969-100075969 on Build GRCh38
rs969788040 Chr.10: 100076163-100076163 on Build GRCh38
rs140546981 Chr.10: 100076052-100076052 on Build GRCh38
rs768945263 Chr.10: 100076048-100076048 on Build GRCh38
rs778531931 Chr.10: 100076008-100076008 on Build GRCh38
rs775743949 Chr.10: 100075934-100075934 on Build GRCh38
rs981429776 Chr.10: 100075958-100075958 on Build GRCh38
rs1052243513 Chr.10: 100076077-100076077 on Build GRCh38
rs370836779 Chr.10: 100075922-100075922 on Build GRCh38
rs367619099 Chr.10: 100076111-100076111 on Build GRCh38
rs141105015 Chr.10: 100076073-100076073 on Build GRCh38
rs12241340 Chr.10: 100075982-100075982 on Build GRCh38
rs777379413 Chr.10: 100076011-100076011 on Build GRCh38
rs377653573 Chr.10: 100075876-100075876 on Build GRCh38
rs145626659 Chr.10: 100076054-100076054 on Build GRCh38
rs759780238 Chr.10: 100075860-100075860 on Build GRCh38
rs908704485 Chr.10: 100076263-100076263 on Build GRCh38
rs1031097637 Chr.10: 100075885-100075885 on Build GRCh38
rs553809968 Chr.10: 100075854-100075854 on Build GRCh38
rs374870878 Chr.10: 100076051-100076051 on Build GRCh38
rs868177651 Chr.10: 100076028-100076028 on Build GRCh38
rs746847569 Chr.10: 100076015-100076015 on Build GRCh38
rs768101001 Chr.10: 100075989-100075989 on Build GRCh38
rs763109927 Chr.10: 100075975-100075975 on Build GRCh38
rs755620359 Chr.10: 100075913-100075913 on Build GRCh38
rs115166271 Chr.10: 100075968-100075968 on Build GRCh38
rs138185881 Chr.10: 100076062-100076062 on Build GRCh38
rs878952186 Chr.10: 100076161-100076161 on Build GRCh38
rs374239992 Chr.10: 100076093-100076093 on Build GRCh38
rs770703494 Chr.10: 100076018-100076018 on Build GRCh38
rs1035238318 Chr.10: 100076222-100076222 on Build GRCh38
rs188667294 Chr.10: 100076155-100076155 on Build GRCh38
rs762079403 Chr.10: 100076143-100076143 on Build GRCh38
rs769973756 Chr.10: 100076129-100076129 on Build GRCh38
rs773167602 Chr.10: 100075959-100075959 on Build GRCh38
rs747924477 Chr.10: 100075947-100075947 on Build GRCh38
rs757656853 Chr.10: 100075988-100075988 on Build GRCh38
rs749411041 Chr.10: 100076133-100076133 on Build GRCh38
rs1011394805 Chr.10: 100076149-100076149 on Build GRCh38
rs1037401752 Chr.10: 100075997-100075997 on Build GRCh38
rs759014388 Chr.10: 100076061-100076061 on Build GRCh38
rs892564693 Chr.10: 100076101-100076101 on Build GRCh38
rs763317686 Chr.10: 100076045-100076045 on Build GRCh38
rs764408494 Chr.10: 100075976-100075976 on Build GRCh38
rs757944645 Chr.10: 100076067-100076067 on Build GRCh38
rs146812654 Chr.10: 100076009-100076009 on Build GRCh38
rs767897122 Chr.10: 100076053-100076053 on Build GRCh38

View Full Product Details