Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100081321-100081827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPN1
Entrez Gene ID
1369
Gene Name
carboxypeptidase N subunit 1
Gene Aliases
CPN, SCPN
UniGene
Hs.2246
Gene Chromosome Location
Chr.10: 100042308-100081885 on Build GRCh38
Transcripts
NM_001308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3433511 c.98G>A p.R33Q Chr.10: 100081528-100081528 on Build GRCh38
23428 c.141G>A p.R47R Chr.10: 100081485-100081485 on Build GRCh38
5721571 c.42C>T p.L14L Chr.10: 100081584-100081584 on Build GRCh38
4011008 c.157C>T p.R53C Chr.10: 100081469-100081469 on Build GRCh38
5612656 c.199G>A p.D67N Chr.10: 100081427-100081427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745729622 Chr.10: 100081792-100081792 on Build GRCh38
rs372205929 Chr.10: 100081410-100081410 on Build GRCh38
rs1052830748 Chr.10: 100081456-100081456 on Build GRCh38
rs200987064 Chr.10: 100081455-100081455 on Build GRCh38
rs189044474 Chr.10: 100081379-100081379 on Build GRCh38
rs762992078 Chr.10: 100081658-100081658 on Build GRCh38
rs373670285 Chr.10: 100081362-100081362 on Build GRCh38
rs1002721852 Chr.10: 100081482-100081482 on Build GRCh38
rs955117066 Chr.10: 100081632-100081632 on Build GRCh38
rs890909438 Chr.10: 100081443-100081443 on Build GRCh38
rs958792180 Chr.10: 100081796-100081796 on Build GRCh38
rs893376063 Chr.10: 100081680-100081680 on Build GRCh38
rs111400589 Chr.10: 100081612-100081612 on Build GRCh38
rs746251575 Chr.10: 100081571-100081571 on Build GRCh38
rs755151996 Chr.10: 100081407-100081407 on Build GRCh38
rs934389254 Chr.10: 100081449-100081449 on Build GRCh38
rs1052808553 Chr.10: 100081701-100081701 on Build GRCh38
rs752640687 Chr.10: 100081652-100081652 on Build GRCh38
rs770270851 Chr.10: 100081572-100081572 on Build GRCh38
rs773064523 Chr.10: 100081536-100081536 on Build GRCh38
rs568940128 Chr.10: 100081671-100081671 on Build GRCh38
rs758527116 Chr.10: 100081555-100081555 on Build GRCh38
rs781573757 Chr.10: 100081563-100081563 on Build GRCh38
rs922192921 Chr.10: 100081616-100081616 on Build GRCh38
rs914281785 Chr.10: 100081716-100081716 on Build GRCh38
rs61733668 Chr.10: 100081475-100081475 on Build GRCh38
rs899596582 Chr.10: 100081633-100081633 on Build GRCh38
rs10883439 Chr.10: 100081396-100081396 on Build GRCh38
rs751664367 Chr.10: 100081667-100081667 on Build GRCh38
rs772203596 Chr.10: 100081440-100081440 on Build GRCh38
rs757536670 Chr.10: 100081592-100081592 on Build GRCh38
rs549281938 Chr.10: 100081512-100081512 on Build GRCh38
rs766323805 Chr.10: 100081541-100081541 on Build GRCh38
rs1010521425 Chr.10: 100081688-100081688 on Build GRCh38
rs370527794 Chr.10: 100081567-100081567 on Build GRCh38
rs781275676 Chr.10: 100081675-100081675 on Build GRCh38
rs553176799 Chr.10: 100081580-100081580 on Build GRCh38
rs765273218 Chr.10: 100081567-100081567 on Build GRCh38
rs751706282 Chr.10: 100081525-100081525 on Build GRCh38
rs909193327 Chr.10: 100081384-100081384 on Build GRCh38
rs771747138 Chr.10: 100081584-100081584 on Build GRCh38
rs748205584 Chr.10: 100081499-100081499 on Build GRCh38
rs933494733 Chr.10: 100081422-100081422 on Build GRCh38
rs573116144 Chr.10: 100081747-100081747 on Build GRCh38
rs368682241 Chr.10: 100081467-100081467 on Build GRCh38
rs771355307 Chr.10: 100081515-100081515 on Build GRCh38
rs747648440 Chr.10: 100081581-100081581 on Build GRCh38
rs750176724 Chr.10: 100081481-100081481 on Build GRCh38
rs776976696 Chr.10: 100081516-100081516 on Build GRCh38
rs757598430 Chr.10: 100081560-100081560 on Build GRCh38
rs781183240 Chr.10: 100081394-100081394 on Build GRCh38
rs776417344 Chr.10: 100081608-100081608 on Build GRCh38
rs752752245 Chr.10: 100081675-100081678 on Build GRCh38
rs200920833 Chr.10: 100081640-100081640 on Build GRCh38
rs934463565 Chr.10: 100081694-100081694 on Build GRCh38
rs763644870 Chr.10: 100081372-100081372 on Build GRCh38
rs538591971 Chr.10: 100081619-100081619 on Build GRCh38
rs562025469 Chr.10: 100081578-100081578 on Build GRCh38
rs760354309 Chr.10: 100081602-100081602 on Build GRCh38
rs149401779 Chr.10: 100081487-100081487 on Build GRCh38
rs1002848938 Chr.10: 100081769-100081769 on Build GRCh38
rs567825978 Chr.10: 100081529-100081529 on Build GRCh38
rs754901371 Chr.10: 100081549-100081549 on Build GRCh38
rs765087377 Chr.10: 100081551-100081551 on Build GRCh38
rs758431094 Chr.10: 100081473-100081473 on Build GRCh38
rs758547188 Chr.10: 100081500-100081500 on Build GRCh38
rs368462704 Chr.10: 100081377-100081377 on Build GRCh38
rs376709188 Chr.10: 100081370-100081370 on Build GRCh38
rs770737401 Chr.10: 100081605-100081605 on Build GRCh38
rs753854211 Chr.10: 100081489-100081489 on Build GRCh38
rs748449996 Chr.10: 100081421-100081421 on Build GRCh38
rs370027071 Chr.10: 100081405-100081405 on Build GRCh38
rs760515524 Chr.10: 100081538-100081538 on Build GRCh38
rs866703720 Chr.10: 100081495-100081495 on Build GRCh38
rs751823708 Chr.10: 100081556-100081556 on Build GRCh38
rs755504443 Chr.10: 100081484-100081504 on Build GRCh38
rs1044009378 Chr.10: 100081786-100081786 on Build GRCh38
rs777184310 Chr.10: 100081457-100081457 on Build GRCh38
rs766413889 Chr.10: 100081488-100081488 on Build GRCh38
rs779001062 Chr.10: 100081497-100081497 on Build GRCh38
rs777979903 Chr.10: 100081511-100081511 on Build GRCh38
rs150259958 Chr.10: 100081403-100081403 on Build GRCh38
rs764210605 Chr.10: 100081659-100081659 on Build GRCh38
rs200035303 Chr.10: 100081533-100081533 on Build GRCh38
rs201817147 Chr.10: 100081546-100081546 on Build GRCh38
rs766899820 Chr.10: 100081358-100081358 on Build GRCh38
rs1039448436 Chr.10: 100081537-100081537 on Build GRCh38
rs754360549 Chr.10: 100081439-100081440 on Build GRCh38
rs1052336812 Chr.10: 100081439-100081439 on Build GRCh38
rs375887047 Chr.10: 100081451-100081451 on Build GRCh38
rs761479872 Chr.10: 100081469-100081469 on Build GRCh38
rs754989179 Chr.10: 100081496-100081496 on Build GRCh38
rs950831152 Chr.10: 100081363-100081363 on Build GRCh38
rs373785594 Chr.10: 100081553-100081553 on Build GRCh38
rs756075767 Chr.10: 100081402-100081402 on Build GRCh38
rs746438789 Chr.10: 100081523-100081523 on Build GRCh38

View Full Product Details