Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100150751-100151263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERLIN1
Entrez Gene ID
10613
Gene Name
ER lipid raft associated 1
Gene Aliases
C10orf69, Erlin-1, KE04, KEO4, SPFH1, SPG62
UniGene
Hs.150087
Gene Chromosome Location
Chr.10: 100150090-100186057 on Build GRCh38
Transcripts
NM_001100626.1 NM_006459.3 XM_005269442.3 XM_017015541.1

SNP Detail

SNP ID Chromosome Location
rs966010524 Chr.10: 100150898-100150898 on Build GRCh38
rs1036572472 Chr.10: 100151229-100151229 on Build GRCh38
rs1049659852 Chr.10: 100150791-100150791 on Build GRCh38
rs930412547 Chr.10: 100151199-100151199 on Build GRCh38
rs931027206 Chr.10: 100151132-100151132 on Build GRCh38
rs938325001 Chr.10: 100151070-100151070 on Build GRCh38
rs575780178 Chr.10: 100150938-100150938 on Build GRCh38
rs971018547 Chr.10: 100151104-100151104 on Build GRCh38
rs977321324 Chr.10: 100151045-100151045 on Build GRCh38
rs1000007649 Chr.10: 100151016-100151016 on Build GRCh38
rs561215465 Chr.10: 100150953-100150953 on Build GRCh38
rs1021039504 Chr.10: 100151009-100151009 on Build GRCh38
rs1040740453 Chr.10: 100150944-100150944 on Build GRCh38
rs1024682704 Chr.10: 100151093-100151093 on Build GRCh38
rs901832921 Chr.10: 100150968-100150968 on Build GRCh38
rs918977321 Chr.10: 100151123-100151123 on Build GRCh38
rs543305727 Chr.10: 100150781-100150781 on Build GRCh38
rs573202250 Chr.10: 100150981-100150981 on Build GRCh38
rs545217029 Chr.10: 100150789-100150789 on Build GRCh38
rs916923769 Chr.10: 100150790-100150790 on Build GRCh38
rs991414520 Chr.10: 100151085-100151085 on Build GRCh38
rs910726626 Chr.10: 100151225-100151225 on Build GRCh38
rs557507680 Chr.10: 100150854-100150854 on Build GRCh38
rs967259639 Chr.10: 100151019-100151019 on Build GRCh38
rs539835716 Chr.10: 100151086-100151086 on Build GRCh38
rs943644281 Chr.10: 100150942-100150942 on Build GRCh38
rs943566167 Chr.10: 100151230-100151230 on Build GRCh38
rs757396134 Chr.10: 100150895-100150895 on Build GRCh38
rs1005528317 Chr.10: 100150833-100150833 on Build GRCh38
rs930795270 Chr.10: 100150915-100150915 on Build GRCh38
rs1032849686 Chr.10: 100151021-100151021 on Build GRCh38
rs903953612 Chr.10: 100151010-100151010 on Build GRCh38
rs545934477 Chr.10: 100151182-100151182 on Build GRCh38
rs542827361 Chr.10: 100150941-100150941 on Build GRCh38
rs971732283 Chr.10: 100151117-100151117 on Build GRCh38
rs887930130 Chr.10: 100151148-100151148 on Build GRCh38
rs564778511 Chr.10: 100151134-100151134 on Build GRCh38
rs752052060 Chr.10: 100150886-100150886 on Build GRCh38
rs985323930 Chr.10: 100151222-100151222 on Build GRCh38
rs554166425 Chr.10: 100150826-100150826 on Build GRCh38
rs958661400 Chr.10: 100150782-100150782 on Build GRCh38
rs902066098 Chr.10: 100151238-100151238 on Build GRCh38
rs764626849 Chr.10: 100150836-100150836 on Build GRCh38
rs928568423 Chr.10: 100151062-100151062 on Build GRCh38
rs577154069 Chr.10: 100151084-100151084 on Build GRCh38
rs781487044 Chr.10: 100150998-100150998 on Build GRCh38
rs919037718 Chr.10: 100150903-100150903 on Build GRCh38
rs887970635 Chr.10: 100150809-100150809 on Build GRCh38
rs1048166995 Chr.10: 100151135-100151135 on Build GRCh38

View Full Product Details