Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101694259-101694764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW4
Entrez Gene ID
6468
Gene Name
F-box and WD repeat domain containing 4
Gene Aliases
DAC, FBW4, FBWD4, SHFM3, SHSF3
UniGene
Hs.500822
Gene Chromosome Location
Chr.10: 101610664-101695295 on Build GRCh38
Transcripts
NM_001323541.1 NM_022039.3 NR_136613.1 XM_005270053.2 XM_017016570.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861778 c.245G>A p.R82Q Chr.10: 101694396-101694396 on Build GRCh38
6209300 c.187A>G p.S63G Chr.10: 101694454-101694454 on Build GRCh38
6029473 c.232T>A p.S78T Chr.10: 101694409-101694409 on Build GRCh38
6430196 c.7G>C p.A3P Chr.10: 101694634-101694634 on Build GRCh38
6391939 c.142C>A p.L48I Chr.10: 101694499-101694499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117664315 Chr.10: 101694666-101694666 on Build GRCh38
rs749970497 Chr.10: 101694517-101694517 on Build GRCh38
rs982766630 Chr.10: 101694692-101694692 on Build GRCh38
rs899623870 Chr.10: 101694543-101694543 on Build GRCh38
rs564717447 Chr.10: 101694682-101694682 on Build GRCh38
rs769235550 Chr.10: 101694642-101694642 on Build GRCh38
rs765859592 Chr.10: 101694654-101694654 on Build GRCh38
rs768227529 Chr.10: 101694596-101694597 on Build GRCh38
rs886046644 Chr.10: 101694669-101694669 on Build GRCh38
rs554605788 Chr.10: 101694397-101694397 on Build GRCh38
rs989947842 Chr.10: 101694363-101694363 on Build GRCh38
rs932393555 Chr.10: 101694445-101694445 on Build GRCh38
rs908840207 Chr.10: 101694734-101694734 on Build GRCh38
rs560966094 Chr.10: 101694600-101694601 on Build GRCh38
rs996537957 Chr.10: 101694544-101694544 on Build GRCh38
rs545689445 Chr.10: 101694521-101694521 on Build GRCh38
rs779631751 Chr.10: 101694421-101694421 on Build GRCh38
rs763485940 Chr.10: 101694512-101694512 on Build GRCh38
rs866337627 Chr.10: 101694492-101694492 on Build GRCh38
rs761288374 Chr.10: 101694399-101694399 on Build GRCh38
rs886046642 Chr.10: 101694396-101694396 on Build GRCh38
rs886046645 Chr.10: 101694697-101694697 on Build GRCh38
rs1032014243 Chr.10: 101694587-101694587 on Build GRCh38
rs780365225 Chr.10: 101694601-101694603 on Build GRCh38
rs61382490 Chr.10: 101694645-101694645 on Build GRCh38
rs746085418 Chr.10: 101694622-101694622 on Build GRCh38
rs866579785 Chr.10: 101694506-101694506 on Build GRCh38
rs747039293 Chr.10: 101694441-101694441 on Build GRCh38
rs1038498449 Chr.10: 101694541-101694541 on Build GRCh38
rs553281120 Chr.10: 101694431-101694431 on Build GRCh38
rs578233191 Chr.10: 101694455-101694455 on Build GRCh38
rs535857775 Chr.10: 101694336-101694336 on Build GRCh38
rs983120646 Chr.10: 101694322-101694322 on Build GRCh38
rs778726647 Chr.10: 101694427-101694427 on Build GRCh38
rs781004014 Chr.10: 101694601-101694606 on Build GRCh38
rs957203741 Chr.10: 101694359-101694359 on Build GRCh38
rs777535899 Chr.10: 101694595-101694595 on Build GRCh38
rs759169990 Chr.10: 101694665-101694665 on Build GRCh38
rs771078614 Chr.10: 101694600-101694600 on Build GRCh38
rs867123934 Chr.10: 101694435-101694435 on Build GRCh38
rs886046643 Chr.10: 101694613-101694613 on Build GRCh38
rs758387288 Chr.10: 101694585-101694585 on Build GRCh38
rs765845910 Chr.10: 101694395-101694395 on Build GRCh38
rs752571607 Chr.10: 101694670-101694670 on Build GRCh38
rs939326551 Chr.10: 101694582-101694582 on Build GRCh38
rs759941552 Chr.10: 101694391-101694391 on Build GRCh38
rs775901446 Chr.10: 101694635-101694635 on Build GRCh38
rs754699416 Chr.10: 101694426-101694426 on Build GRCh38
rs765140285 Chr.10: 101694447-101694448 on Build GRCh38
rs753427358 Chr.10: 101694567-101694567 on Build GRCh38
rs754822071 Chr.10: 101694569-101694569 on Build GRCh38
rs61761937 Chr.10: 101694572-101694572 on Build GRCh38
rs575669609 Chr.10: 101694603-101694603 on Build GRCh38
rs775928431 Chr.10: 101694509-101694509 on Build GRCh38
rs538515803 Chr.10: 101694433-101694433 on Build GRCh38
rs751266824 Chr.10: 101694637-101694637 on Build GRCh38
rs533934270 Chr.10: 101694429-101694429 on Build GRCh38
rs759994322 Chr.10: 101694466-101694466 on Build GRCh38
rs777889376 Chr.10: 101694440-101694440 on Build GRCh38
rs116070021 Chr.10: 101694649-101694649 on Build GRCh38
rs906822840 Chr.10: 101694578-101694578 on Build GRCh38
rs1015495232 Chr.10: 101694343-101694343 on Build GRCh38
rs747092369 Chr.10: 101694597-101694597 on Build GRCh38
rs777161038 Chr.10: 101694382-101694382 on Build GRCh38
rs750083482 Chr.10: 101694410-101694410 on Build GRCh38
rs920977314 Chr.10: 101694430-101694430 on Build GRCh38
rs770271071 Chr.10: 101694497-101694497 on Build GRCh38
rs964998218 Chr.10: 101694364-101694364 on Build GRCh38
rs752630394 Chr.10: 101694576-101694576 on Build GRCh38
rs572854136 Chr.10: 101694425-101694425 on Build GRCh38
rs766021845 Chr.10: 101694547-101694547 on Build GRCh38
rs766984798 Chr.10: 101694515-101694515 on Build GRCh38
rs981283734 Chr.10: 101694661-101694661 on Build GRCh38
rs952638644 Chr.10: 101694299-101694299 on Build GRCh38
rs767049873 Chr.10: 101694405-101694405 on Build GRCh38
rs766902317 Chr.10: 101694340-101694340 on Build GRCh38
rs776928234 Chr.10: 101694462-101694462 on Build GRCh38
rs772089125 Chr.10: 101694436-101694436 on Build GRCh38
rs111600818 Chr.10: 101694634-101694634 on Build GRCh38
rs886046646 Chr.10: 101694732-101694732 on Build GRCh38
rs755740500 Chr.10: 101694413-101694413 on Build GRCh38
rs1047776825 Chr.10: 101694493-101694493 on Build GRCh38
rs371115156 Chr.10: 101694294-101694294 on Build GRCh38
rs776329149 Chr.10: 101694655-101694655 on Build GRCh38

View Full Product Details