Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102358417-102358912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBF1
Entrez Gene ID
8729
Gene Name
golgi brefeldin A resistant guanine nucleotide exchange factor 1
Gene Aliases
ARF1GEF
UniGene
Hs.290243
Gene Chromosome Location
Chr.10: 102245498-102382899 on Build GRCh38
Transcripts
NM_001199378.1 NM_001199379.1 NM_004193.2 XM_005270261.1 XM_006718047.1 XM_006718048.1 XM_006718049.1 XM_006718050.1 XM_011540312.1 XM_011540313.1 XM_017016861.1 XM_017016862.1 XM_017016863.1 XM_017016864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5810073 c.831A>T p.A277A Chr.10: 102358549-102358549 on Build GRCh38
4572859 c.887T>C p.F296S Chr.10: 102358605-102358605 on Build GRCh38
2059012 c.1011+6G>A p.? Chr.10: 102358735-102358735 on Build GRCh38
6669760 c.810G>A p.V270V Chr.10: 102358528-102358528 on Build GRCh38
4943762 c.1011+3A>G p.? Chr.10: 102358732-102358732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76633616 Chr.10: 102358877-102358877 on Build GRCh38
rs771792890 Chr.10: 102358466-102358469 on Build GRCh38
rs770326659 Chr.10: 102358494-102358494 on Build GRCh38
rs759928680 Chr.10: 102358485-102358485 on Build GRCh38
rs776022982 Chr.10: 102358606-102358606 on Build GRCh38
rs771031741 Chr.10: 102358494-102358494 on Build GRCh38
rs566020789 Chr.10: 102358571-102358573 on Build GRCh38
rs1027931688 Chr.10: 102358802-102358802 on Build GRCh38
rs907205958 Chr.10: 102358688-102358688 on Build GRCh38
rs541437770 Chr.10: 102358535-102358535 on Build GRCh38
rs747416775 Chr.10: 102358477-102358477 on Build GRCh38
rs891943686 Chr.10: 102358640-102358640 on Build GRCh38
rs1023385466 Chr.10: 102358670-102358670 on Build GRCh38
rs781159917 Chr.10: 102358712-102358712 on Build GRCh38
rs1039189545 Chr.10: 102358509-102358509 on Build GRCh38
rs758632275 Chr.10: 102358552-102358552 on Build GRCh38
rs888013268 Chr.10: 102358447-102358447 on Build GRCh38
rs772810062 Chr.10: 102358615-102358615 on Build GRCh38
rs942222776 Chr.10: 102358832-102358832 on Build GRCh38
rs1033609763 Chr.10: 102358753-102358753 on Build GRCh38
rs983777180 Chr.10: 102358805-102358805 on Build GRCh38
rs760364849 Chr.10: 102358487-102358492 on Build GRCh38
rs1045813948 Chr.10: 102358674-102358674 on Build GRCh38
rs1039344957 Chr.10: 102358839-102358839 on Build GRCh38
rs777762275 Chr.10: 102358569-102358569 on Build GRCh38
rs776700772 Chr.10: 102358540-102358542 on Build GRCh38
rs1030448036 Chr.10: 102358566-102358566 on Build GRCh38
rs770971071 Chr.10: 102358571-102358571 on Build GRCh38
rs950573774 Chr.10: 102358852-102358852 on Build GRCh38
rs756704424 Chr.10: 102358724-102358724 on Build GRCh38
rs895042579 Chr.10: 102358780-102358780 on Build GRCh38
rs747107445 Chr.10: 102358570-102358570 on Build GRCh38
rs1052262771 Chr.10: 102358885-102358885 on Build GRCh38
rs574205961 Chr.10: 102358533-102358533 on Build GRCh38
rs774112751 Chr.10: 102358512-102358512 on Build GRCh38
rs929734368 Chr.10: 102358726-102358726 on Build GRCh38
rs777068534 Chr.10: 102358484-102358484 on Build GRCh38
rs781705779 Chr.10: 102358574-102358574 on Build GRCh38
rs752293431 Chr.10: 102358606-102358608 on Build GRCh38
rs990705544 Chr.10: 102358668-102358668 on Build GRCh38
rs143179363 Chr.10: 102358669-102358669 on Build GRCh38
rs142955445 Chr.10: 102358666-102358666 on Build GRCh38
rs200416208 Chr.10: 102358517-102358517 on Build GRCh38
rs1055008640 Chr.10: 102358588-102358588 on Build GRCh38
rs959201524 Chr.10: 102358469-102358469 on Build GRCh38
rs761593227 Chr.10: 102358513-102358513 on Build GRCh38
rs773010053 Chr.10: 102358483-102358501 on Build GRCh38
rs776602416 Chr.10: 102358659-102358659 on Build GRCh38
rs142579610 Chr.10: 102358521-102358521 on Build GRCh38
rs771472459 Chr.10: 102358481-102358481 on Build GRCh38
rs752490724 Chr.10: 102358685-102358685 on Build GRCh38
rs34232826 Chr.10: 102358860-102358861 on Build GRCh38
rs771859452 Chr.10: 102358611-102358611 on Build GRCh38
rs777518753 Chr.10: 102358547-102358547 on Build GRCh38
rs936184855 Chr.10: 102358573-102358573 on Build GRCh38
rs368983947 Chr.10: 102358711-102358711 on Build GRCh38
rs994748766 Chr.10: 102358792-102358792 on Build GRCh38
rs756830536 Chr.10: 102358836-102358836 on Build GRCh38
rs41293034 Chr.10: 102358516-102358516 on Build GRCh38
rs578059401 Chr.10: 102358789-102358789 on Build GRCh38
rs746399050 Chr.10: 102358583-102358583 on Build GRCh38
rs759171243 Chr.10: 102358542-102358543 on Build GRCh38
rs759502796 Chr.10: 102358663-102358663 on Build GRCh38
rs150542654 Chr.10: 102358527-102358527 on Build GRCh38
rs867006242 Chr.10: 102358608-102358608 on Build GRCh38
rs758256403 Chr.10: 102358702-102358702 on Build GRCh38
rs749669745 Chr.10: 102358742-102358742 on Build GRCh38
rs900661576 Chr.10: 102358854-102358854 on Build GRCh38
rs747747808 Chr.10: 102358609-102358609 on Build GRCh38
rs751773741 Chr.10: 102358709-102358709 on Build GRCh38
rs770240595 Chr.10: 102358596-102358596 on Build GRCh38
rs764360902 Chr.10: 102358703-102358703 on Build GRCh38
rs760219072 Chr.10: 102358643-102358643 on Build GRCh38
rs758138730 Chr.10: 102358538-102358538 on Build GRCh38
rs911580775 Chr.10: 102358528-102358528 on Build GRCh38
rs765856982 Chr.10: 102358657-102358657 on Build GRCh38
rs909548505 Chr.10: 102358810-102358810 on Build GRCh38

View Full Product Details