Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102675642-102676143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL3
Entrez Gene ID
403
Gene Name
ADP ribosylation factor like GTPase 3
Gene Aliases
ARFL3
UniGene
Hs.182215
Gene Chromosome Location
Chr.10: 102673727-102714433 on Build GRCh38
Transcripts
NM_004311.3 XM_017016260.1

SNP Detail

SNP ID Chromosome Location
rs974112848 Chr.10: 102676081-102676081 on Build GRCh38
rs920002551 Chr.10: 102676089-102676089 on Build GRCh38
rs1037309721 Chr.10: 102676080-102676080 on Build GRCh38
rs953851685 Chr.10: 102675919-102675919 on Build GRCh38
rs948810205 Chr.10: 102676055-102676055 on Build GRCh38
rs898628860 Chr.10: 102675748-102675748 on Build GRCh38
rs548583529 Chr.10: 102675994-102675994 on Build GRCh38
rs987140367 Chr.10: 102676038-102676038 on Build GRCh38
rs779697500 Chr.10: 102675857-102675857 on Build GRCh38
rs150463931 Chr.10: 102675758-102675759 on Build GRCh38
rs1001257456 Chr.10: 102675928-102675928 on Build GRCh38
rs1047296568 Chr.10: 102675804-102675804 on Build GRCh38
rs888629032 Chr.10: 102676062-102676062 on Build GRCh38
rs1001678056 Chr.10: 102675844-102675845 on Build GRCh38
rs994338439 Chr.10: 102675760-102675760 on Build GRCh38
rs748688065 Chr.10: 102675843-102675843 on Build GRCh38
rs528770714 Chr.10: 102676020-102676020 on Build GRCh38
rs370434926 Chr.10: 102676021-102676021 on Build GRCh38
rs544289992 Chr.10: 102675812-102675812 on Build GRCh38
rs1023952370 Chr.10: 102675842-102675842 on Build GRCh38
rs190916339 Chr.10: 102675872-102675872 on Build GRCh38
rs565136139 Chr.10: 102676061-102676061 on Build GRCh38
rs989351055 Chr.10: 102675929-102675929 on Build GRCh38
rs988596228 Chr.10: 102675953-102675953 on Build GRCh38
rs573195809 Chr.10: 102675696-102675696 on Build GRCh38
rs186421167 Chr.10: 102675819-102675819 on Build GRCh38
rs1013075261 Chr.10: 102675791-102675791 on Build GRCh38
rs1014279606 Chr.10: 102675841-102675841 on Build GRCh38
rs755783067 Chr.10: 102675681-102675681 on Build GRCh38
rs746621280 Chr.10: 102675944-102675944 on Build GRCh38
rs547405777 Chr.10: 102675943-102675943 on Build GRCh38
rs377447077 Chr.10: 102675680-102675680 on Build GRCh38
rs146155890 Chr.10: 102675837-102675838 on Build GRCh38
rs1032726417 Chr.10: 102675942-102675942 on Build GRCh38
rs915079553 Chr.10: 102675946-102675947 on Build GRCh38
rs11818519 Chr.10: 102675707-102675707 on Build GRCh38
rs1004849962 Chr.10: 102676078-102676078 on Build GRCh38
rs772692566 Chr.10: 102675988-102675988 on Build GRCh38
rs976890447 Chr.10: 102675862-102675862 on Build GRCh38
rs938717608 Chr.10: 102675995-102675995 on Build GRCh38
rs576964304 Chr.10: 102675795-102675795 on Build GRCh38
rs558794083 Chr.10: 102675746-102675746 on Build GRCh38
rs4146427 Chr.10: 102675989-102675989 on Build GRCh38
rs546745422 Chr.10: 102676039-102676039 on Build GRCh38
rs1028105275 Chr.10: 102675903-102675903 on Build GRCh38

View Full Product Details