Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102872306-102872853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AS3MT
Entrez Gene ID
57412
Gene Name
arsenite methyltransferase
Gene Aliases
CYT19
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102869453-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157540 c.171-1G>T p.? Chr.10: 102872447-102872447 on Build GRCh38
5639972 c.238G>A p.G80R Chr.10: 102872515-102872515 on Build GRCh38
683514 c.299G>C p.G100A Chr.10: 102872576-102872576 on Build GRCh38
3666683 c.173A>T p.Y58F Chr.10: 102872450-102872450 on Build GRCh38
6055785 c.299G>T p.G100V Chr.10: 102872576-102872576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376960692 Chr.10: 102872545-102872545 on Build GRCh38
rs1016091485 Chr.10: 102872690-102872690 on Build GRCh38
rs932398439 Chr.10: 102872659-102872659 on Build GRCh38
rs371877430 Chr.10: 102872363-102872370 on Build GRCh38
rs759484385 Chr.10: 102872569-102872569 on Build GRCh38
rs749709556 Chr.10: 102872644-102872644 on Build GRCh38
rs545209508 Chr.10: 102872373-102872373 on Build GRCh38
rs746395314 Chr.10: 102872463-102872463 on Build GRCh38
rs764946632 Chr.10: 102872572-102872572 on Build GRCh38
rs753599180 Chr.10: 102872404-102872404 on Build GRCh38
rs756749151 Chr.10: 102872635-102872635 on Build GRCh38
rs968300059 Chr.10: 102872539-102872539 on Build GRCh38
rs757323706 Chr.10: 102872593-102872593 on Build GRCh38
rs776452061 Chr.10: 102872449-102872451 on Build GRCh38
rs556868245 Chr.10: 102872495-102872495 on Build GRCh38
rs751566360 Chr.10: 102872589-102872589 on Build GRCh38
rs200044145 Chr.10: 102872546-102872546 on Build GRCh38
rs34028617 Chr.10: 102872430-102872430 on Build GRCh38
rs1049548098 Chr.10: 102872672-102872672 on Build GRCh38
rs767715476 Chr.10: 102872599-102872599 on Build GRCh38
rs922272020 Chr.10: 102872823-102872823 on Build GRCh38
rs974515423 Chr.10: 102872353-102872353 on Build GRCh38
rs566118458 Chr.10: 102872828-102872828 on Build GRCh38
rs766387558 Chr.10: 102872403-102872403 on Build GRCh38
rs897147257 Chr.10: 102872727-102872727 on Build GRCh38
rs963306724 Chr.10: 102872334-102872337 on Build GRCh38
rs1015574876 Chr.10: 102872391-102872391 on Build GRCh38
rs961388465 Chr.10: 102872392-102872392 on Build GRCh38
rs765039549 Chr.10: 102872414-102872414 on Build GRCh38
rs776502872 Chr.10: 102872554-102872554 on Build GRCh38
rs141498202 Chr.10: 102872785-102872785 on Build GRCh38
rs372624494 Chr.10: 102872354-102872361 on Build GRCh38
rs771669666 Chr.10: 102872778-102872778 on Build GRCh38
rs941234755 Chr.10: 102872691-102872691 on Build GRCh38
rs772843530 Chr.10: 102872402-102872402 on Build GRCh38
rs537534393 Chr.10: 102872820-102872820 on Build GRCh38
rs764428592 Chr.10: 102872588-102872588 on Build GRCh38
rs375290999 Chr.10: 102872729-102872729 on Build GRCh38
rs750890056 Chr.10: 102872604-102872604 on Build GRCh38
rs555945086 Chr.10: 102872824-102872824 on Build GRCh38
rs574687813 Chr.10: 102872338-102872338 on Build GRCh38
rs771414706 Chr.10: 102872397-102872397 on Build GRCh38
rs200487609 Chr.10: 102872519-102872519 on Build GRCh38
rs986866139 Chr.10: 102872422-102872422 on Build GRCh38
rs17115188 Chr.10: 102872815-102872815 on Build GRCh38
rs1031246432 Chr.10: 102872810-102872810 on Build GRCh38
rs780567314 Chr.10: 102872490-102872490 on Build GRCh38
rs762373668 Chr.10: 102872647-102872647 on Build GRCh38
rs1022435555 Chr.10: 102872792-102872792 on Build GRCh38
rs978299033 Chr.10: 102872808-102872808 on Build GRCh38
rs777712128 Chr.10: 102872433-102872433 on Build GRCh38
rs549165947 Chr.10: 102872510-102872510 on Build GRCh38
rs528090351 Chr.10: 102872580-102872580 on Build GRCh38
rs968208859 Chr.10: 102872793-102872793 on Build GRCh38
rs749172135 Chr.10: 102872464-102872464 on Build GRCh38
rs147889757 Chr.10: 102872657-102872657 on Build GRCh38
rs747816486 Chr.10: 102872679-102872679 on Build GRCh38
rs370458438 Chr.10: 102872550-102872550 on Build GRCh38
rs772572895 Chr.10: 102872531-102872531 on Build GRCh38
rs373783784 Chr.10: 102872523-102872523 on Build GRCh38
rs767361004 Chr.10: 102872476-102872485 on Build GRCh38
rs750316257 Chr.10: 102872631-102872631 on Build GRCh38
rs115224622 Chr.10: 102872754-102872754 on Build GRCh38
rs759286397 Chr.10: 102872409-102872409 on Build GRCh38
rs1003612304 Chr.10: 102872417-102872417 on Build GRCh38
rs144713814 Chr.10: 102872544-102872544 on Build GRCh38
rs371442321 Chr.10: 102872436-102872436 on Build GRCh38
rs149960556 Chr.10: 102872829-102872829 on Build GRCh38
rs770222107 Chr.10: 102872488-102872488 on Build GRCh38
rs560096528 Chr.10: 102872439-102872439 on Build GRCh38
rs780550865 Chr.10: 102872638-102872638 on Build GRCh38
rs757071866 Chr.10: 102872420-102872421 on Build GRCh38
rs768336799 Chr.10: 102872594-102872594 on Build GRCh38
rs751370259 Chr.10: 102872402-102872402 on Build GRCh38
rs888334052 Chr.10: 102872361-102872361 on Build GRCh38
rs374532111 Chr.10: 102872425-102872425 on Build GRCh38
rs551984793 Chr.10: 102872814-102872814 on Build GRCh38
rs1035433748 Chr.10: 102872777-102872777 on Build GRCh38
rs796610749 Chr.10: 102872401-102872401 on Build GRCh38
rs763075282 Chr.10: 102872584-102872584 on Build GRCh38
rs1036975342 Chr.10: 102872707-102872707 on Build GRCh38
rs80317306 Chr.10: 102872460-102872460 on Build GRCh38

View Full Product Details