Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103479059-103479558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALHM3
Entrez Gene ID
119395
Gene Name
calcium homeostasis modulator 3
Gene Aliases
FAM26A, bA225H22.7
UniGene
Hs.364624
Gene Chromosome Location
Chr.10: 103472804-103479240 on Build GRCh38
Transcripts
NM_001129742.1

SNP Detail

SNP ID Chromosome Location
rs533096075 Chr.10: 103479142-103479142 on Build GRCh38
rs1048528953 Chr.10: 103479310-103479310 on Build GRCh38
rs1015806450 Chr.10: 103479328-103479328 on Build GRCh38
rs545957409 Chr.10: 103479508-103479508 on Build GRCh38
rs868291508 Chr.10: 103479364-103479364 on Build GRCh38
rs1013775680 Chr.10: 103479162-103479162 on Build GRCh38
rs986708676 Chr.10: 103479383-103479383 on Build GRCh38
rs60099062 Chr.10: 103479164-103479164 on Build GRCh38
rs893030721 Chr.10: 103479493-103479504 on Build GRCh38
rs536122332 Chr.10: 103479449-103479449 on Build GRCh38
rs1048775312 Chr.10: 103479530-103479530 on Build GRCh38
rs193229202 Chr.10: 103479318-103479318 on Build GRCh38
rs915349282 Chr.10: 103479399-103479399 on Build GRCh38
rs1004807340 Chr.10: 103479317-103479317 on Build GRCh38
rs779454380 Chr.10: 103479329-103479329 on Build GRCh38
rs553976327 Chr.10: 103479453-103479453 on Build GRCh38
rs562538902 Chr.10: 103479507-103479507 on Build GRCh38
rs529186998 Chr.10: 103479363-103479363 on Build GRCh38
rs758989184 Chr.10: 103479384-103479384 on Build GRCh38
rs1047479213 Chr.10: 103479126-103479126 on Build GRCh38
rs966532796 Chr.10: 103479433-103479433 on Build GRCh38
rs551518070 Chr.10: 103479143-103479143 on Build GRCh38
rs780742151 Chr.10: 103479386-103479386 on Build GRCh38
rs969521678 Chr.10: 103479201-103479201 on Build GRCh38
rs927897432 Chr.10: 103479489-103479489 on Build GRCh38
rs11594852 Chr.10: 103479272-103479272 on Build GRCh38
rs886190280 Chr.10: 103479534-103479534 on Build GRCh38
rs1023800901 Chr.10: 103479183-103479183 on Build GRCh38
rs1055230759 Chr.10: 103479192-103479192 on Build GRCh38
rs994196871 Chr.10: 103479116-103479116 on Build GRCh38
rs568705391 Chr.10: 103479448-103479448 on Build GRCh38
rs370132122 Chr.10: 103479166-103479166 on Build GRCh38
rs371530917 Chr.10: 103479314-103479315 on Build GRCh38
rs764285977 Chr.10: 103479186-103479186 on Build GRCh38
rs1555823 Chr.10: 103479506-103479506 on Build GRCh38
rs746313523 Chr.10: 103479361-103479361 on Build GRCh38
rs896765170 Chr.10: 103479153-103479153 on Build GRCh38
rs756701832 Chr.10: 103479079-103479080 on Build GRCh38
rs955314289 Chr.10: 103479359-103479359 on Build GRCh38
rs912870500 Chr.10: 103479344-103479344 on Build GRCh38
rs954150700 Chr.10: 103479125-103479125 on Build GRCh38
rs989295408 Chr.10: 103479149-103479149 on Build GRCh38
rs1038585362 Chr.10: 103479491-103479491 on Build GRCh38
rs536651066 Chr.10: 103479304-103479304 on Build GRCh38
rs942510697 Chr.10: 103479487-103479487 on Build GRCh38
rs927875339 Chr.10: 103479172-103479172 on Build GRCh38
rs757651906 Chr.10: 103479301-103479301 on Build GRCh38
rs1000954462 Chr.10: 103479210-103479210 on Build GRCh38

View Full Product Details