Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116058779-116059282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA1
Entrez Gene ID
2674
Gene Name
GDNF family receptor alpha 1
Gene Aliases
GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1
UniGene
Hs.388347
Gene Chromosome Location
Chr.10: 116056925-116273645 on Build GRCh38
Transcripts
NM_001145453.2 NM_005264.5 NM_145793.3 XM_005269696.1 XM_011539634.1

SNP Detail

SNP ID Chromosome Location
rs76376736 Chr.10: 116059163-116059163 on Build GRCh38
rs527419060 Chr.10: 116059137-116059137 on Build GRCh38
rs988636535 Chr.10: 116059021-116059021 on Build GRCh38
rs867060607 Chr.10: 116059064-116059064 on Build GRCh38
rs1033078395 Chr.10: 116058826-116058826 on Build GRCh38
rs180554 Chr.10: 116059049-116059049 on Build GRCh38
rs2052012 Chr.10: 116058958-116058958 on Build GRCh38
rs940502427 Chr.10: 116059087-116059087 on Build GRCh38
rs1001108541 Chr.10: 116059000-116059000 on Build GRCh38
rs988239252 Chr.10: 116058843-116058843 on Build GRCh38
rs896222402 Chr.10: 116059100-116059100 on Build GRCh38
rs879685634 Chr.10: 116059152-116059152 on Build GRCh38
rs529363359 Chr.10: 116058832-116058832 on Build GRCh38
rs988897275 Chr.10: 116058990-116058990 on Build GRCh38
rs75268639 Chr.10: 116058970-116058970 on Build GRCh38
rs748473039 Chr.10: 116059089-116059089 on Build GRCh38
rs937002043 Chr.10: 116058870-116058870 on Build GRCh38
rs551708768 Chr.10: 116059164-116059164 on Build GRCh38
rs550991312 Chr.10: 116058844-116058844 on Build GRCh38
rs186307553 Chr.10: 116058846-116058846 on Build GRCh38
rs1011906017 Chr.10: 116058929-116058929 on Build GRCh38
rs949829986 Chr.10: 116058849-116058849 on Build GRCh38
rs147345918 Chr.10: 116059014-116059014 on Build GRCh38
rs4367865 Chr.10: 116058830-116058830 on Build GRCh38
rs971235609 Chr.10: 116059057-116059057 on Build GRCh38
rs919594657 Chr.10: 116059136-116059136 on Build GRCh38
rs906094838 Chr.10: 116058940-116058940 on Build GRCh38
rs905754922 Chr.10: 116058866-116058866 on Build GRCh38
rs192063975 Chr.10: 116059016-116059016 on Build GRCh38
rs368830182 Chr.10: 116059068-116059068 on Build GRCh38
rs1007920212 Chr.10: 116059175-116059175 on Build GRCh38
rs189104185 Chr.10: 116059142-116059142 on Build GRCh38
rs149332532 Chr.10: 116058963-116058963 on Build GRCh38
rs940386936 Chr.10: 116058888-116058888 on Build GRCh38
rs780210364 Chr.10: 116059020-116059020 on Build GRCh38
rs1038473381 Chr.10: 116058876-116058876 on Build GRCh38
rs994357597 Chr.10: 116058909-116058909 on Build GRCh38
rs1018496604 Chr.10: 116058998-116058998 on Build GRCh38
rs975101010 Chr.10: 116058844-116058844 on Build GRCh38
rs180553 Chr.10: 116059072-116059072 on Build GRCh38
rs937265138 Chr.10: 116059083-116059083 on Build GRCh38
rs577546202 Chr.10: 116059046-116059046 on Build GRCh38
rs551672873 Chr.10: 116058939-116058939 on Build GRCh38
rs911856389 Chr.10: 116058840-116058840 on Build GRCh38
rs554996897 Chr.10: 116058859-116058859 on Build GRCh38
rs61203890 Chr.10: 116058904-116058904 on Build GRCh38
rs572511814 Chr.10: 116058880-116058880 on Build GRCh38
rs375727767 Chr.10: 116058803-116058803 on Build GRCh38
rs183245964 Chr.10: 116059088-116059088 on Build GRCh38
rs951393116 Chr.10: 116058992-116058992 on Build GRCh38
rs544357087 Chr.10: 116059144-116059144 on Build GRCh38
rs1056143338 Chr.10: 116058900-116058900 on Build GRCh38
rs772590550 Chr.10: 116059145-116059145 on Build GRCh38
rs546706997 Chr.10: 116058813-116058813 on Build GRCh38
rs566727979 Chr.10: 116059228-116059228 on Build GRCh38
rs925296882 Chr.10: 116058809-116058809 on Build GRCh38
rs373783787 Chr.10: 116058999-116058999 on Build GRCh38
rs562922140 Chr.10: 116059162-116059162 on Build GRCh38
rs556161225 Chr.10: 116059032-116059032 on Build GRCh38
rs891999298 Chr.10: 116059180-116059180 on Build GRCh38

View Full Product Details