Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116062197-116062701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA1
Entrez Gene ID
2674
Gene Name
GDNF family receptor alpha 1
Gene Aliases
GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1
UniGene
Hs.388347
Gene Chromosome Location
Chr.10: 116056925-116273645 on Build GRCh38
Transcripts
NM_001145453.2 NM_005264.5 NM_145793.3 XM_005269696.1 XM_011539634.1

SNP Detail

SNP ID Chromosome Location
rs182055027 Chr.10: 116062450-116062450 on Build GRCh38
rs998949140 Chr.10: 116062245-116062245 on Build GRCh38
rs902715224 Chr.10: 116062436-116062436 on Build GRCh38
rs371108019 Chr.10: 116062600-116062600 on Build GRCh38
rs545864247 Chr.10: 116062356-116062356 on Build GRCh38
rs969593400 Chr.10: 116062665-116062665 on Build GRCh38
rs1025435953 Chr.10: 116062223-116062223 on Build GRCh38
rs1043669045 Chr.10: 116062547-116062547 on Build GRCh38
rs145758999 Chr.10: 116062598-116062598 on Build GRCh38
rs946932173 Chr.10: 116062364-116062364 on Build GRCh38
rs947699480 Chr.10: 116062523-116062523 on Build GRCh38
rs751166980 Chr.10: 116062292-116062292 on Build GRCh38
rs1023876716 Chr.10: 116062296-116062296 on Build GRCh38
rs922993907 Chr.10: 116062457-116062457 on Build GRCh38
rs772159164 Chr.10: 116062329-116062332 on Build GRCh38
rs1013399887 Chr.10: 116062618-116062618 on Build GRCh38
rs760647955 Chr.10: 116062466-116062469 on Build GRCh38
rs540968181 Chr.10: 116062334-116062334 on Build GRCh38
rs1035739376 Chr.10: 116062278-116062278 on Build GRCh38
rs908889373 Chr.10: 116062396-116062396 on Build GRCh38
rs143591190 Chr.10: 116062261-116062261 on Build GRCh38
rs1050636830 Chr.10: 116062505-116062505 on Build GRCh38
rs1025972755 Chr.10: 116062553-116062554 on Build GRCh38
rs768403936 Chr.10: 116062492-116062493 on Build GRCh38
rs889283037 Chr.10: 116062508-116062508 on Build GRCh38
rs764260029 Chr.10: 116062293-116062293 on Build GRCh38
rs1057252312 Chr.10: 116062588-116062588 on Build GRCh38
rs1006908421 Chr.10: 116062544-116062545 on Build GRCh38
rs1051213413 Chr.10: 116062482-116062482 on Build GRCh38
rs762058710 Chr.10: 116062559-116062559 on Build GRCh38
rs1043953737 Chr.10: 116062366-116062366 on Build GRCh38
rs115468103 Chr.10: 116062229-116062229 on Build GRCh38
rs998834311 Chr.10: 116062551-116062551 on Build GRCh38
rs958416099 Chr.10: 116062621-116062621 on Build GRCh38
rs532882830 Chr.10: 116062550-116062550 on Build GRCh38
rs933055855 Chr.10: 116062458-116062458 on Build GRCh38
rs375288059 Chr.10: 116062652-116062652 on Build GRCh38
rs971804166 Chr.10: 116062455-116062455 on Build GRCh38
rs573344195 Chr.10: 116062551-116062552 on Build GRCh38
rs995460456 Chr.10: 116062553-116062553 on Build GRCh38
rs1006139601 Chr.10: 116062670-116062670 on Build GRCh38
rs36114266 Chr.10: 116062234-116062235 on Build GRCh38
rs891180998 Chr.10: 116062507-116062507 on Build GRCh38
rs373387421 Chr.10: 116062242-116062243 on Build GRCh38
rs985105010 Chr.10: 116062374-116062374 on Build GRCh38
rs960549481 Chr.10: 116062290-116062290 on Build GRCh38
rs566155541 Chr.10: 116062667-116062667 on Build GRCh38

View Full Product Details