Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116341683-116342200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC172
Entrez Gene ID
374355
Gene Name
coiled-coil domain containing 172
Gene Aliases
C10orf96
UniGene
Hs.233407
Gene Chromosome Location
Chr.10: 116324097-116380029 on Build GRCh38
Transcripts
NM_198515.2 XM_006717820.3 XM_011539771.2 XM_017016206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246568 c.352G>A p.D118N Chr.10: 116342105-116342105 on Build GRCh38
1675593 c.376G>A p.E126K Chr.10: 116342129-116342129 on Build GRCh38
6030521 c.402G>T p.K134N Chr.10: 116342155-116342155 on Build GRCh38
7078564 c.387G>T p.M129I Chr.10: 116342140-116342140 on Build GRCh38
1474382 c.353delA p.D118fs*4 Chr.10: 116342106-116342106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778200621 Chr.10: 116342166-116342166 on Build GRCh38
rs370853531 Chr.10: 116342071-116342071 on Build GRCh38
rs754378767 Chr.10: 116342131-116342131 on Build GRCh38
rs748213847 Chr.10: 116342139-116342139 on Build GRCh38
rs773534759 Chr.10: 116342033-116342033 on Build GRCh38
rs899229584 Chr.10: 116341963-116341963 on Build GRCh38
rs763674212 Chr.10: 116342053-116342053 on Build GRCh38
rs759456413 Chr.10: 116342044-116342044 on Build GRCh38
rs139809289 Chr.10: 116342052-116342052 on Build GRCh38
rs763255969 Chr.10: 116342127-116342127 on Build GRCh38
rs765328815 Chr.10: 116342112-116342112 on Build GRCh38
rs750987400 Chr.10: 116342066-116342066 on Build GRCh38
rs773589618 Chr.10: 116341987-116341987 on Build GRCh38
rs1034792176 Chr.10: 116341899-116341899 on Build GRCh38
rs771434552 Chr.10: 116342040-116342041 on Build GRCh38
rs976345845 Chr.10: 116342065-116342065 on Build GRCh38
rs755387650 Chr.10: 116342132-116342132 on Build GRCh38
rs561198614 Chr.10: 116341919-116341919 on Build GRCh38
rs201698300 Chr.10: 116342107-116342107 on Build GRCh38
rs772377437 Chr.10: 116342028-116342028 on Build GRCh38
rs931836084 Chr.10: 116341889-116341889 on Build GRCh38
rs866518323 Chr.10: 116342042-116342042 on Build GRCh38
rs756538004 Chr.10: 116342097-116342097 on Build GRCh38
rs768785665 Chr.10: 116341998-116341998 on Build GRCh38
rs1056677266 Chr.10: 116342141-116342141 on Build GRCh38
rs374073681 Chr.10: 116341997-116341997 on Build GRCh38
rs750061944 Chr.10: 116342095-116342095 on Build GRCh38
rs902393800 Chr.10: 116341788-116341788 on Build GRCh38
rs774234460 Chr.10: 116342001-116342001 on Build GRCh38
rs983106638 Chr.10: 116341994-116341994 on Build GRCh38
rs759582063 Chr.10: 116342073-116342074 on Build GRCh38
rs748421213 Chr.10: 116342010-116342010 on Build GRCh38
rs777108726 Chr.10: 116342044-116342047 on Build GRCh38
rs144107549 Chr.10: 116341766-116341766 on Build GRCh38
rs139493735 Chr.10: 116341976-116341976 on Build GRCh38
rs868571860 Chr.10: 116342111-116342111 on Build GRCh38
rs532636333 Chr.10: 116342102-116342102 on Build GRCh38
rs572443026 Chr.10: 116341858-116341858 on Build GRCh38
rs892715952 Chr.10: 116342170-116342170 on Build GRCh38
rs779078810 Chr.10: 116342135-116342135 on Build GRCh38
rs767204397 Chr.10: 116342085-116342085 on Build GRCh38
rs78019944 Chr.10: 116342159-116342159 on Build GRCh38
rs77733004 Chr.10: 116341913-116341913 on Build GRCh38
rs772259916 Chr.10: 116342165-116342165 on Build GRCh38
rs1003872286 Chr.10: 116341813-116341813 on Build GRCh38
rs200926297 Chr.10: 116341993-116341993 on Build GRCh38
rs753631347 Chr.10: 116341810-116341810 on Build GRCh38
rs764161004 Chr.10: 116342144-116342145 on Build GRCh38
rs752679290 Chr.10: 116342116-116342118 on Build GRCh38
rs868051108 Chr.10: 116342060-116342060 on Build GRCh38
rs894994195 Chr.10: 116341907-116341907 on Build GRCh38
rs375961590 Chr.10: 116342047-116342047 on Build GRCh38
rs987399310 Chr.10: 116341901-116341901 on Build GRCh38
rs760974417 Chr.10: 116341996-116341999 on Build GRCh38
rs34746230 Chr.10: 116342032-116342032 on Build GRCh38
rs73374933 Chr.10: 116341924-116341924 on Build GRCh38

View Full Product Details