Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116668166-116668669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf82
Entrez Gene ID
143379
Gene Name
chromosome 10 open reading frame 82
Gene Aliases
-
UniGene
Hs.121347
Gene Chromosome Location
Chr.10: 116662211-116670305 on Build GRCh38
Transcripts
NM_144661.2 XM_011539340.2 XM_011539341.1 XM_011539342.2 XM_011539343.2 XM_011539344.2 XM_011539345.2 XM_011539346.2 XM_011539347.2 XM_017015753.1 XM_017015754.1 XM_017015755.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894617 c.26G>C p.R9T Chr.10: 116668287-116668287 on Build GRCh38
4820450 c.31C>T p.L11L Chr.10: 116668282-116668282 on Build GRCh38
6894619 c.26G>C p.R9T Chr.10: 116668287-116668287 on Build GRCh38
6343090 c.21C>A p.F7L Chr.10: 116668292-116668292 on Build GRCh38
6894618 c.26G>C p.R9T Chr.10: 116668287-116668287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543742423 Chr.10: 116668258-116668258 on Build GRCh38
rs368633769 Chr.10: 116668214-116668214 on Build GRCh38
rs782058275 Chr.10: 116668287-116668287 on Build GRCh38
rs781798887 Chr.10: 116668242-116668242 on Build GRCh38
rs1039719177 Chr.10: 116668337-116668337 on Build GRCh38
rs781859772 Chr.10: 116668301-116668301 on Build GRCh38
rs782489940 Chr.10: 116668235-116668235 on Build GRCh38
rs781921024 Chr.10: 116668219-116668219 on Build GRCh38
rs2901117 Chr.10: 116668268-116668268 on Build GRCh38
rs183605940 Chr.10: 116668339-116668339 on Build GRCh38
rs532285933 Chr.10: 116668495-116668495 on Build GRCh38
rs373639179 Chr.10: 116668390-116668391 on Build GRCh38
rs1025594758 Chr.10: 116668519-116668519 on Build GRCh38
rs201351111 Chr.10: 116668259-116668259 on Build GRCh38
rs782083735 Chr.10: 116668277-116668277 on Build GRCh38
rs17095092 Chr.10: 116668188-116668188 on Build GRCh38
rs375511136 Chr.10: 116668352-116668352 on Build GRCh38
rs147507078 Chr.10: 116668314-116668314 on Build GRCh38
rs560907374 Chr.10: 116668363-116668363 on Build GRCh38
rs782382774 Chr.10: 116668212-116668212 on Build GRCh38
rs565785000 Chr.10: 116668571-116668571 on Build GRCh38
rs782779889 Chr.10: 116668232-116668232 on Build GRCh38
rs782803718 Chr.10: 116668294-116668294 on Build GRCh38
rs781784283 Chr.10: 116668284-116668284 on Build GRCh38
rs781893010 Chr.10: 116668325-116668325 on Build GRCh38
rs782608656 Chr.10: 116668309-116668309 on Build GRCh38
rs377561725 Chr.10: 116668353-116668353 on Build GRCh38
rs782578620 Chr.10: 116668316-116668316 on Build GRCh38
rs140130227 Chr.10: 116668320-116668321 on Build GRCh38
rs375309699 Chr.10: 116668234-116668234 on Build GRCh38
rs369905967 Chr.10: 116668246-116668246 on Build GRCh38
rs781883666 Chr.10: 116668220-116668220 on Build GRCh38
rs192588615 Chr.10: 116668403-116668403 on Build GRCh38
rs782163689 Chr.10: 116668265-116668265 on Build GRCh38
rs372882482 Chr.10: 116668279-116668279 on Build GRCh38
rs782626974 Chr.10: 116668250-116668250 on Build GRCh38
rs892605450 Chr.10: 116668451-116668451 on Build GRCh38
rs144531690 Chr.10: 116668455-116668455 on Build GRCh38
rs782272854 Chr.10: 116668247-116668247 on Build GRCh38
rs753753005 Chr.10: 116668192-116668192 on Build GRCh38
rs781997877 Chr.10: 116668276-116668276 on Build GRCh38
rs970948188 Chr.10: 116668520-116668520 on Build GRCh38
rs782527148 Chr.10: 116668306-116668306 on Build GRCh38
rs782571141 Chr.10: 116668546-116668546 on Build GRCh38
rs187406999 Chr.10: 116668383-116668383 on Build GRCh38
rs183222341 Chr.10: 116668595-116668595 on Build GRCh38
rs782557315 Chr.10: 116668245-116668245 on Build GRCh38
rs782293879 Chr.10: 116668554-116668554 on Build GRCh38
rs534801848 Chr.10: 116668596-116668596 on Build GRCh38
rs950543789 Chr.10: 116668605-116668605 on Build GRCh38
rs782454511 Chr.10: 116668389-116668389 on Build GRCh38
rs552108367 Chr.10: 116668501-116668501 on Build GRCh38
rs782071344 Chr.10: 116668222-116668222 on Build GRCh38
rs782178347 Chr.10: 116668253-116668253 on Build GRCh38
rs374210971 Chr.10: 116668213-116668213 on Build GRCh38
rs782353554 Chr.10: 116668248-116668248 on Build GRCh38
rs782554342 Chr.10: 116668238-116668238 on Build GRCh38
rs564875326 Chr.10: 116668318-116668318 on Build GRCh38
rs973531970 Chr.10: 116668580-116668580 on Build GRCh38
rs782540012 Chr.10: 116668331-116668331 on Build GRCh38
rs981031432 Chr.10: 116668527-116668527 on Build GRCh38
rs781827442 Chr.10: 116668315-116668315 on Build GRCh38
rs138253124 Chr.10: 116668374-116668375 on Build GRCh38
rs371343136 Chr.10: 116668224-116668224 on Build GRCh38
rs987309103 Chr.10: 116668632-116668632 on Build GRCh38
rs782116470 Chr.10: 116668227-116668227 on Build GRCh38
rs372010572 Chr.10: 116668295-116668295 on Build GRCh38

View Full Product Details