Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116871026-116871575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
NM_001242699.1 XM_005269815.3 XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3686424 c.1191T>A p.A397A Chr.10: 116871259-116871259 on Build GRCh38
6499912 c.1120C>T p.Q374* Chr.10: 116871188-116871188 on Build GRCh38
6895163 c.1108G>A p.D370N Chr.10: 116871176-116871176 on Build GRCh38
7003792 c.1160G>T p.G387V Chr.10: 116871228-116871228 on Build GRCh38
4852575 c.1165G>C p.E389Q Chr.10: 116871233-116871233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535458856 Chr.10: 116871087-116871087 on Build GRCh38
rs534551504 Chr.10: 116871316-116871316 on Build GRCh38
rs930697263 Chr.10: 116871093-116871093 on Build GRCh38
rs950698635 Chr.10: 116871363-116871365 on Build GRCh38
rs761041562 Chr.10: 116871398-116871402 on Build GRCh38
rs569337651 Chr.10: 116871204-116871204 on Build GRCh38
rs949851779 Chr.10: 116871141-116871141 on Build GRCh38
rs541398336 Chr.10: 116871370-116871370 on Build GRCh38
rs574954551 Chr.10: 116871540-116871540 on Build GRCh38
rs966258229 Chr.10: 116871354-116871354 on Build GRCh38
rs1017462569 Chr.10: 116871329-116871329 on Build GRCh38
rs1020437810 Chr.10: 116871321-116871321 on Build GRCh38
rs745765645 Chr.10: 116871252-116871252 on Build GRCh38
rs759723060 Chr.10: 116871214-116871214 on Build GRCh38
rs572824970 Chr.10: 116871324-116871324 on Build GRCh38
rs533418738 Chr.10: 116871067-116871067 on Build GRCh38
rs771710593 Chr.10: 116871550-116871553 on Build GRCh38
rs998451010 Chr.10: 116871277-116871277 on Build GRCh38
rs958580965 Chr.10: 116871499-116871499 on Build GRCh38
rs1055331514 Chr.10: 116871111-116871111 on Build GRCh38
rs748793187 Chr.10: 116871297-116871297 on Build GRCh38
rs561461892 Chr.10: 116871528-116871528 on Build GRCh38
rs745427148 Chr.10: 116871228-116871228 on Build GRCh38
rs538495243 Chr.10: 116871400-116871400 on Build GRCh38
rs769399046 Chr.10: 116871267-116871267 on Build GRCh38
rs551637225 Chr.10: 116871293-116871293 on Build GRCh38
rs773557581 Chr.10: 116871395-116871397 on Build GRCh38
rs755278012 Chr.10: 116871137-116871137 on Build GRCh38
rs781348236 Chr.10: 116871238-116871238 on Build GRCh38
rs776188982 Chr.10: 116871355-116871355 on Build GRCh38
rs757108146 Chr.10: 116871223-116871223 on Build GRCh38
rs1007887804 Chr.10: 116871323-116871323 on Build GRCh38
rs900750425 Chr.10: 116871183-116871183 on Build GRCh38
rs981675046 Chr.10: 116871359-116871359 on Build GRCh38
rs991460920 Chr.10: 116871401-116871401 on Build GRCh38
rs773891436 Chr.10: 116871143-116871143 on Build GRCh38
rs183734752 Chr.10: 116871066-116871066 on Build GRCh38
rs566890704 Chr.10: 116871069-116871069 on Build GRCh38
rs886633744 Chr.10: 116871115-116871115 on Build GRCh38
rs918119791 Chr.10: 116871451-116871451 on Build GRCh38
rs558333547 Chr.10: 116871208-116871208 on Build GRCh38
rs578109845 Chr.10: 116871292-116871292 on Build GRCh38
rs959650048 Chr.10: 116871400-116871401 on Build GRCh38
rs927233471 Chr.10: 116871485-116871485 on Build GRCh38
rs766345842 Chr.10: 116871107-116871107 on Build GRCh38
rs751411499 Chr.10: 116871165-116871165 on Build GRCh38
rs902867220 Chr.10: 116871187-116871187 on Build GRCh38
rs1026793970 Chr.10: 116871357-116871357 on Build GRCh38
rs12251703 Chr.10: 116871318-116871318 on Build GRCh38
rs971285869 Chr.10: 116871352-116871352 on Build GRCh38
rs920685286 Chr.10: 116871070-116871070 on Build GRCh38
rs753905484 Chr.10: 116871114-116871114 on Build GRCh38
rs779206215 Chr.10: 116871159-116871159 on Build GRCh38
rs920570522 Chr.10: 116871530-116871530 on Build GRCh38
rs549519008 Chr.10: 116871174-116871174 on Build GRCh38
rs981407759 Chr.10: 116871055-116871055 on Build GRCh38
rs918076143 Chr.10: 116871134-116871134 on Build GRCh38
rs990792176 Chr.10: 116871523-116871523 on Build GRCh38
rs770377603 Chr.10: 116871264-116871264 on Build GRCh38
rs533304526 Chr.10: 116871062-116871062 on Build GRCh38
rs538446997 Chr.10: 116871207-116871207 on Build GRCh38
rs760847393 Chr.10: 116871090-116871090 on Build GRCh38

View Full Product Details