Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119651662-119652152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG3
Entrez Gene ID
9531
Gene Name
BCL2 associated athanogene 3
Gene Aliases
BAG-3, BIS, CAIR-1, MFM6
UniGene
Hs.523309
Gene Chromosome Location
Chr.10: 119651347-119677819 on Build GRCh38
Transcripts
NM_004281.3 XM_005270287.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609567 c.75A>G p.G25G Chr.10: 119651750-119651750 on Build GRCh38
1296990 c.3G>A p.M1I Chr.10: 119651678-119651678 on Build GRCh38
6268291 c.39G>T p.A13A Chr.10: 119651714-119651714 on Build GRCh38
4012086 c.44G>A p.G15D Chr.10: 119651719-119651719 on Build GRCh38
1346127 c.67delC p.G25fs*186 Chr.10: 119651742-119651742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543209392 Chr.10: 119651874-119651874 on Build GRCh38
rs137965903 Chr.10: 119651700-119651700 on Build GRCh38
rs988033291 Chr.10: 119651840-119651840 on Build GRCh38
rs112546447 Chr.10: 119651898-119651898 on Build GRCh38
rs2577336 Chr.10: 119652124-119652124 on Build GRCh38
rs201329879 Chr.10: 119651803-119651803 on Build GRCh38
rs2736422 Chr.10: 119651941-119651941 on Build GRCh38
rs727504929 Chr.10: 119651789-119651789 on Build GRCh38
rs138785335 Chr.10: 119651848-119651848 on Build GRCh38
rs764255227 Chr.10: 119651862-119651862 on Build GRCh38
rs970509662 Chr.10: 119652126-119652126 on Build GRCh38
rs563146298 Chr.10: 119651970-119651970 on Build GRCh38
rs1013686226 Chr.10: 119651959-119651959 on Build GRCh38
rs794728980 Chr.10: 119651817-119651817 on Build GRCh38
rs956429406 Chr.10: 119651682-119651682 on Build GRCh38
rs1017727288 Chr.10: 119652093-119652093 on Build GRCh38
rs772652580 Chr.10: 119651750-119651750 on Build GRCh38
rs964684316 Chr.10: 119651747-119651747 on Build GRCh38
rs764177199 Chr.10: 119651784-119651784 on Build GRCh38
rs981891257 Chr.10: 119652133-119652133 on Build GRCh38
rs778921699 Chr.10: 119651804-119651804 on Build GRCh38
rs772351208 Chr.10: 119651741-119651742 on Build GRCh38
rs145934400 Chr.10: 119651777-119651777 on Build GRCh38
rs1038378989 Chr.10: 119652064-119652064 on Build GRCh38
rs142391650 Chr.10: 119651749-119651749 on Build GRCh38
rs777752849 Chr.10: 119651688-119651688 on Build GRCh38
rs989198918 Chr.10: 119651947-119651947 on Build GRCh38
rs543356821 Chr.10: 119651926-119651926 on Build GRCh38
rs1030653325 Chr.10: 119651933-119651933 on Build GRCh38
rs777267711 Chr.10: 119651813-119651813 on Build GRCh38
rs537615906 Chr.10: 119651802-119651802 on Build GRCh38
rs727502895 Chr.10: 119651724-119651724 on Build GRCh38
rs866659159 Chr.10: 119651729-119651729 on Build GRCh38
rs200072558 Chr.10: 119651778-119651778 on Build GRCh38
rs1033425829 Chr.10: 119652117-119652117 on Build GRCh38
rs948948012 Chr.10: 119652075-119652075 on Build GRCh38
rs576579239 Chr.10: 119652044-119652044 on Build GRCh38
rs963830528 Chr.10: 119651969-119651969 on Build GRCh38
rs1046307004 Chr.10: 119652116-119652116 on Build GRCh38
rs926308865 Chr.10: 119651909-119651909 on Build GRCh38
rs890591345 Chr.10: 119652047-119652047 on Build GRCh38
rs779929078 Chr.10: 119651703-119651703 on Build GRCh38
rs778790962 Chr.10: 119651878-119651878 on Build GRCh38
rs988426421 Chr.10: 119652122-119652122 on Build GRCh38
rs375604400 Chr.10: 119651891-119651891 on Build GRCh38
rs727504938 Chr.10: 119651868-119651868 on Build GRCh38
rs141858419 Chr.10: 119651849-119651849 on Build GRCh38
rs751595824 Chr.10: 119651785-119651785 on Build GRCh38
rs759915726 Chr.10: 119651770-119651770 on Build GRCh38
rs727502896 Chr.10: 119651730-119651730 on Build GRCh38
rs1024599221 Chr.10: 119651924-119651924 on Build GRCh38
rs990738396 Chr.10: 119651781-119651782 on Build GRCh38
rs112168846 Chr.10: 119652119-119652119 on Build GRCh38
rs747846089 Chr.10: 119651742-119651742 on Build GRCh38
rs768378511 Chr.10: 119651718-119651718 on Build GRCh38
rs866492709 Chr.10: 119651946-119651946 on Build GRCh38
rs771609568 Chr.10: 119651746-119651746 on Build GRCh38
rs535344112 Chr.10: 119651753-119651753 on Build GRCh38
rs982823685 Chr.10: 119652021-119652021 on Build GRCh38
rs763170023 Chr.10: 119651851-119651851 on Build GRCh38
rs766796091 Chr.10: 119651763-119651763 on Build GRCh38
rs866794627 Chr.10: 119652035-119652035 on Build GRCh38
rs771633629 Chr.10: 119651811-119651811 on Build GRCh38
rs746527768 Chr.10: 119651826-119651826 on Build GRCh38
rs747820097 Chr.10: 119651808-119651808 on Build GRCh38
rs372083121 Chr.10: 119651771-119651771 on Build GRCh38
rs557837780 Chr.10: 119651870-119651870 on Build GRCh38
rs899449396 Chr.10: 119651953-119651953 on Build GRCh38
rs727505283 Chr.10: 119651775-119651782 on Build GRCh38
rs745806982 Chr.10: 119651693-119651693 on Build GRCh38
rs1003238782 Chr.10: 119651928-119651929 on Build GRCh38
rs397516880 Chr.10: 119651786-119651786 on Build GRCh38
rs548800649 Chr.10: 119651768-119651768 on Build GRCh38
rs900576454 Chr.10: 119652094-119652094 on Build GRCh38
rs1022143849 Chr.10: 119651956-119651956 on Build GRCh38
rs2736421 Chr.10: 119651937-119651937 on Build GRCh38
rs922576649 Chr.10: 119651987-119651987 on Build GRCh38
rs777100532 Chr.10: 119651842-119651842 on Build GRCh38
rs752436149 Chr.10: 119651881-119651881 on Build GRCh38
rs367917821 Chr.10: 119651709-119651709 on Build GRCh38
rs915994252 Chr.10: 119652072-119652072 on Build GRCh38
rs937701647 Chr.10: 119651912-119651912 on Build GRCh38
rs929544723 Chr.10: 119652020-119652020 on Build GRCh38
rs956461511 Chr.10: 119651934-119651934 on Build GRCh38
rs727502897 Chr.10: 119651747-119651747 on Build GRCh38
rs771379373 Chr.10: 119651828-119651828 on Build GRCh38
rs561298142 Chr.10: 119651875-119651875 on Build GRCh38
rs1000232601 Chr.10: 119652096-119652096 on Build GRCh38
rs1009005365 Chr.10: 119652071-119652071 on Build GRCh38
rs751609529 Chr.10: 119651867-119651867 on Build GRCh38

View Full Product Details