Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121743335-121743870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATE1
Entrez Gene ID
11101
Gene Name
arginyltransferase 1
Gene Aliases
-
UniGene
Hs.632080
Gene Chromosome Location
Chr.10: 121740421-121928801 on Build GRCh38
Transcripts
NM_001001976.2 NM_001288734.1 NM_001288735.1 NM_001288736.1 NM_007041.3 NR_110006.1 XM_005269458.2 XM_005269459.3 XM_017015597.1 XM_017015598.1 XM_017015599.1 XM_017015600.1 XM_017015602.1 XM_017015604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6189527 c.1524G>T p.K508N Chr.10: 121743713-121743713 on Build GRCh38
4760894 c.1396A>G p.T466A Chr.10: 121743841-121743841 on Build GRCh38
4166695 c.1447G>A p.G483S Chr.10: 121743790-121743790 on Build GRCh38
1736448 c.1428G>C p.K476N Chr.10: 121743809-121743809 on Build GRCh38
2056360 c.1398G>A p.T466T Chr.10: 121743839-121743839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778484084 Chr.10: 121743661-121743661 on Build GRCh38
rs746264073 Chr.10: 121743784-121743784 on Build GRCh38
rs773221487 Chr.10: 121743702-121743702 on Build GRCh38
rs764849374 Chr.10: 121743725-121743725 on Build GRCh38
rs752601356 Chr.10: 121743733-121743733 on Build GRCh38
rs548673018 Chr.10: 121743499-121743500 on Build GRCh38
rs920815494 Chr.10: 121743627-121743627 on Build GRCh38
rs201116177 Chr.10: 121743677-121743677 on Build GRCh38
rs555194800 Chr.10: 121743836-121743836 on Build GRCh38
rs749513221 Chr.10: 121743846-121743846 on Build GRCh38
rs979329477 Chr.10: 121743647-121743647 on Build GRCh38
rs373910135 Chr.10: 121743789-121743789 on Build GRCh38
rs765112117 Chr.10: 121743806-121743806 on Build GRCh38
rs377706344 Chr.10: 121743622-121743622 on Build GRCh38
rs1020566103 Chr.10: 121743563-121743567 on Build GRCh38
rs1049349908 Chr.10: 121743785-121743785 on Build GRCh38
rs539545051 Chr.10: 121743549-121743549 on Build GRCh38
rs778204758 Chr.10: 121743668-121743668 on Build GRCh38
rs757825450 Chr.10: 121743743-121743743 on Build GRCh38
rs10886973 Chr.10: 121743672-121743672 on Build GRCh38
rs780094821 Chr.10: 121743788-121743788 on Build GRCh38
rs568741703 Chr.10: 121743375-121743375 on Build GRCh38
rs774946938 Chr.10: 121743807-121743807 on Build GRCh38
rs1010933453 Chr.10: 121743755-121743755 on Build GRCh38
rs754648790 Chr.10: 121743660-121743660 on Build GRCh38
rs763588722 Chr.10: 121743812-121743812 on Build GRCh38
rs1036622116 Chr.10: 121743517-121743517 on Build GRCh38
rs1002752246 Chr.10: 121743489-121743489 on Build GRCh38
rs746096831 Chr.10: 121743673-121743673 on Build GRCh38
rs756520822 Chr.10: 121743787-121743787 on Build GRCh38
rs200562528 Chr.10: 121743839-121743839 on Build GRCh38
rs137975321 Chr.10: 121743757-121743757 on Build GRCh38
rs7919858 Chr.10: 121743456-121743456 on Build GRCh38
rs982308754 Chr.10: 121743467-121743467 on Build GRCh38
rs916690220 Chr.10: 121743450-121743450 on Build GRCh38
rs371199945 Chr.10: 121743651-121743651 on Build GRCh38
rs541588284 Chr.10: 121743730-121743730 on Build GRCh38
rs575175908 Chr.10: 121743795-121743795 on Build GRCh38
rs770816162 Chr.10: 121743797-121743797 on Build GRCh38
rs919278695 Chr.10: 121743751-121743751 on Build GRCh38
rs1009223315 Chr.10: 121743521-121743521 on Build GRCh38
rs760706352 Chr.10: 121743703-121743703 on Build GRCh38
rs953905562 Chr.10: 121743655-121743655 on Build GRCh38
rs780484585 Chr.10: 121743843-121743843 on Build GRCh38
rs755320961 Chr.10: 121743847-121743847 on Build GRCh38
rs753476989 Chr.10: 121743658-121743658 on Build GRCh38
rs752048714 Chr.10: 121743734-121743734 on Build GRCh38
rs1219497 Chr.10: 121743417-121743417 on Build GRCh38
rs138585613 Chr.10: 121743790-121743790 on Build GRCh38
rs752218324 Chr.10: 121743663-121743663 on Build GRCh38
rs767609368 Chr.10: 121743630-121743630 on Build GRCh38
rs749888796 Chr.10: 121743477-121743477 on Build GRCh38
rs752416555 Chr.10: 121743729-121743729 on Build GRCh38
rs759197284 Chr.10: 121743719-121743719 on Build GRCh38
rs188545293 Chr.10: 121743438-121743438 on Build GRCh38
rs1036354762 Chr.10: 121743384-121743384 on Build GRCh38
rs148181636 Chr.10: 121743840-121743840 on Build GRCh38
rs377482651 Chr.10: 121743643-121743643 on Build GRCh38
rs143026183 Chr.10: 121743824-121743824 on Build GRCh38
rs201143870 Chr.10: 121743664-121743664 on Build GRCh38
rs375095340 Chr.10: 121743652-121743652 on Build GRCh38
rs751207983 Chr.10: 121743581-121743581 on Build GRCh38
rs149469002 Chr.10: 121743791-121743791 on Build GRCh38
rs776466464 Chr.10: 121743799-121743799 on Build GRCh38
rs767860330 Chr.10: 121743838-121743838 on Build GRCh38
rs940797932 Chr.10: 121743507-121743507 on Build GRCh38
rs762888773 Chr.10: 121743695-121743695 on Build GRCh38
rs544531894 Chr.10: 121743802-121743802 on Build GRCh38
rs753645736 Chr.10: 121743707-121743707 on Build GRCh38
rs768884904 Chr.10: 121743701-121743701 on Build GRCh38
rs958173314 Chr.10: 121743745-121743745 on Build GRCh38
rs4751850 Chr.10: 121743481-121743481 on Build GRCh38
rs944761427 Chr.10: 121743376-121743376 on Build GRCh38
rs757737187 Chr.10: 121743828-121743828 on Build GRCh38
rs747518564 Chr.10: 121743670-121743670 on Build GRCh38
rs146965571 Chr.10: 121743706-121743706 on Build GRCh38
rs905375901 Chr.10: 121743480-121743480 on Build GRCh38
rs928113386 Chr.10: 121743473-121743473 on Build GRCh38
rs775615068 Chr.10: 121743690-121743690 on Build GRCh38
rs1036389210 Chr.10: 121743714-121743714 on Build GRCh38

View Full Product Details