Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121790088-121790592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATE1
Entrez Gene ID
11101
Gene Name
arginyltransferase 1
Gene Aliases
-
UniGene
Hs.632080
Gene Chromosome Location
Chr.10: 121740421-121928801 on Build GRCh38
Transcripts
NM_001001976.2 NM_001288734.1 NM_001288735.1 NM_001288736.1 NM_007041.3 NR_110006.1 XM_005269458.2 XM_005269459.3 XM_017015597.1 XM_017015598.1 XM_017015599.1 XM_017015600.1 XM_017015602.1 XM_017015604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5700649 c.1378G>C p.V460L Chr.10: 121790169-121790169 on Build GRCh38
4663960 c.1307T>C p.V436A Chr.10: 121790240-121790240 on Build GRCh38
4885433 c.1345A>T p.K449* Chr.10: 121790202-121790202 on Build GRCh38
3751574 c.1371A>C p.P457P Chr.10: 121790176-121790176 on Build GRCh38
4844949 c.1276G>A p.D426N Chr.10: 121790271-121790271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953145184 Chr.10: 121790511-121790511 on Build GRCh38
rs771473936 Chr.10: 121790218-121790218 on Build GRCh38
rs1014782979 Chr.10: 121790301-121790301 on Build GRCh38
rs774873908 Chr.10: 121790409-121790409 on Build GRCh38
rs767045204 Chr.10: 121790235-121790235 on Build GRCh38
rs777923775 Chr.10: 121790183-121790183 on Build GRCh38
rs577854964 Chr.10: 121790541-121790541 on Build GRCh38
rs756705389 Chr.10: 121790156-121790156 on Build GRCh38
rs768938258 Chr.10: 121790195-121790195 on Build GRCh38
rs755232792 Chr.10: 121790175-121790175 on Build GRCh38
rs745788219 Chr.10: 121790190-121790190 on Build GRCh38
rs186754318 Chr.10: 121790567-121790567 on Build GRCh38
rs762282671 Chr.10: 121790429-121790429 on Build GRCh38
rs750834320 Chr.10: 121790147-121790147 on Build GRCh38
rs546961740 Chr.10: 121790162-121790162 on Build GRCh38
rs551602269 Chr.10: 121790418-121790418 on Build GRCh38
rs369494461 Chr.10: 121790309-121790309 on Build GRCh38
rs553630664 Chr.10: 121790510-121790510 on Build GRCh38
rs761270895 Chr.10: 121790136-121790136 on Build GRCh38
rs575222726 Chr.10: 121790320-121790320 on Build GRCh38
rs887123896 Chr.10: 121790350-121790350 on Build GRCh38
rs761057499 Chr.10: 121790234-121790234 on Build GRCh38
rs754297760 Chr.10: 121790164-121790164 on Build GRCh38
rs533720619 Chr.10: 121790113-121790113 on Build GRCh38
rs754420430 Chr.10: 121790236-121790236 on Build GRCh38
rs975607388 Chr.10: 121790485-121790485 on Build GRCh38
rs758712943 Chr.10: 121790281-121790281 on Build GRCh38
rs181695719 Chr.10: 121790263-121790263 on Build GRCh38
rs1014721510 Chr.10: 121790395-121790395 on Build GRCh38
rs769808653 Chr.10: 121790193-121790193 on Build GRCh38
rs987195741 Chr.10: 121790523-121790523 on Build GRCh38
rs759941433 Chr.10: 121790247-121790247 on Build GRCh38
rs981111010 Chr.10: 121790508-121790508 on Build GRCh38
rs773430386 Chr.10: 121790321-121790323 on Build GRCh38
rs779924356 Chr.10: 121790322-121790322 on Build GRCh38
rs1017207005 Chr.10: 121790416-121790416 on Build GRCh38
rs749030906 Chr.10: 121790202-121790202 on Build GRCh38
rs571348327 Chr.10: 121790441-121790441 on Build GRCh38
rs774012038 Chr.10: 121790123-121790123 on Build GRCh38
rs752910094 Chr.10: 121790274-121790274 on Build GRCh38
rs189114692 Chr.10: 121790223-121790223 on Build GRCh38
rs768468088 Chr.10: 121790204-121790204 on Build GRCh38
rs925887426 Chr.10: 121790562-121790562 on Build GRCh38
rs35350755 Chr.10: 121790176-121790176 on Build GRCh38
rs202154262 Chr.10: 121790115-121790115 on Build GRCh38
rs994552658 Chr.10: 121790443-121790443 on Build GRCh38
rs777683373 Chr.10: 121790287-121790287 on Build GRCh38
rs749226066 Chr.10: 121790340-121790340 on Build GRCh38
rs767054130 Chr.10: 121790144-121790144 on Build GRCh38
rs534132268 Chr.10: 121790500-121790500 on Build GRCh38
rs369368805 Chr.10: 121790553-121790553 on Build GRCh38
rs977089136 Chr.10: 121790545-121790545 on Build GRCh38
rs150821970 Chr.10: 121790534-121790534 on Build GRCh38
rs146262449 Chr.10: 121790215-121790215 on Build GRCh38
rs374481978 Chr.10: 121790563-121790563 on Build GRCh38
rs1028935045 Chr.10: 121790501-121790501 on Build GRCh38
rs142354950 Chr.10: 121790533-121790533 on Build GRCh38
rs201984922 Chr.10: 121790207-121790207 on Build GRCh38

View Full Product Details