Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 121913598-121913997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATE1
Entrez Gene ID
11101
Gene Name
arginyltransferase 1
Gene Aliases
-
UniGene
Hs.632080
Gene Chromosome Location
Chr.10: 121740421-121928801 on Build GRCh38
Transcripts
NM_001001976.2 NM_001288734.1 NM_001288735.1 NM_001288736.1 NM_007041.3 NR_110006.1 XM_005269458.2 XM_005269459.3 XM_017015597.1 XM_017015598.1 XM_017015599.1 XM_017015600.1 XM_017015601.1 XM_017015602.1 XM_017015603.1 XM_017015604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4185893 c.289A>G p.M97V Chr.10: 121913838-121913838 on Build GRCh38
415143 c.268C>G p.H90D Chr.10: 121913859-121913859 on Build GRCh38
1179439 c.238C>T p.R80* Chr.10: 121913889-121913889 on Build GRCh38
4482476 c.260C>T p.S87L Chr.10: 121913867-121913867 on Build GRCh38
415142 c.268C>G p.H90D Chr.10: 121913859-121913859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917163362 Chr.10: 121913683-121913683 on Build GRCh38
rs776337589 Chr.10: 121913754-121913754 on Build GRCh38
rs998449974 Chr.10: 121913748-121913748 on Build GRCh38
rs533064066 Chr.10: 121913771-121913771 on Build GRCh38
rs759395648 Chr.10: 121913937-121913937 on Build GRCh38
rs534935836 Chr.10: 121913956-121913956 on Build GRCh38
rs182948311 Chr.10: 121913757-121913757 on Build GRCh38
rs991317528 Chr.10: 121913674-121913674 on Build GRCh38
rs939820516 Chr.10: 121913795-121913795 on Build GRCh38
rs1025656406 Chr.10: 121913642-121913642 on Build GRCh38
rs977023180 Chr.10: 121913689-121913689 on Build GRCh38
rs7092663 Chr.10: 121913651-121913651 on Build GRCh38
rs765777434 Chr.10: 121913767-121913767 on Build GRCh38
rs371143220 Chr.10: 121913831-121913831 on Build GRCh38
rs760223886 Chr.10: 121913927-121913927 on Build GRCh38
rs570324689 Chr.10: 121913765-121913765 on Build GRCh38
rs188695815 Chr.10: 121913667-121913667 on Build GRCh38
rs751728733 Chr.10: 121913837-121913837 on Build GRCh38
rs10749435 Chr.10: 121913824-121913824 on Build GRCh38
rs11553149 Chr.10: 121913845-121913845 on Build GRCh38
rs751570178 Chr.10: 121913742-121913742 on Build GRCh38
rs770570939 Chr.10: 121913935-121913935 on Build GRCh38
rs973889445 Chr.10: 121913798-121913798 on Build GRCh38
rs891154950 Chr.10: 121913922-121913922 on Build GRCh38
rs1001035733 Chr.10: 121913680-121913680 on Build GRCh38
rs190644303 Chr.10: 121913746-121913746 on Build GRCh38
rs373058397 Chr.10: 121913878-121913878 on Build GRCh38
rs745899377 Chr.10: 121913889-121913889 on Build GRCh38
rs910335921 Chr.10: 121913731-121913731 on Build GRCh38
rs749212432 Chr.10: 121913743-121913743 on Build GRCh38
rs376433060 Chr.10: 121913766-121913766 on Build GRCh38
rs761634106 Chr.10: 121913756-121913756 on Build GRCh38
rs1017428415 Chr.10: 121913776-121913776 on Build GRCh38
rs1049743954 Chr.10: 121913644-121913644 on Build GRCh38
rs150419506 Chr.10: 121913655-121913655 on Build GRCh38
rs187192255 Chr.10: 121913808-121913808 on Build GRCh38
rs958859221 Chr.10: 121913797-121913797 on Build GRCh38
rs769994478 Chr.10: 121913898-121913898 on Build GRCh38
rs112026227 Chr.10: 121913687-121913687 on Build GRCh38
rs11200251 Chr.10: 121913691-121913691 on Build GRCh38
rs545941516 Chr.10: 121913664-121913664 on Build GRCh38
rs577287001 Chr.10: 121913660-121913660 on Build GRCh38
rs368040838 Chr.10: 121913902-121913902 on Build GRCh38
rs924580452 Chr.10: 121913940-121913940 on Build GRCh38
rs932760063 Chr.10: 121913872-121913872 on Build GRCh38
rs1021114068 Chr.10: 121913666-121913666 on Build GRCh38
rs771758054 Chr.10: 121913903-121913903 on Build GRCh38
rs765694044 Chr.10: 121913804-121913804 on Build GRCh38
rs575519403 Chr.10: 121913676-121913678 on Build GRCh38
rs935741225 Chr.10: 121913708-121913708 on Build GRCh38
rs757544917 Chr.10: 121913844-121913844 on Build GRCh38
rs74512463 Chr.10: 121913744-121913744 on Build GRCh38
rs773098055 Chr.10: 121913917-121913917 on Build GRCh38

View Full Product Details