Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122271199-122271702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD16
Entrez Gene ID
118663
Gene Name
BTB domain containing 16
Gene Aliases
C10orf87
UniGene
Hs.422466
Gene Chromosome Location
Chr.10: 122270745-122338180 on Build GRCh38
Transcripts
NM_001318189.1 NM_144587.3 XM_011539239.2 XM_011539240.2 XM_011539241.2 XM_011539242.2

SNP Detail

SNP ID Chromosome Location
rs902658292 Chr.10: 122271399-122271399 on Build GRCh38
rs1042494632 Chr.10: 122271599-122271599 on Build GRCh38
rs79732505 Chr.10: 122271432-122271432 on Build GRCh38
rs528983294 Chr.10: 122271418-122271418 on Build GRCh38
rs531378485 Chr.10: 122271502-122271502 on Build GRCh38
rs751455694 Chr.10: 122271485-122271485 on Build GRCh38
rs34479636 Chr.10: 122271276-122271277 on Build GRCh38
rs910000853 Chr.10: 122271383-122271383 on Build GRCh38
rs535548831 Chr.10: 122271646-122271646 on Build GRCh38
rs1005353109 Chr.10: 122271513-122271513 on Build GRCh38
rs1046699791 Chr.10: 122271649-122271649 on Build GRCh38
rs7069328 Chr.10: 122271226-122271226 on Build GRCh38
rs546773953 Chr.10: 122271620-122271620 on Build GRCh38
rs1042154548 Chr.10: 122271384-122271384 on Build GRCh38
rs1018491292 Chr.10: 122271308-122271308 on Build GRCh38
rs61258522 Chr.10: 122271366-122271366 on Build GRCh38
rs966034760 Chr.10: 122271341-122271341 on Build GRCh38
rs981282962 Chr.10: 122271297-122271297 on Build GRCh38
rs973966707 Chr.10: 122271524-122271524 on Build GRCh38
rs919842249 Chr.10: 122271525-122271525 on Build GRCh38
rs564412468 Chr.10: 122271317-122271317 on Build GRCh38
rs970728061 Chr.10: 122271484-122271484 on Build GRCh38
rs1032615162 Chr.10: 122271255-122271255 on Build GRCh38
rs570047019 Chr.10: 122271377-122271377 on Build GRCh38
rs965771021 Chr.10: 122271619-122271619 on Build GRCh38
rs895709013 Chr.10: 122271511-122271511 on Build GRCh38
rs1001498986 Chr.10: 122271251-122271251 on Build GRCh38
rs970823026 Chr.10: 122271286-122271286 on Build GRCh38
rs544167241 Chr.10: 122271233-122271233 on Build GRCh38
rs1001045324 Chr.10: 122271409-122271409 on Build GRCh38
rs939887083 Chr.10: 122271339-122271339 on Build GRCh38
rs144548599 Chr.10: 122271607-122271607 on Build GRCh38
rs959322235 Chr.10: 122271258-122271258 on Build GRCh38
rs577720527 Chr.10: 122271343-122271343 on Build GRCh38
rs990852029 Chr.10: 122271271-122271271 on Build GRCh38
rs566749772 Chr.10: 122271645-122271645 on Build GRCh38
rs903022575 Chr.10: 122271565-122271565 on Build GRCh38
rs529462793 Chr.10: 122271664-122271664 on Build GRCh38
rs918747574 Chr.10: 122271359-122271359 on Build GRCh38
rs1012233849 Chr.10: 122271468-122271468 on Build GRCh38
rs953988158 Chr.10: 122271568-122271568 on Build GRCh38
rs1041640216 Chr.10: 122271530-122271530 on Build GRCh38
rs1025007841 Chr.10: 122271478-122271478 on Build GRCh38
rs761402491 Chr.10: 122271641-122271641 on Build GRCh38
rs895438115 Chr.10: 122271462-122271462 on Build GRCh38
rs190513533 Chr.10: 122271591-122271591 on Build GRCh38
rs908342522 Chr.10: 122271334-122271334 on Build GRCh38
rs74161404 Chr.10: 122271573-122271573 on Build GRCh38
rs768389106 Chr.10: 122271523-122271523 on Build GRCh38
rs748867846 Chr.10: 122271472-122271472 on Build GRCh38
rs936831155 Chr.10: 122271615-122271615 on Build GRCh38
rs766142177 Chr.10: 122271681-122271681 on Build GRCh38
rs976981311 Chr.10: 122271634-122271634 on Build GRCh38
rs770275175 Chr.10: 122271381-122271381 on Build GRCh38
rs1015011371 Chr.10: 122271515-122271515 on Build GRCh38
rs1049424654 Chr.10: 122271376-122271376 on Build GRCh38
rs919850471 Chr.10: 122271363-122271363 on Build GRCh38
rs912200991 Chr.10: 122271588-122271588 on Build GRCh38
rs781767870 Chr.10: 122271281-122271281 on Build GRCh38
rs902751507 Chr.10: 122271245-122271245 on Build GRCh38

View Full Product Details