Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122573632-122574129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMBT1
Entrez Gene ID
1755
Gene Name
deleted in malignant brain tumors 1
Gene Aliases
GP340, SAG, muclin
UniGene
Hs.279611
Gene Chromosome Location
Chr.10: 122560665-122643736 on Build GRCh38
Transcripts
NM_001320644.1 NM_004406.2 NM_007329.2 NM_017579.2 XM_006717660.3 XM_006717665.3 XM_011539388.2 XM_011539389.2 XM_011539390.2 XM_011539391.2 XM_011539392.2 XM_011539393.2 XM_011539394.2 XM_011539395.2 XM_011539396.2 XM_011539398.2 XM_011539399.2 XM_011539400.2 XM_011539401.2 XM_011539402.2 XM_011539403.2 XM_011539405.2 XM_011539407.2 XM_011539408.2 XM_011539409.2 XM_011539410.2 XM_011539411.2 XM_011539413.2 XM_011539414.2 XM_011539415.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6409984 c.242T>A p.L81Q Chr.10: 122573721-122573721 on Build GRCh38
332532 c.278C>A p.A93E Chr.10: 122573757-122573757 on Build GRCh38
6300858 c.250T>A p.S84T Chr.10: 122573729-122573729 on Build GRCh38
5074087 c.283+5C>T p.? Chr.10: 122573767-122573767 on Build GRCh38
6128865 c.283+1G>A p.? Chr.10: 122573763-122573763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752795958 Chr.10: 122573833-122573833 on Build GRCh38
rs374018592 Chr.10: 122573670-122573670 on Build GRCh38
rs78241404 Chr.10: 122573830-122573830 on Build GRCh38
rs746399168 Chr.10: 122573688-122573688 on Build GRCh38
rs995773343 Chr.10: 122574070-122574070 on Build GRCh38
rs774667838 Chr.10: 122573968-122573968 on Build GRCh38
rs765879961 Chr.10: 122574018-122574018 on Build GRCh38
rs906986868 Chr.10: 122574054-122574054 on Build GRCh38
rs371052369 Chr.10: 122573840-122573840 on Build GRCh38
rs144485254 Chr.10: 122573682-122573682 on Build GRCh38
rs747860291 Chr.10: 122573956-122573956 on Build GRCh38
rs780288065 Chr.10: 122573694-122573694 on Build GRCh38
rs141625742 Chr.10: 122573781-122573781 on Build GRCh38
rs776744023 Chr.10: 122573666-122573677 on Build GRCh38
rs1038767497 Chr.10: 122573978-122573978 on Build GRCh38
rs760074122 Chr.10: 122573973-122573973 on Build GRCh38
rs756275346 Chr.10: 122573834-122573834 on Build GRCh38
rs183419973 Chr.10: 122573927-122573927 on Build GRCh38
rs192475474 Chr.10: 122573876-122573876 on Build GRCh38
rs942938620 Chr.10: 122573918-122573918 on Build GRCh38
rs773229656 Chr.10: 122573727-122573727 on Build GRCh38
rs187793854 Chr.10: 122574010-122574010 on Build GRCh38
rs200322788 Chr.10: 122573716-122573716 on Build GRCh38
rs764966265 Chr.10: 122573671-122573671 on Build GRCh38
rs780808484 Chr.10: 122573685-122573685 on Build GRCh38
rs556122762 Chr.10: 122573958-122573958 on Build GRCh38
rs1015781596 Chr.10: 122573841-122573841 on Build GRCh38
rs915535168 Chr.10: 122573899-122573899 on Build GRCh38
rs772226024 Chr.10: 122573810-122573810 on Build GRCh38
rs115554496 Chr.10: 122573790-122573790 on Build GRCh38
rs961831946 Chr.10: 122573662-122573662 on Build GRCh38
rs934629364 Chr.10: 122573997-122573997 on Build GRCh38
rs753817108 Chr.10: 122573668-122573668 on Build GRCh38
rs762851442 Chr.10: 122573760-122573760 on Build GRCh38
rs934125763 Chr.10: 122573957-122573957 on Build GRCh38
rs748395172 Chr.10: 122573808-122573808 on Build GRCh38
rs142815957 Chr.10: 122573672-122573672 on Build GRCh38
rs903128623 Chr.10: 122573769-122573769 on Build GRCh38
rs887180075 Chr.10: 122573966-122573966 on Build GRCh38
rs975629166 Chr.10: 122573936-122573936 on Build GRCh38
rs922769792 Chr.10: 122573949-122573949 on Build GRCh38
rs1044112579 Chr.10: 122574039-122574039 on Build GRCh38
rs765283729 Chr.10: 122573740-122573740 on Build GRCh38
rs775274238 Chr.10: 122573759-122573759 on Build GRCh38
rs906892147 Chr.10: 122573816-122573816 on Build GRCh38
rs774606331 Chr.10: 122573708-122573708 on Build GRCh38
rs974642425 Chr.10: 122573916-122573916 on Build GRCh38
rs776862468 Chr.10: 122573965-122573965 on Build GRCh38
rs1054657807 Chr.10: 122573999-122573999 on Build GRCh38
rs954248227 Chr.10: 122574078-122574078 on Build GRCh38
rs756066037 Chr.10: 122573772-122573772 on Build GRCh38
rs556349374 Chr.10: 122573659-122573659 on Build GRCh38
rs113418659 Chr.10: 122574071-122574071 on Build GRCh38
rs148017393 Chr.10: 122573711-122573711 on Build GRCh38
rs767566236 Chr.10: 122573667-122573667 on Build GRCh38
rs2277235 Chr.10: 122573792-122573792 on Build GRCh38
rs755415245 Chr.10: 122573793-122573793 on Build GRCh38
rs549536495 Chr.10: 122573920-122573920 on Build GRCh38
rs759408245 Chr.10: 122573731-122573731 on Build GRCh38
rs751229971 Chr.10: 122573681-122573681 on Build GRCh38
rs1017696849 Chr.10: 122574075-122574075 on Build GRCh38
rs948446835 Chr.10: 122574021-122574021 on Build GRCh38
rs779212979 Chr.10: 122573794-122573794 on Build GRCh38
rs150522937 Chr.10: 122574085-122574085 on Build GRCh38
rs756919017 Chr.10: 122573768-122573768 on Build GRCh38
rs76627533 Chr.10: 122574065-122574065 on Build GRCh38
rs763357596 Chr.10: 122574066-122574066 on Build GRCh38
rs763894212 Chr.10: 122573674-122573674 on Build GRCh38
rs1005940271 Chr.10: 122573755-122573755 on Build GRCh38
rs538479333 Chr.10: 122573979-122573979 on Build GRCh38
rs187544343 Chr.10: 122573691-122573691 on Build GRCh38
rs540634398 Chr.10: 122573707-122573707 on Build GRCh38
rs749729999 Chr.10: 122573702-122573702 on Build GRCh38
rs897369174 Chr.10: 122574069-122574069 on Build GRCh38
rs367761326 Chr.10: 122573903-122573903 on Build GRCh38
rs759728909 Chr.10: 122573800-122573800 on Build GRCh38
rs547437585 Chr.10: 122573967-122573967 on Build GRCh38
rs1024151452 Chr.10: 122574044-122574044 on Build GRCh38
rs551877365 Chr.10: 122573897-122573897 on Build GRCh38
rs376363880 Chr.10: 122573811-122573811 on Build GRCh38
rs571125124 Chr.10: 122573923-122573923 on Build GRCh38
rs780641214 Chr.10: 122573857-122573857 on Build GRCh38
rs530064249 Chr.10: 122574055-122574055 on Build GRCh38
rs772174631 Chr.10: 122573712-122573712 on Build GRCh38
rs75236604 Chr.10: 122573861-122573861 on Build GRCh38
rs538767094 Chr.10: 122573673-122573673 on Build GRCh38

View Full Product Details