Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122832093-122832598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUZD1
Entrez Gene ID
50624
Gene Name
CUB and zona pellucida like domains 1
Gene Aliases
ERG-1, ERG1, ITMAP1, UO-44
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122846175 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3435333 c.1676A>G p.E559G Chr.10: 122832426-122832426 on Build GRCh38
1202714 c.1802A>G p.Y601C Chr.10: 122832300-122832300 on Build GRCh38
4675662 c.1787G>A p.R596Q Chr.10: 122832315-122832315 on Build GRCh38
3435332 c.1766T>G p.V589G Chr.10: 122832336-122832336 on Build GRCh38
2020792 c.1733C>A p.A578D Chr.10: 122832369-122832369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753825900 Chr.10: 122832384-122832384 on Build GRCh38
rs539790743 Chr.10: 122832145-122832145 on Build GRCh38
rs995566035 Chr.10: 122832193-122832193 on Build GRCh38
rs762017332 Chr.10: 122832352-122832352 on Build GRCh38
rs187639528 Chr.10: 122832216-122832216 on Build GRCh38
rs956329267 Chr.10: 122832477-122832477 on Build GRCh38
rs143224907 Chr.10: 122832343-122832343 on Build GRCh38
rs570837090 Chr.10: 122832140-122832140 on Build GRCh38
rs774402353 Chr.10: 122832347-122832347 on Build GRCh38
rs949163760 Chr.10: 122832205-122832205 on Build GRCh38
rs763527312 Chr.10: 122832398-122832398 on Build GRCh38
rs777382719 Chr.10: 122832313-122832313 on Build GRCh38
rs189938140 Chr.10: 122832290-122832290 on Build GRCh38
rs367765206 Chr.10: 122832254-122832254 on Build GRCh38
rs199652779 Chr.10: 122832358-122832358 on Build GRCh38
rs764305148 Chr.10: 122832289-122832289 on Build GRCh38
rs760014426 Chr.10: 122832276-122832276 on Build GRCh38
rs770291640 Chr.10: 122832337-122832337 on Build GRCh38
rs112129106 Chr.10: 122832338-122832338 on Build GRCh38
rs61730786 Chr.10: 122832441-122832441 on Build GRCh38
rs780504501 Chr.10: 122832479-122832479 on Build GRCh38
rs779866817 Chr.10: 122832353-122832353 on Build GRCh38
rs917582603 Chr.10: 122832183-122832183 on Build GRCh38
rs182694427 Chr.10: 122832125-122832125 on Build GRCh38
rs961767913 Chr.10: 122832186-122832186 on Build GRCh38
rs150722442 Chr.10: 122832428-122832428 on Build GRCh38
rs758158635 Chr.10: 122832431-122832431 on Build GRCh38
rs746053733 Chr.10: 122832471-122832471 on Build GRCh38
rs756807763 Chr.10: 122832469-122832469 on Build GRCh38
rs753711457 Chr.10: 122832298-122832298 on Build GRCh38
rs757997185 Chr.10: 122832309-122832309 on Build GRCh38
rs778621800 Chr.10: 122832299-122832299 on Build GRCh38
rs539803177 Chr.10: 122832498-122832498 on Build GRCh38
rs755934811 Chr.10: 122832246-122832246 on Build GRCh38
rs907477751 Chr.10: 122832466-122832466 on Build GRCh38
rs905040061 Chr.10: 122832259-122832259 on Build GRCh38
rs771583975 Chr.10: 122832239-122832239 on Build GRCh38
rs924437049 Chr.10: 122832329-122832329 on Build GRCh38
rs374600302 Chr.10: 122832340-122832340 on Build GRCh38
rs763205663 Chr.10: 122832287-122832287 on Build GRCh38
rs1015732863 Chr.10: 122832126-122832126 on Build GRCh38
rs139868556 Chr.10: 122832316-122832316 on Build GRCh38
rs776974640 Chr.10: 122832265-122832265 on Build GRCh38
rs772641813 Chr.10: 122832244-122832244 on Build GRCh38
rs747802150 Chr.10: 122832300-122832300 on Build GRCh38
rs201620290 Chr.10: 122832443-122832443 on Build GRCh38
rs537927731 Chr.10: 122832429-122832429 on Build GRCh38
rs759400818 Chr.10: 122832399-122832399 on Build GRCh38
rs1027440061 Chr.10: 122832212-122832212 on Build GRCh38
rs200173778 Chr.10: 122832315-122832315 on Build GRCh38
rs765686672 Chr.10: 122832277-122832277 on Build GRCh38
rs145984488 Chr.10: 122832292-122832292 on Build GRCh38
rs771517753 Chr.10: 122832258-122832258 on Build GRCh38
rs374469698 Chr.10: 122832227-122832227 on Build GRCh38
rs746422142 Chr.10: 122832491-122832491 on Build GRCh38
rs555509336 Chr.10: 122832317-122832317 on Build GRCh38
rs372686495 Chr.10: 122832327-122832327 on Build GRCh38
rs775871596 Chr.10: 122832283-122832283 on Build GRCh38
rs948370801 Chr.10: 122832348-122832348 on Build GRCh38
rs182527423 Chr.10: 122832356-122832356 on Build GRCh38

View Full Product Details