Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123050963-123051465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADSB
Entrez Gene ID
36
Gene Name
acyl-CoA dehydrogenase, short/branched chain
Gene Aliases
2-MEBCAD, ACAD7, SBCAD
UniGene
Hs.81934
Gene Chromosome Location
Chr.10: 123008913-123058290 on Build GRCh38
Transcripts
NM_001609.3 XM_017016194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5661202 c.1128+3_1128+7delAAAAA p.? Chr.10: 123051189-123051193 on Build GRCh38
7414813 c.1066C>A p.L356I Chr.10: 123051124-123051124 on Build GRCh38
6591697 c.1062T>C p.A354A Chr.10: 123051120-123051120 on Build GRCh38
41001 c.1043T>G p.L348* Chr.10: 123051101-123051101 on Build GRCh38
6591703 c.1100C>T p.A367V Chr.10: 123051158-123051158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781276495 Chr.10: 123051154-123051154 on Build GRCh38
rs758834353 Chr.10: 123051222-123051222 on Build GRCh38
rs546637502 Chr.10: 123050988-123050988 on Build GRCh38
rs779679250 Chr.10: 123051305-123051305 on Build GRCh38
rs984708859 Chr.10: 123051428-123051428 on Build GRCh38
rs781238146 Chr.10: 123051004-123051004 on Build GRCh38
rs779328148 Chr.10: 123051098-123051098 on Build GRCh38
rs150034487 Chr.10: 123051073-123051073 on Build GRCh38
rs765353247 Chr.10: 123051206-123051207 on Build GRCh38
rs759590509 Chr.10: 123051189-123051196 on Build GRCh38
rs749816710 Chr.10: 123051014-123051014 on Build GRCh38
rs768193051 Chr.10: 123051399-123051399 on Build GRCh38
rs10571424 Chr.10: 123051211-123051212 on Build GRCh38
rs201268218 Chr.10: 123051078-123051078 on Build GRCh38
rs773585591 Chr.10: 123051189-123051191 on Build GRCh38
rs753732388 Chr.10: 123051189-123051192 on Build GRCh38
rs988557350 Chr.10: 123051246-123051246 on Build GRCh38
rs12357783 Chr.10: 123051185-123051185 on Build GRCh38
rs761798774 Chr.10: 123051174-123051174 on Build GRCh38
rs775065448 Chr.10: 123051154-123051154 on Build GRCh38
rs927165744 Chr.10: 123051398-123051398 on Build GRCh38
rs1029629205 Chr.10: 123051037-123051037 on Build GRCh38
rs776628240 Chr.10: 123051046-123051046 on Build GRCh38
rs202176300 Chr.10: 123051017-123051017 on Build GRCh38
rs376677303 Chr.10: 123051162-123051162 on Build GRCh38
rs745376960 Chr.10: 123051004-123051005 on Build GRCh38
rs772365552 Chr.10: 123051189-123051194 on Build GRCh38
rs374470713 Chr.10: 123051022-123051022 on Build GRCh38
rs763893052 Chr.10: 123051060-123051060 on Build GRCh38
rs199895628 Chr.10: 123051221-123051221 on Build GRCh38
rs753497781 Chr.10: 123051199-123051199 on Build GRCh38
rs773468834 Chr.10: 123051042-123051042 on Build GRCh38
rs555232353 Chr.10: 123051422-123051422 on Build GRCh38
rs778102958 Chr.10: 123051123-123051123 on Build GRCh38
rs754282140 Chr.10: 123051189-123051197 on Build GRCh38
rs902318429 Chr.10: 123051120-123051120 on Build GRCh38
rs755539165 Chr.10: 123051084-123051084 on Build GRCh38
rs766873238 Chr.10: 123051188-123051189 on Build GRCh38
rs61532872 Chr.10: 123051167-123051167 on Build GRCh38
rs771062697 Chr.10: 123051043-123051043 on Build GRCh38
rs760423996 Chr.10: 123051189-123051189 on Build GRCh38
rs1049896649 Chr.10: 123051059-123051059 on Build GRCh38
rs773082568 Chr.10: 123051184-123051184 on Build GRCh38
rs759013344 Chr.10: 123051206-123051206 on Build GRCh38
rs762666946 Chr.10: 123051187-123051188 on Build GRCh38
rs937283643 Chr.10: 123051401-123051401 on Build GRCh38
rs886046780 Chr.10: 123051120-123051120 on Build GRCh38
rs971441845 Chr.10: 123051314-123051314 on Build GRCh38
rs762963935 Chr.10: 123051050-123051050 on Build GRCh38
rs751432029 Chr.10: 123051231-123051231 on Build GRCh38
rs1023410406 Chr.10: 123051317-123051317 on Build GRCh38
rs557241397 Chr.10: 123051148-123051148 on Build GRCh38
rs3833736 Chr.10: 123051230-123051231 on Build GRCh38
rs778127134 Chr.10: 123051224-123051224 on Build GRCh38
rs565672481 Chr.10: 123051010-123051010 on Build GRCh38
rs1002929952 Chr.10: 123051330-123051330 on Build GRCh38
rs981150534 Chr.10: 123051393-123051393 on Build GRCh38
rs764786805 Chr.10: 123051214-123051214 on Build GRCh38
rs745900584 Chr.10: 123051158-123051158 on Build GRCh38
rs761554120 Chr.10: 123051063-123051063 on Build GRCh38
rs769292815 Chr.10: 123051036-123051039 on Build GRCh38
rs779698379 Chr.10: 123051001-123051001 on Build GRCh38
rs779357737 Chr.10: 123051020-123051020 on Build GRCh38
rs748420411 Chr.10: 123051031-123051031 on Build GRCh38
rs748810576 Chr.10: 123051324-123051324 on Build GRCh38
rs754353574 Chr.10: 123051190-123051192 on Build GRCh38
rs77550691 Chr.10: 123051208-123051212 on Build GRCh38
rs748662471 Chr.10: 123051105-123051105 on Build GRCh38
rs536501578 Chr.10: 123051400-123051400 on Build GRCh38
rs200548567 Chr.10: 123051118-123051118 on Build GRCh38
rs57339164 Chr.10: 123051072-123051072 on Build GRCh38
rs534689734 Chr.10: 123050987-123050987 on Build GRCh38
rs974288564 Chr.10: 123051003-123051003 on Build GRCh38
rs772447595 Chr.10: 123051041-123051041 on Build GRCh38
rs145295182 Chr.10: 123051159-123051159 on Build GRCh38
rs779384817 Chr.10: 123051191-123051192 on Build GRCh38
rs751874923 Chr.10: 123051229-123051229 on Build GRCh38
rs1015694438 Chr.10: 123051380-123051380 on Build GRCh38
rs752968972 Chr.10: 123051178-123051178 on Build GRCh38
rs774205809 Chr.10: 123051160-123051160 on Build GRCh38
rs755735951 Chr.10: 123050999-123050999 on Build GRCh38
rs760982989 Chr.10: 123051189-123051195 on Build GRCh38
rs774237425 Chr.10: 123051061-123051061 on Build GRCh38
rs767398802 Chr.10: 123051064-123051064 on Build GRCh38

View Full Product Details