Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123157609-123158115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB3
Entrez Gene ID
9184
Gene Name
BUB3, mitotic checkpoint protein
Gene Aliases
BUB3L, hBUB3
UniGene
Hs.418533
Gene Chromosome Location
Chr.10: 123154244-123165370 on Build GRCh38
Transcripts
NM_001007793.2 NM_004725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027319 c.349G>A p.D117N Chr.10: 123157812-123157812 on Build GRCh38
1346529 c.401_402delTC p.Q136fs*2 Chr.10: 123157864-123157865 on Build GRCh38
243262 c.350A>G p.D117G Chr.10: 123157813-123157813 on Build GRCh38
5753429 c.356C>T p.T119I Chr.10: 123157819-123157819 on Build GRCh38
3806654 c.358G>C p.V120L Chr.10: 123157821-123157821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762926964 Chr.10: 123157898-123157898 on Build GRCh38
rs968494443 Chr.10: 123157678-123157678 on Build GRCh38
rs545060918 Chr.10: 123157999-123157999 on Build GRCh38
rs749922735 Chr.10: 123157862-123157862 on Build GRCh38
rs900640794 Chr.10: 123157653-123157653 on Build GRCh38
rs528125942 Chr.10: 123157711-123157722 on Build GRCh38
rs907995641 Chr.10: 123158028-123158028 on Build GRCh38
rs749795696 Chr.10: 123157868-123157868 on Build GRCh38
rs920313581 Chr.10: 123157692-123157692 on Build GRCh38
rs773429750 Chr.10: 123157890-123157890 on Build GRCh38
rs982390063 Chr.10: 123158032-123158032 on Build GRCh38
rs930697201 Chr.10: 123157744-123157744 on Build GRCh38
rs746168607 Chr.10: 123157702-123157702 on Build GRCh38
rs760763404 Chr.10: 123157730-123157730 on Build GRCh38
rs763864787 Chr.10: 123157752-123157752 on Build GRCh38
rs749627889 Chr.10: 123157702-123157702 on Build GRCh38
rs996388487 Chr.10: 123157666-123157666 on Build GRCh38
rs2304550 Chr.10: 123157722-123157722 on Build GRCh38
rs944723067 Chr.10: 123157781-123157781 on Build GRCh38
rs775793839 Chr.10: 123157711-123157712 on Build GRCh38
rs751389575 Chr.10: 123157906-123157906 on Build GRCh38
rs773391701 Chr.10: 123157725-123157725 on Build GRCh38
rs536855037 Chr.10: 123157917-123157917 on Build GRCh38
rs552924732 Chr.10: 123157994-123157994 on Build GRCh38
rs765103671 Chr.10: 123157736-123157736 on Build GRCh38
rs371827854 Chr.10: 123157966-123157966 on Build GRCh38
rs776015878 Chr.10: 123157693-123157693 on Build GRCh38
rs762726735 Chr.10: 123157747-123157747 on Build GRCh38
rs780428660 Chr.10: 123157867-123157867 on Build GRCh38
rs763917712 Chr.10: 123157904-123157904 on Build GRCh38
rs893845299 Chr.10: 123157938-123157938 on Build GRCh38
rs1041015830 Chr.10: 123157804-123157804 on Build GRCh38
rs557900034 Chr.10: 123157847-123157847 on Build GRCh38
rs767750435 Chr.10: 123157723-123157723 on Build GRCh38
rs146410251 Chr.10: 123157791-123157791 on Build GRCh38
rs763140348 Chr.10: 123157712-123157712 on Build GRCh38
rs201044946 Chr.10: 123157886-123157886 on Build GRCh38
rs768996460 Chr.10: 123157707-123157707 on Build GRCh38
rs944667323 Chr.10: 123157797-123157797 on Build GRCh38
rs796942021 Chr.10: 123157988-123157988 on Build GRCh38
rs980437179 Chr.10: 123157994-123157994 on Build GRCh38
rs367666323 Chr.10: 123157888-123157888 on Build GRCh38
rs992532387 Chr.10: 123157971-123157971 on Build GRCh38
rs771041322 Chr.10: 123157894-123157894 on Build GRCh38
rs970037010 Chr.10: 123157990-123157990 on Build GRCh38
rs983472038 Chr.10: 123158024-123158024 on Build GRCh38
rs1024257601 Chr.10: 123157988-123157988 on Build GRCh38
rs746428257 Chr.10: 123157681-123157681 on Build GRCh38
rs751263267 Chr.10: 123157758-123157758 on Build GRCh38
rs1035240806 Chr.10: 123158077-123158077 on Build GRCh38
rs760679689 Chr.10: 123157891-123157891 on Build GRCh38
rs770235518 Chr.10: 123157684-123157684 on Build GRCh38
rs955660931 Chr.10: 123157924-123157924 on Build GRCh38
rs886420742 Chr.10: 123157753-123157753 on Build GRCh38
rs117847680 Chr.10: 123157640-123157640 on Build GRCh38
rs774777776 Chr.10: 123157720-123157720 on Build GRCh38
rs1031103928 Chr.10: 123157937-123157937 on Build GRCh38
rs1040252213 Chr.10: 123157851-123157851 on Build GRCh38
rs549131386 Chr.10: 123157996-123157996 on Build GRCh38
rs971818569 Chr.10: 123157667-123157667 on Build GRCh38
rs555630197 Chr.10: 123157949-123157949 on Build GRCh38
rs569788197 Chr.10: 123157897-123157897 on Build GRCh38
rs901212009 Chr.10: 123157853-123157853 on Build GRCh38
rs752616479 Chr.10: 123157738-123157738 on Build GRCh38
rs1019212606 Chr.10: 123157638-123157638 on Build GRCh38
rs930494911 Chr.10: 123157701-123157701 on Build GRCh38
rs573888360 Chr.10: 123157956-123157956 on Build GRCh38
rs769197778 Chr.10: 123157871-123157871 on Build GRCh38
rs944691770 Chr.10: 123158031-123158031 on Build GRCh38
rs373148123 Chr.10: 123158001-123158001 on Build GRCh38

View Full Product Details