Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 123164895-123165441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB3
Entrez Gene ID
9184
Gene Name
BUB3, mitotic checkpoint protein
Gene Aliases
BUB3L, hBUB3
UniGene
Hs.418533
Gene Chromosome Location
Chr.10: 123154244-123165370 on Build GRCh38
Transcripts
NM_001007793.2 NM_004725.3

SNP Detail

SNP ID Chromosome Location
rs368167736 Chr.10: 123165068-123165068 on Build GRCh38
rs989112477 Chr.10: 123164926-123164926 on Build GRCh38
rs969882601 Chr.10: 123165235-123165235 on Build GRCh38
rs1036689439 Chr.10: 123164980-123164980 on Build GRCh38
rs759900033 Chr.10: 123165040-123165040 on Build GRCh38
rs538788291 Chr.10: 123164935-123164935 on Build GRCh38
rs755382311 Chr.10: 123165081-123165081 on Build GRCh38
rs543778311 Chr.10: 123165309-123165309 on Build GRCh38
rs916244291 Chr.10: 123164927-123164927 on Build GRCh38
rs34736387 Chr.10: 123164979-123164979 on Build GRCh38
rs778430582 Chr.10: 123165112-123165112 on Build GRCh38
rs944887630 Chr.10: 123164949-123164949 on Build GRCh38
rs940510360 Chr.10: 123164965-123164965 on Build GRCh38
rs558020639 Chr.10: 123165261-123165261 on Build GRCh38
rs1031266881 Chr.10: 123165226-123165226 on Build GRCh38
rs768038353 Chr.10: 123165279-123165279 on Build GRCh38
rs539299654 Chr.10: 123165227-123165227 on Build GRCh38
rs1000290886 Chr.10: 123165169-123165169 on Build GRCh38
rs14634 Chr.10: 123165073-123165073 on Build GRCh38
rs1050291401 Chr.10: 123165370-123165370 on Build GRCh38
rs372160983 Chr.10: 123165104-123165104 on Build GRCh38
rs930585723 Chr.10: 123165340-123165340 on Build GRCh38
rs911234142 Chr.10: 123165404-123165404 on Build GRCh38
rs927694682 Chr.10: 123165302-123165302 on Build GRCh38
rs1041106071 Chr.10: 123165142-123165142 on Build GRCh38
rs766299601 Chr.10: 123165079-123165079 on Build GRCh38
rs550722372 Chr.10: 123164970-123164970 on Build GRCh38
rs776894806 Chr.10: 123165033-123165033 on Build GRCh38
rs907879972 Chr.10: 123165346-123165346 on Build GRCh38
rs202101285 Chr.10: 123165019-123165019 on Build GRCh38
rs750432366 Chr.10: 123165056-123165056 on Build GRCh38
rs796211287 Chr.10: 123164969-123164969 on Build GRCh38
rs558050542 Chr.10: 123165007-123165007 on Build GRCh38
rs763193404 Chr.10: 123165050-123165050 on Build GRCh38
rs553915820 Chr.10: 123165131-123165131 on Build GRCh38
rs775715440 Chr.10: 123165047-123165047 on Build GRCh38
rs962192078 Chr.10: 123165306-123165306 on Build GRCh38
rs36090124 Chr.10: 123164969-123164970 on Build GRCh38
rs1040533623 Chr.10: 123164989-123164989 on Build GRCh38
rs771500312 Chr.10: 123165004-123165004 on Build GRCh38
rs569112571 Chr.10: 123164970-123164971 on Build GRCh38
rs756086719 Chr.10: 123165063-123165063 on Build GRCh38
rs748775104 Chr.10: 123165274-123165274 on Build GRCh38
rs3168039 Chr.10: 123165363-123165363 on Build GRCh38
rs398097262 Chr.10: 123164978-123164979 on Build GRCh38
rs766350866 Chr.10: 123165003-123165004 on Build GRCh38
rs753597737 Chr.10: 123165084-123165084 on Build GRCh38
rs573712284 Chr.10: 123165391-123165391 on Build GRCh38
rs747494124 Chr.10: 123165012-123165012 on Build GRCh38
rs767432233 Chr.10: 123165296-123165296 on Build GRCh38
rs944904179 Chr.10: 123165401-123165401 on Build GRCh38
rs188288241 Chr.10: 123165113-123165113 on Build GRCh38
rs879050212 Chr.10: 123164967-123164967 on Build GRCh38
rs1014133548 Chr.10: 123165216-123165216 on Build GRCh38
rs1007413760 Chr.10: 123165127-123165127 on Build GRCh38
rs765391664 Chr.10: 123165044-123165044 on Build GRCh38
rs771328391 Chr.10: 123165021-123165021 on Build GRCh38
rs576293332 Chr.10: 123165290-123165290 on Build GRCh38
rs999661447 Chr.10: 123165186-123165186 on Build GRCh38
rs555447529 Chr.10: 123165389-123165389 on Build GRCh38
rs397845492 Chr.10: 123164979-123164980 on Build GRCh38
rs764258230 Chr.10: 123165053-123165053 on Build GRCh38
rs571356622 Chr.10: 123164928-123164928 on Build GRCh38
rs754827243 Chr.10: 123164990-123164990 on Build GRCh38
rs982285680 Chr.10: 123165297-123165297 on Build GRCh38
rs192785062 Chr.10: 123165171-123165171 on Build GRCh38
rs920425253 Chr.10: 123165312-123165312 on Build GRCh38
rs927818048 Chr.10: 123164960-123164960 on Build GRCh38
rs536537724 Chr.10: 123165286-123165290 on Build GRCh38
rs969031525 Chr.10: 123165282-123165282 on Build GRCh38

View Full Product Details