Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124801696-124802198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175B
Entrez Gene ID
23172
Gene Name
family with sequence similarity 175 member B
Gene Aliases
ABRO1, KIAA0157
UniGene
Hs.280695
Gene Chromosome Location
Chr.10: 124801785-124836670 on Build GRCh38
Transcripts
NM_032182.3 XM_011539554.1 XM_017015986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658945 c.35C>G p.A12G Chr.10: 124801864-124801864 on Build GRCh38
7077351 c.30C>T p.F10F Chr.10: 124801859-124801859 on Build GRCh38
1745684 c.17C>A p.S6* Chr.10: 124801846-124801846 on Build GRCh38
5729107 c.19G>A p.G7S Chr.10: 124801848-124801848 on Build GRCh38
3670482 c.46C>A p.H16N Chr.10: 124801875-124801875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766328832 Chr.10: 124801944-124801944 on Build GRCh38
rs770793359 Chr.10: 124801912-124801912 on Build GRCh38
rs752052816 Chr.10: 124801779-124801779 on Build GRCh38
rs770830784 Chr.10: 124801840-124801840 on Build GRCh38
rs975626078 Chr.10: 124802018-124802018 on Build GRCh38
rs368569863 Chr.10: 124801856-124801856 on Build GRCh38
rs908300846 Chr.10: 124802117-124802117 on Build GRCh38
rs775211497 Chr.10: 124801934-124801934 on Build GRCh38
rs113642808 Chr.10: 124802054-124802054 on Build GRCh38
rs546043551 Chr.10: 124802123-124802123 on Build GRCh38
rs543064229 Chr.10: 124801901-124801901 on Build GRCh38
rs780399727 Chr.10: 124801796-124801796 on Build GRCh38
rs1028176738 Chr.10: 124802046-124802046 on Build GRCh38
rs146309339 Chr.10: 124802012-124802012 on Build GRCh38
rs141996152 Chr.10: 124801885-124801885 on Build GRCh38
rs748216897 Chr.10: 124801911-124801911 on Build GRCh38
rs780489848 Chr.10: 124801886-124801886 on Build GRCh38
rs1000872672 Chr.10: 124801929-124801929 on Build GRCh38
rs760424948 Chr.10: 124801931-124801932 on Build GRCh38
rs768021357 Chr.10: 124801874-124801874 on Build GRCh38
rs375094564 Chr.10: 124801816-124801816 on Build GRCh38
rs751004673 Chr.10: 124801863-124801863 on Build GRCh38
rs904915928 Chr.10: 124801905-124801905 on Build GRCh38
rs763611372 Chr.10: 124801862-124801862 on Build GRCh38
rs111953777 Chr.10: 124801783-124801783 on Build GRCh38
rs1038210570 Chr.10: 124801772-124801772 on Build GRCh38
rs973699994 Chr.10: 124802122-124802122 on Build GRCh38
rs761365510 Chr.10: 124801952-124801952 on Build GRCh38
rs755190063 Chr.10: 124801902-124801902 on Build GRCh38
rs1050942806 Chr.10: 124801790-124801790 on Build GRCh38
rs572730335 Chr.10: 124801854-124801854 on Build GRCh38
rs772283958 Chr.10: 124801798-124801798 on Build GRCh38
rs781415113 Chr.10: 124801785-124801785 on Build GRCh38
rs367574016 Chr.10: 124801791-124801791 on Build GRCh38
rs979518534 Chr.10: 124802064-124802064 on Build GRCh38
rs541583246 Chr.10: 124802014-124802014 on Build GRCh38
rs373425523 Chr.10: 124802010-124802010 on Build GRCh38
rs773758188 Chr.10: 124801949-124801949 on Build GRCh38
rs935575812 Chr.10: 124801812-124801812 on Build GRCh38
rs757736355 Chr.10: 124801867-124801867 on Build GRCh38
rs1025123387 Chr.10: 124801997-124801997 on Build GRCh38
rs555968180 Chr.10: 124801989-124801989 on Build GRCh38
rs890935694 Chr.10: 124802162-124802162 on Build GRCh38
rs554175572 Chr.10: 124801754-124801754 on Build GRCh38
rs772891508 Chr.10: 124801865-124801866 on Build GRCh38
rs554720157 Chr.10: 124801732-124801732 on Build GRCh38
rs988165586 Chr.10: 124802146-124802146 on Build GRCh38
rs982529547 Chr.10: 124802069-124802069 on Build GRCh38
rs769567275 Chr.10: 124801927-124801927 on Build GRCh38
rs566688527 Chr.10: 124801763-124801763 on Build GRCh38
rs534035946 Chr.10: 124801768-124801768 on Build GRCh38
rs995142665 Chr.10: 124801718-124801718 on Build GRCh38
rs375090139 Chr.10: 124801941-124801941 on Build GRCh38
rs779134441 Chr.10: 124801906-124801906 on Build GRCh38
rs576643997 Chr.10: 124802119-124802119 on Build GRCh38
rs982726747 Chr.10: 124801735-124801735 on Build GRCh38
rs992860429 Chr.10: 124801958-124801958 on Build GRCh38
rs759323884 Chr.10: 124801853-124801853 on Build GRCh38
rs527888761 Chr.10: 124802106-124802106 on Build GRCh38
rs1051480548 Chr.10: 124802147-124802147 on Build GRCh38
rs749507269 Chr.10: 124801892-124801892 on Build GRCh38
rs186519499 Chr.10: 124801721-124801721 on Build GRCh38
rs560702444 Chr.10: 124802036-124802036 on Build GRCh38
rs925282799 Chr.10: 124802065-124802065 on Build GRCh38
rs767933709 Chr.10: 124802034-124802034 on Build GRCh38
rs1033897846 Chr.10: 124801937-124801937 on Build GRCh38
rs372327539 Chr.10: 124801925-124801925 on Build GRCh38
rs762512173 Chr.10: 124801859-124801859 on Build GRCh38
rs536035972 Chr.10: 124801725-124801725 on Build GRCh38
rs746205803 Chr.10: 124801787-124801787 on Build GRCh38
rs191790856 Chr.10: 124801945-124801945 on Build GRCh38
rs953447212 Chr.10: 124802046-124802047 on Build GRCh38
rs374773441 Chr.10: 124801774-124801774 on Build GRCh38
rs576903748 Chr.10: 124801914-124801914 on Build GRCh38
rs773339009 Chr.10: 124801821-124801821 on Build GRCh38
rs564489558 Chr.10: 124802159-124802159 on Build GRCh38
rs775121709 Chr.10: 124801857-124801857 on Build GRCh38
rs371772141 Chr.10: 124801805-124801805 on Build GRCh38
rs750803687 Chr.10: 124801884-124801884 on Build GRCh38
rs932563952 Chr.10: 124802140-124802140 on Build GRCh38

View Full Product Details