Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124835901-124836438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175B
Entrez Gene ID
23172
Gene Name
family with sequence similarity 175 member B
Gene Aliases
ABRO1, KIAA0157
UniGene
Hs.280695
Gene Chromosome Location
Chr.10: 124801785-124836670 on Build GRCh38
Transcripts
NM_032182.3 XM_011539554.1 XM_017015986.1

SNP Detail

SNP ID Chromosome Location
rs550318516 Chr.10: 124836263-124836263 on Build GRCh38
rs1000669215 Chr.10: 124836344-124836344 on Build GRCh38
rs988842630 Chr.10: 124836083-124836083 on Build GRCh38
rs938051735 Chr.10: 124836316-124836316 on Build GRCh38
rs1047875152 Chr.10: 124836375-124836375 on Build GRCh38
rs981695205 Chr.10: 124836411-124836411 on Build GRCh38
rs1028811099 Chr.10: 124836006-124836006 on Build GRCh38
rs770736200 Chr.10: 124836037-124836037 on Build GRCh38
rs1029584540 Chr.10: 124836123-124836123 on Build GRCh38
rs868565389 Chr.10: 124836133-124836133 on Build GRCh38
rs918180688 Chr.10: 124836340-124836340 on Build GRCh38
rs1016598318 Chr.10: 124836069-124836069 on Build GRCh38
rs996764682 Chr.10: 124836096-124836096 on Build GRCh38
rs370854129 Chr.10: 124836062-124836062 on Build GRCh38
rs901807472 Chr.10: 124836002-124836002 on Build GRCh38
rs893257460 Chr.10: 124836366-124836366 on Build GRCh38
rs564703763 Chr.10: 124836177-124836177 on Build GRCh38
rs139492992 Chr.10: 124836330-124836330 on Build GRCh38
rs1056674924 Chr.10: 124835975-124835975 on Build GRCh38
rs896643204 Chr.10: 124835993-124835993 on Build GRCh38
rs540578494 Chr.10: 124836072-124836072 on Build GRCh38
rs888947425 Chr.10: 124836313-124836313 on Build GRCh38
rs145727957 Chr.10: 124836005-124836005 on Build GRCh38
rs542800378 Chr.10: 124835954-124835954 on Build GRCh38
rs532164995 Chr.10: 124836214-124836214 on Build GRCh38
rs561073590 Chr.10: 124835988-124835988 on Build GRCh38
rs548333525 Chr.10: 124836354-124836354 on Build GRCh38
rs968086188 Chr.10: 124836274-124836274 on Build GRCh38
rs908997964 Chr.10: 124836276-124836276 on Build GRCh38
rs971365351 Chr.10: 124836376-124836376 on Build GRCh38
rs953209336 Chr.10: 124836014-124836014 on Build GRCh38
rs1032579867 Chr.10: 124836352-124836352 on Build GRCh38
rs1035979305 Chr.10: 124835942-124835942 on Build GRCh38
rs963742613 Chr.10: 124836090-124836090 on Build GRCh38
rs1050280211 Chr.10: 124836308-124836308 on Build GRCh38
rs1047215852 Chr.10: 124836011-124836011 on Build GRCh38
rs901503313 Chr.10: 124836300-124836300 on Build GRCh38
rs1010314939 Chr.10: 124836367-124836367 on Build GRCh38
rs937488185 Chr.10: 124836185-124836185 on Build GRCh38
rs926616307 Chr.10: 124836303-124836303 on Build GRCh38
rs759056605 Chr.10: 124836267-124836267 on Build GRCh38
rs776224485 Chr.10: 124836247-124836247 on Build GRCh38
rs940442664 Chr.10: 124836283-124836283 on Build GRCh38
rs887976302 Chr.10: 124836408-124836408 on Build GRCh38
rs1005331513 Chr.10: 124836068-124836068 on Build GRCh38
rs1007403765 Chr.10: 124836034-124836034 on Build GRCh38
rs114104870 Chr.10: 124836292-124836292 on Build GRCh38

View Full Product Details