Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124992605-124993113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBP2
Entrez Gene ID
1488
Gene Name
C-terminal binding protein 2
Gene Aliases
-
UniGene
Hs.501345
Gene Chromosome Location
Chr.10: 124987849-125161061 on Build GRCh38
Transcripts
NM_001083914.2 NM_001290214.2 NM_001290215.2 NM_001321012.1 NM_001321013.1 NM_001321014.1 NM_001329.3 NM_022802.2 XM_005269561.2 XM_005269564.2 XM_005269567.2 XM_005269568.4 XM_005269569.2 XM_005269571.2 XM_005269572.3 XM_006717642.2 XM_011539349.2 XM_011539351.1 XM_011539353.1 XM_011539354.1 XM_011539355.1 XM_011539357.1 XM_011539358.1 XM_017015756.1 XM_017015757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620583 c.2672A>G p.E891G Chr.10: 124992800-124992800 on Build GRCh38
3806674 c.1155C>T p.Y385Y Chr.10: 124992697-124992697 on Build GRCh38
5009183 c.2662C>T p.R888C Chr.10: 124992810-124992810 on Build GRCh38
5468484 c.1098G>A p.A366A Chr.10: 124992754-124992754 on Build GRCh38
2021350 c.2762A>G p.N921S Chr.10: 124992710-124992710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962994917 Chr.10: 124993077-124993077 on Build GRCh38
rs568038974 Chr.10: 124992642-124992642 on Build GRCh38
rs765737846 Chr.10: 124992634-124992634 on Build GRCh38
rs904524739 Chr.10: 124992932-124992932 on Build GRCh38
rs529005643 Chr.10: 124992645-124992645 on Build GRCh38
rs749342138 Chr.10: 124992662-124992662 on Build GRCh38
rs1026175779 Chr.10: 124992999-124992999 on Build GRCh38
rs923141772 Chr.10: 124992738-124992738 on Build GRCh38
rs866913452 Chr.10: 124992696-124992696 on Build GRCh38
rs759102800 Chr.10: 124992781-124992781 on Build GRCh38
rs934436565 Chr.10: 124992926-124992926 on Build GRCh38
rs1052292135 Chr.10: 124992776-124992776 on Build GRCh38
rs553142350 Chr.10: 124993055-124993055 on Build GRCh38
rs933220172 Chr.10: 124992745-124992745 on Build GRCh38
rs773115590 Chr.10: 124992736-124992736 on Build GRCh38
rs1043088615 Chr.10: 124992802-124992802 on Build GRCh38
rs200861963 Chr.10: 124992846-124992846 on Build GRCh38
rs768609784 Chr.10: 124992644-124992644 on Build GRCh38
rs529399734 Chr.10: 124992948-124992949 on Build GRCh38
rs771919969 Chr.10: 124992729-124992729 on Build GRCh38
rs555366446 Chr.10: 124992972-124992972 on Build GRCh38
rs946041017 Chr.10: 124992834-124992834 on Build GRCh38
rs921527712 Chr.10: 124992860-124992860 on Build GRCh38
rs950517351 Chr.10: 124993023-124993023 on Build GRCh38
rs1004580931 Chr.10: 124992991-124992991 on Build GRCh38
rs750131607 Chr.10: 124992649-124992649 on Build GRCh38
rs769009810 Chr.10: 124992674-124992674 on Build GRCh38
rs905908889 Chr.10: 124992813-124992813 on Build GRCh38
rs377717591 Chr.10: 124992862-124992862 on Build GRCh38
rs903344972 Chr.10: 124992877-124992877 on Build GRCh38
rs3217482 Chr.10: 124992652-124992652 on Build GRCh38
rs147571820 Chr.10: 124992754-124992754 on Build GRCh38
rs983311391 Chr.10: 124993053-124993053 on Build GRCh38
rs367903762 Chr.10: 124992650-124992650 on Build GRCh38
rs771120202 Chr.10: 124992685-124992685 on Build GRCh38
rs752036884 Chr.10: 124992816-124992816 on Build GRCh38
rs146951852 Chr.10: 124992704-124992704 on Build GRCh38
rs80242128 Chr.10: 124992752-124992752 on Build GRCh38
rs781290075 Chr.10: 124992697-124992697 on Build GRCh38
rs1014399516 Chr.10: 124992980-124992980 on Build GRCh38
rs374316412 Chr.10: 124992860-124992860 on Build GRCh38
rs753459316 Chr.10: 124992755-124992755 on Build GRCh38
rs750666703 Chr.10: 124992840-124992842 on Build GRCh38
rs201538793 Chr.10: 124992705-124992705 on Build GRCh38
rs573665603 Chr.10: 124992981-124992981 on Build GRCh38
rs766794112 Chr.10: 124992973-124992974 on Build GRCh38
rs746021868 Chr.10: 124992852-124992852 on Build GRCh38
rs539692752 Chr.10: 124992653-124992653 on Build GRCh38
rs764492791 Chr.10: 124992826-124992826 on Build GRCh38
rs895468681 Chr.10: 124992635-124992635 on Build GRCh38
rs1001810477 Chr.10: 124992934-124992934 on Build GRCh38
rs79291988 Chr.10: 124992984-124992984 on Build GRCh38
rs3213907 Chr.10: 124992650-124992650 on Build GRCh38
rs947404043 Chr.10: 124992800-124992800 on Build GRCh38
rs368805350 Chr.10: 124992841-124992841 on Build GRCh38
rs57979465 Chr.10: 124992647-124992647 on Build GRCh38
rs891789193 Chr.10: 124992787-124992787 on Build GRCh38
rs764607238 Chr.10: 124992650-124992651 on Build GRCh38
rs1056038363 Chr.10: 124992952-124992952 on Build GRCh38
rs1043226387 Chr.10: 124992865-124992865 on Build GRCh38
rs769702581 Chr.10: 124992713-124992713 on Build GRCh38
rs921783606 Chr.10: 124992722-124992722 on Build GRCh38
rs753365078 Chr.10: 124992651-124992651 on Build GRCh38
rs761576519 Chr.10: 124992992-124992992 on Build GRCh38
rs537329635 Chr.10: 124992966-124992966 on Build GRCh38
rs761680341 Chr.10: 124992647-124992647 on Build GRCh38
rs767576398 Chr.10: 124992830-124992831 on Build GRCh38
rs755501222 Chr.10: 124992652-124992652 on Build GRCh38
rs895508763 Chr.10: 124992949-124992951 on Build GRCh38
rs753090390 Chr.10: 124992657-124992657 on Build GRCh38
rs1038819003 Chr.10: 124992728-124992728 on Build GRCh38
rs781404394 Chr.10: 124992851-124992851 on Build GRCh38
rs185859463 Chr.10: 124992663-124992663 on Build GRCh38
rs113197376 Chr.10: 124993084-124993084 on Build GRCh38
rs547141454 Chr.10: 124992646-124992646 on Build GRCh38
rs767150375 Chr.10: 124992648-124992648 on Build GRCh38
rs774349850 Chr.10: 124992720-124992720 on Build GRCh38
rs200318561 Chr.10: 124992839-124992839 on Build GRCh38
rs913278897 Chr.10: 124992809-124992809 on Build GRCh38
rs878963358 Chr.10: 124992740-124992740 on Build GRCh38
rs886154330 Chr.10: 124992987-124992987 on Build GRCh38
rs570048983 Chr.10: 124992864-124992864 on Build GRCh38
rs3213906 Chr.10: 124992726-124992726 on Build GRCh38
rs753283053 Chr.10: 124992799-124992799 on Build GRCh38
rs751891313 Chr.10: 124992838-124992838 on Build GRCh38
rs994732521 Chr.10: 124992994-124993002 on Build GRCh38
rs761919168 Chr.10: 124992826-124992831 on Build GRCh38
rs758782963 Chr.10: 124992810-124992810 on Build GRCh38

View Full Product Details