Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125656085-125656584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX36
Entrez Gene ID
387718
Gene Name
testis expressed 36
Gene Aliases
C10orf122, bA383C5.1
UniGene
Hs.148259
Gene Chromosome Location
Chr.10: 125576522-125683144 on Build GRCh38
Transcripts
NM_001128202.2 NM_001318133.1 XM_005269817.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2021473 c.346C>A p.P116T Chr.10: 125656115-125656115 on Build GRCh38
4165419 c.320T>A p.F107Y Chr.10: 125656141-125656141 on Build GRCh38
1584840 c.325C>A p.L109I Chr.10: 125656136-125656136 on Build GRCh38
5594274 c.347C>T p.P116L Chr.10: 125656114-125656114 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554781535 Chr.10: 125656154-125656154 on Build GRCh38
rs546321983 Chr.10: 125656459-125656459 on Build GRCh38
rs912404907 Chr.10: 125656110-125656110 on Build GRCh38
rs1047113325 Chr.10: 125656503-125656503 on Build GRCh38
rs746076407 Chr.10: 125656164-125656164 on Build GRCh38
rs763872498 Chr.10: 125656178-125656180 on Build GRCh38
rs950961778 Chr.10: 125656557-125656557 on Build GRCh38
rs936380262 Chr.10: 125656200-125656200 on Build GRCh38
rs145370162 Chr.10: 125656497-125656497 on Build GRCh38
rs12252504 Chr.10: 125656495-125656495 on Build GRCh38
rs943381269 Chr.10: 125656346-125656346 on Build GRCh38
rs949680563 Chr.10: 125656274-125656274 on Build GRCh38
rs762990876 Chr.10: 125656249-125656250 on Build GRCh38
rs936432398 Chr.10: 125656438-125656438 on Build GRCh38
rs367550179 Chr.10: 125656176-125656176 on Build GRCh38
rs939172371 Chr.10: 125656410-125656410 on Build GRCh38
rs1042269607 Chr.10: 125656371-125656371 on Build GRCh38
rs1016388901 Chr.10: 125656349-125656349 on Build GRCh38
rs569075758 Chr.10: 125656506-125656506 on Build GRCh38
rs550884162 Chr.10: 125656505-125656505 on Build GRCh38
rs149208707 Chr.10: 125656504-125656504 on Build GRCh38
rs1012335465 Chr.10: 125656437-125656437 on Build GRCh38
rs115949385 Chr.10: 125656406-125656406 on Build GRCh38
rs987946550 Chr.10: 125656322-125656322 on Build GRCh38
rs1046534128 Chr.10: 125656289-125656289 on Build GRCh38
rs373093013 Chr.10: 125656195-125656195 on Build GRCh38
rs754179260 Chr.10: 125656170-125656170 on Build GRCh38
rs150029867 Chr.10: 125656355-125656355 on Build GRCh38
rs71486542 Chr.10: 125656248-125656249 on Build GRCh38
rs987147001 Chr.10: 125656383-125656383 on Build GRCh38
rs542689256 Chr.10: 125656362-125656362 on Build GRCh38
rs559428988 Chr.10: 125656413-125656413 on Build GRCh38
rs781717191 Chr.10: 125656157-125656157 on Build GRCh38
rs536481030 Chr.10: 125656547-125656547 on Build GRCh38
rs775809467 Chr.10: 125656213-125656213 on Build GRCh38
rs949642742 Chr.10: 125656469-125656469 on Build GRCh38
rs973577010 Chr.10: 125656199-125656199 on Build GRCh38
rs977808870 Chr.10: 125656414-125656414 on Build GRCh38
rs770807784 Chr.10: 125656493-125656493 on Build GRCh38
rs533984132 Chr.10: 125656171-125656171 on Build GRCh38
rs200297774 Chr.10: 125656207-125656207 on Build GRCh38
rs899221778 Chr.10: 125656533-125656533 on Build GRCh38
rs66461124 Chr.10: 125656250-125656251 on Build GRCh38
rs71486543 Chr.10: 125656550-125656551 on Build GRCh38
rs4962477 Chr.10: 125656401-125656401 on Build GRCh38
rs745796039 Chr.10: 125656237-125656237 on Build GRCh38
rs779149393 Chr.10: 125656187-125656187 on Build GRCh38
rs769845763 Chr.10: 125656318-125656318 on Build GRCh38
rs995031150 Chr.10: 125656509-125656509 on Build GRCh38
rs545323219 Chr.10: 125656320-125656320 on Build GRCh38
rs528088304 Chr.10: 125656447-125656447 on Build GRCh38
rs553222902 Chr.10: 125656221-125656221 on Build GRCh38
rs780552847 Chr.10: 125656160-125656160 on Build GRCh38
rs781542332 Chr.10: 125656186-125656186 on Build GRCh38
rs761804541 Chr.10: 125656423-125656423 on Build GRCh38
rs945111127 Chr.10: 125656120-125656120 on Build GRCh38
rs535254616 Chr.10: 125656250-125656263 on Build GRCh38
rs756593760 Chr.10: 125656155-125656155 on Build GRCh38
rs899252887 Chr.10: 125656350-125656350 on Build GRCh38
rs886629256 Chr.10: 125656312-125656312 on Build GRCh38
rs184831551 Chr.10: 125656167-125656167 on Build GRCh38
rs916249953 Chr.10: 125656461-125656461 on Build GRCh38
rs577969339 Chr.10: 125656296-125656296 on Build GRCh38
rs189673609 Chr.10: 125656358-125656358 on Build GRCh38
rs911949551 Chr.10: 125656339-125656339 on Build GRCh38
rs953965825 Chr.10: 125656366-125656366 on Build GRCh38

View Full Product Details