Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125661755-125662253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX36
Entrez Gene ID
387718
Gene Name
testis expressed 36
Gene Aliases
C10orf122, bA383C5.1
UniGene
Hs.148259
Gene Chromosome Location
Chr.10: 125576522-125683144 on Build GRCh38
Transcripts
NM_001128202.2 NM_001318133.1 XM_005269817.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1346610 c.68T>C p.L23P Chr.10: 125661961-125661961 on Build GRCh38
5667931 c.147G>A p.E49E Chr.10: 125661882-125661882 on Build GRCh38
5991385 c.183G>T p.K61N Chr.10: 125661846-125661846 on Build GRCh38
4452400 c.182A>T p.K61M Chr.10: 125661847-125661847 on Build GRCh38
1584838 c.141A>G p.Q47Q Chr.10: 125661888-125661888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763254561 Chr.10: 125661934-125661934 on Build GRCh38
rs942034669 Chr.10: 125661895-125661895 on Build GRCh38
rs1022302131 Chr.10: 125661787-125661787 on Build GRCh38
rs987401355 Chr.10: 125662206-125662206 on Build GRCh38
rs372277807 Chr.10: 125661994-125661994 on Build GRCh38
rs573776440 Chr.10: 125661924-125661924 on Build GRCh38
rs199742415 Chr.10: 125661913-125661913 on Build GRCh38
rs191947873 Chr.10: 125661930-125661930 on Build GRCh38
rs974263443 Chr.10: 125662044-125662044 on Build GRCh38
rs549646168 Chr.10: 125662117-125662117 on Build GRCh38
rs779914158 Chr.10: 125661866-125661866 on Build GRCh38
rs919254385 Chr.10: 125662201-125662201 on Build GRCh38
rs368590911 Chr.10: 125661980-125661980 on Build GRCh38
rs903456766 Chr.10: 125662043-125662043 on Build GRCh38
rs546740464 Chr.10: 125662231-125662231 on Build GRCh38
rs758468420 Chr.10: 125661839-125661839 on Build GRCh38
rs932894367 Chr.10: 125661882-125661882 on Build GRCh38
rs992163262 Chr.10: 125661939-125661939 on Build GRCh38
rs1038803273 Chr.10: 125661965-125661965 on Build GRCh38
rs760215142 Chr.10: 125661955-125661955 on Build GRCh38
rs891355589 Chr.10: 125662146-125662146 on Build GRCh38
rs374762971 Chr.10: 125661853-125661853 on Build GRCh38
rs915956846 Chr.10: 125661892-125661892 on Build GRCh38
rs1033677327 Chr.10: 125661921-125661921 on Build GRCh38
rs899525341 Chr.10: 125661863-125661863 on Build GRCh38
rs533012924 Chr.10: 125661944-125661944 on Build GRCh38
rs9422915 Chr.10: 125661917-125661917 on Build GRCh38
rs143081958 Chr.10: 125661778-125661778 on Build GRCh38
rs2128640 Chr.10: 125662191-125662191 on Build GRCh38
rs777907804 Chr.10: 125661841-125661841 on Build GRCh38
rs867051539 Chr.10: 125661879-125661879 on Build GRCh38
rs941125430 Chr.10: 125661848-125661848 on Build GRCh38
rs189072771 Chr.10: 125661827-125661827 on Build GRCh38
rs752945530 Chr.10: 125661831-125661831 on Build GRCh38
rs556911081 Chr.10: 125661799-125661799 on Build GRCh38
rs541914476 Chr.10: 125662174-125662174 on Build GRCh38
rs891379798 Chr.10: 125661910-125661910 on Build GRCh38
rs866430986 Chr.10: 125662086-125662086 on Build GRCh38
rs991439195 Chr.10: 125661785-125661785 on Build GRCh38
rs563277519 Chr.10: 125662071-125662071 on Build GRCh38
rs756055562 Chr.10: 125661854-125661854 on Build GRCh38
rs184784989 Chr.10: 125662183-125662183 on Build GRCh38
rs967647844 Chr.10: 125661793-125661793 on Build GRCh38
rs755355754 Chr.10: 125661823-125661823 on Build GRCh38
rs780776521 Chr.10: 125662052-125662052 on Build GRCh38
rs573658433 Chr.10: 125662096-125662096 on Build GRCh38
rs151185267 Chr.10: 125661925-125661925 on Build GRCh38
rs971316028 Chr.10: 125661958-125661958 on Build GRCh38
rs545101288 Chr.10: 125661793-125661793 on Build GRCh38
rs553135405 Chr.10: 125661809-125661809 on Build GRCh38
rs999049654 Chr.10: 125662062-125662062 on Build GRCh38
rs983030815 Chr.10: 125661830-125661830 on Build GRCh38
rs949827234 Chr.10: 125661829-125661829 on Build GRCh38
rs945655983 Chr.10: 125662175-125662175 on Build GRCh38
rs766936702 Chr.10: 125661796-125661796 on Build GRCh38
rs1051324928 Chr.10: 125661893-125661893 on Build GRCh38
rs1023040013 Chr.10: 125662159-125662159 on Build GRCh38
rs543689469 Chr.10: 125661911-125661911 on Build GRCh38
rs932279925 Chr.10: 125662113-125662113 on Build GRCh38
rs199568733 Chr.10: 125661967-125661967 on Build GRCh38
rs903921456 Chr.10: 125661914-125661914 on Build GRCh38
rs950746329 Chr.10: 125662202-125662202 on Build GRCh38
rs201711904 Chr.10: 125661950-125661950 on Build GRCh38
rs941104483 Chr.10: 125662028-125662028 on Build GRCh38
rs760031091 Chr.10: 125661984-125661984 on Build GRCh38

View Full Product Details