Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125722738-125723244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDRF1
Entrez Gene ID
26098
Gene Name
erythroid differentiation regulatory factor 1
Gene Aliases
C10orf137
UniGene
Hs.468688
Gene Chromosome Location
Chr.10: 125719515-125764143 on Build GRCh38
Transcripts
NM_001202438.1 NM_015608.2 NR_110857.1 NR_110858.1 NR_110859.1 XM_017016066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6613829 c.357T>C p.N119N Chr.10: 125723107-125723107 on Build GRCh38
3435452 c.364G>A p.D122N Chr.10: 125723114-125723114 on Build GRCh38
5640143 c.347C>T p.S116L Chr.10: 125723097-125723097 on Build GRCh38
3435453 c.364G>A p.D122N Chr.10: 125723114-125723114 on Build GRCh38
5640144 c.347C>T p.S116L Chr.10: 125723097-125723097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765231718 Chr.10: 125722972-125722972 on Build GRCh38
rs927824751 Chr.10: 125722778-125722781 on Build GRCh38
rs117524794 Chr.10: 125723134-125723134 on Build GRCh38
rs375821509 Chr.10: 125723049-125723049 on Build GRCh38
rs1006640213 Chr.10: 125722866-125722866 on Build GRCh38
rs556223924 Chr.10: 125722820-125722820 on Build GRCh38
rs1017975938 Chr.10: 125722896-125722896 on Build GRCh38
rs781684131 Chr.10: 125723131-125723131 on Build GRCh38
rs762489054 Chr.10: 125723071-125723071 on Build GRCh38
rs897034216 Chr.10: 125723184-125723184 on Build GRCh38
rs888214787 Chr.10: 125722848-125722848 on Build GRCh38
rs757617620 Chr.10: 125723122-125723122 on Build GRCh38
rs780373128 Chr.10: 125723152-125723152 on Build GRCh38
rs560452892 Chr.10: 125723034-125723034 on Build GRCh38
rs759193427 Chr.10: 125723046-125723046 on Build GRCh38
rs752228912 Chr.10: 125723051-125723051 on Build GRCh38
rs896997257 Chr.10: 125722804-125722804 on Build GRCh38
rs746960555 Chr.10: 125723163-125723163 on Build GRCh38
rs952471997 Chr.10: 125722843-125722843 on Build GRCh38
rs577340033 Chr.10: 125722769-125722769 on Build GRCh38
rs17153469 Chr.10: 125723156-125723156 on Build GRCh38
rs906541702 Chr.10: 125722772-125722772 on Build GRCh38
rs993745710 Chr.10: 125722990-125722990 on Build GRCh38
rs948180752 Chr.10: 125723144-125723144 on Build GRCh38
rs977423371 Chr.10: 125722941-125722941 on Build GRCh38
rs565057695 Chr.10: 125723185-125723185 on Build GRCh38
rs965059607 Chr.10: 125722914-125722914 on Build GRCh38
rs1018162353 Chr.10: 125723159-125723159 on Build GRCh38
rs941090596 Chr.10: 125722899-125722900 on Build GRCh38
rs373852221 Chr.10: 125723150-125723150 on Build GRCh38
rs748089615 Chr.10: 125722787-125722787 on Build GRCh38
rs777630022 Chr.10: 125723162-125723162 on Build GRCh38
rs773194506 Chr.10: 125723145-125723145 on Build GRCh38
rs574633907 Chr.10: 125722912-125722912 on Build GRCh38
rs76903239 Chr.10: 125723219-125723219 on Build GRCh38
rs1036679001 Chr.10: 125722915-125722915 on Build GRCh38
rs372101903 Chr.10: 125723180-125723180 on Build GRCh38
rs1003227806 Chr.10: 125722777-125722777 on Build GRCh38
rs1026742705 Chr.10: 125722832-125722832 on Build GRCh38
rs764553488 Chr.10: 125723086-125723086 on Build GRCh38
rs369134194 Chr.10: 125723120-125723120 on Build GRCh38
rs371316473 Chr.10: 125723179-125723179 on Build GRCh38
rs776333335 Chr.10: 125723038-125723038 on Build GRCh38
rs74162849 Chr.10: 125722957-125722957 on Build GRCh38
rs141738339 Chr.10: 125722781-125722781 on Build GRCh38
rs756416043 Chr.10: 125723151-125723151 on Build GRCh38
rs760428103 Chr.10: 125723021-125723021 on Build GRCh38
rs776965454 Chr.10: 125722953-125722953 on Build GRCh38

View Full Product Details