Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125831295-125831799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCCIP
Entrez Gene ID
56647
Gene Name
BRCA2 and CDKN1A interacting protein
Gene Aliases
TOK-1, TOK1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125823535-125853695 on Build GRCh38
Transcripts
NM_016567.3 NM_078468.2 NM_078469.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5782475 c.456C>T p.F152F Chr.10: 125831464-125831464 on Build GRCh38
5782476 c.456C>T p.F152F Chr.10: 125831464-125831464 on Build GRCh38
2021732 c.436C>T p.Q146* Chr.10: 125831444-125831444 on Build GRCh38
6451183 c.449T>C p.L150P Chr.10: 125831457-125831457 on Build GRCh38
6199973 c.594G>A p.Q198Q Chr.10: 125831602-125831602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777164629 Chr.10: 125831642-125831643 on Build GRCh38
rs148258244 Chr.10: 125831608-125831608 on Build GRCh38
rs914667610 Chr.10: 125831346-125831346 on Build GRCh38
rs751739190 Chr.10: 125831386-125831386 on Build GRCh38
rs879179818 Chr.10: 125831609-125831609 on Build GRCh38
rs554549451 Chr.10: 125831321-125831321 on Build GRCh38
rs749781661 Chr.10: 125831424-125831424 on Build GRCh38
rs757988268 Chr.10: 125831643-125831643 on Build GRCh38
rs780531810 Chr.10: 125831422-125831422 on Build GRCh38
rs750752017 Chr.10: 125831419-125831419 on Build GRCh38
rs766279198 Chr.10: 125831372-125831372 on Build GRCh38
rs968820658 Chr.10: 125831347-125831347 on Build GRCh38
rs17153605 Chr.10: 125831479-125831479 on Build GRCh38
rs375856702 Chr.10: 125831374-125831374 on Build GRCh38
rs369785811 Chr.10: 125831508-125831508 on Build GRCh38
rs766630392 Chr.10: 125831557-125831557 on Build GRCh38
rs777411270 Chr.10: 125831582-125831582 on Build GRCh38
rs144879420 Chr.10: 125831460-125831460 on Build GRCh38
rs756437566 Chr.10: 125831421-125831421 on Build GRCh38
rs977598181 Chr.10: 125831405-125831405 on Build GRCh38
rs947716824 Chr.10: 125831489-125831489 on Build GRCh38
rs759383074 Chr.10: 125831477-125831478 on Build GRCh38
rs527964424 Chr.10: 125831500-125831500 on Build GRCh38
rs748687255 Chr.10: 125831383-125831383 on Build GRCh38
rs754241115 Chr.10: 125831558-125831558 on Build GRCh38
rs755434191 Chr.10: 125831570-125831570 on Build GRCh38
rs533326147 Chr.10: 125831656-125831656 on Build GRCh38
rs963983643 Chr.10: 125831762-125831765 on Build GRCh38
rs143179971 Chr.10: 125831581-125831581 on Build GRCh38
rs1051263239 Chr.10: 125831751-125831751 on Build GRCh38
rs767665746 Chr.10: 125831417-125831417 on Build GRCh38
rs755527658 Chr.10: 125831425-125831425 on Build GRCh38
rs978689677 Chr.10: 125831332-125831332 on Build GRCh38
rs770669388 Chr.10: 125831590-125831590 on Build GRCh38
rs773657797 Chr.10: 125831769-125831771 on Build GRCh38
rs771703694 Chr.10: 125831447-125831447 on Build GRCh38
rs964456382 Chr.10: 125831322-125831322 on Build GRCh38
rs746755352 Chr.10: 125831455-125831455 on Build GRCh38
rs764260363 Chr.10: 125831383-125831383 on Build GRCh38
rs560582713 Chr.10: 125831480-125831480 on Build GRCh38
rs778360218 Chr.10: 125831428-125831428 on Build GRCh38
rs886792383 Chr.10: 125831567-125831567 on Build GRCh38
rs199996010 Chr.10: 125831403-125831403 on Build GRCh38
rs201532843 Chr.10: 125831450-125831450 on Build GRCh38
rs374632268 Chr.10: 125831548-125831548 on Build GRCh38
rs774670252 Chr.10: 125831729-125831729 on Build GRCh38
rs1038619352 Chr.10: 125831724-125831724 on Build GRCh38
rs917019924 Chr.10: 125831440-125831440 on Build GRCh38
rs577038501 Chr.10: 125831357-125831357 on Build GRCh38
rs887011195 Chr.10: 125831727-125831727 on Build GRCh38
rs147798836 Chr.10: 125831426-125831426 on Build GRCh38
rs781107076 Chr.10: 125831591-125831591 on Build GRCh38
rs765306821 Chr.10: 125831476-125831476 on Build GRCh38
rs141189146 Chr.10: 125831444-125831444 on Build GRCh38
rs745690875 Chr.10: 125831600-125831600 on Build GRCh38
rs763012804 Chr.10: 125831639-125831639 on Build GRCh38
rs533936814 Chr.10: 125831320-125831320 on Build GRCh38
rs752577217 Chr.10: 125831521-125831522 on Build GRCh38
rs759527887 Chr.10: 125831462-125831462 on Build GRCh38
rs765306663 Chr.10: 125831520-125831521 on Build GRCh38
rs779411460 Chr.10: 125831573-125831573 on Build GRCh38
rs145046178 Chr.10: 125831459-125831459 on Build GRCh38
rs767693579 Chr.10: 125831764-125831764 on Build GRCh38
rs775583762 Chr.10: 125831626-125831626 on Build GRCh38
rs763326410 Chr.10: 125831537-125831537 on Build GRCh38
rs11596442 Chr.10: 125831744-125831744 on Build GRCh38
rs746631713 Chr.10: 125831588-125831588 on Build GRCh38
rs772030242 Chr.10: 125831654-125831654 on Build GRCh38
rs756347867 Chr.10: 125831458-125831458 on Build GRCh38
rs925849363 Chr.10: 125831334-125831334 on Build GRCh38
rs760407684 Chr.10: 125831369-125831369 on Build GRCh38

View Full Product Details