Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127008651-127009157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK1
Entrez Gene ID
1793
Gene Name
dedicator of cytokinesis 1
Gene Aliases
DOCK180, ced5
UniGene
Hs.159195
Gene Chromosome Location
Chr.10: 126905409-127452517 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6200139 c.1048C>T p.P350S Chr.10: 127008794-127008794 on Build GRCh38
160555 c.1058+1G>C p.? Chr.10: 127008805-127008805 on Build GRCh38
3435569 c.1049C>T p.P350L Chr.10: 127008795-127008795 on Build GRCh38
6410336 c.1035G>T p.K345N Chr.10: 127008781-127008781 on Build GRCh38
6362447 c.1057C>A p.P353T Chr.10: 127008803-127008803 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1011182137 Chr.10: 127008927-127008927 on Build GRCh38
rs767563624 Chr.10: 127008823-127008823 on Build GRCh38
rs960664337 Chr.10: 127008961-127008961 on Build GRCh38
rs761849675 Chr.10: 127008816-127008816 on Build GRCh38
rs1026506259 Chr.10: 127008974-127008974 on Build GRCh38
rs560821920 Chr.10: 127008800-127008800 on Build GRCh38
rs371110999 Chr.10: 127008981-127008981 on Build GRCh38
rs1000850623 Chr.10: 127008957-127008957 on Build GRCh38
rs746697217 Chr.10: 127008750-127008750 on Build GRCh38
rs529146709 Chr.10: 127008999-127008999 on Build GRCh38
rs951826725 Chr.10: 127008978-127008978 on Build GRCh38
rs769746568 Chr.10: 127008810-127008810 on Build GRCh38
rs185863599 Chr.10: 127008826-127008826 on Build GRCh38
rs9418678 Chr.10: 127008814-127008814 on Build GRCh38
rs979848299 Chr.10: 127008809-127008809 on Build GRCh38
rs768633815 Chr.10: 127008953-127008953 on Build GRCh38
rs376208957 Chr.10: 127008837-127008837 on Build GRCh38
rs556342533 Chr.10: 127008768-127008768 on Build GRCh38
rs35541322 Chr.10: 127008754-127008754 on Build GRCh38
rs777477340 Chr.10: 127008742-127008742 on Build GRCh38
rs927039868 Chr.10: 127008813-127008813 on Build GRCh38
rs902016955 Chr.10: 127008908-127008908 on Build GRCh38
rs778515009 Chr.10: 127008697-127008697 on Build GRCh38
rs577483562 Chr.10: 127008718-127008718 on Build GRCh38
rs1052962893 Chr.10: 127008916-127008916 on Build GRCh38
rs905152358 Chr.10: 127008936-127008936 on Build GRCh38
rs892541764 Chr.10: 127008923-127008923 on Build GRCh38
rs531637774 Chr.10: 127008929-127008929 on Build GRCh38
rs779968531 Chr.10: 127008933-127008933 on Build GRCh38
rs759431307 Chr.10: 127008784-127008784 on Build GRCh38
rs189445897 Chr.10: 127008872-127008872 on Build GRCh38
rs564373523 Chr.10: 127008887-127008887 on Build GRCh38
rs747635970 Chr.10: 127008713-127008713 on Build GRCh38
rs964167307 Chr.10: 127009092-127009092 on Build GRCh38
rs993164239 Chr.10: 127008972-127008972 on Build GRCh38
rs532383917 Chr.10: 127009048-127009048 on Build GRCh38
rs956720980 Chr.10: 127008988-127008988 on Build GRCh38
rs771518886 Chr.10: 127008716-127008716 on Build GRCh38
rs9418679 Chr.10: 127008832-127008832 on Build GRCh38
rs758336480 Chr.10: 127008693-127008693 on Build GRCh38
rs527259862 Chr.10: 127008840-127008840 on Build GRCh38
rs181834958 Chr.10: 127008982-127008982 on Build GRCh38
rs910057607 Chr.10: 127009013-127009013 on Build GRCh38
rs780821337 Chr.10: 127008952-127008952 on Build GRCh38
rs1024269710 Chr.10: 127009010-127009010 on Build GRCh38
rs988752455 Chr.10: 127008691-127008691 on Build GRCh38
rs374245459 Chr.10: 127008787-127008787 on Build GRCh38
rs753156369 Chr.10: 127008712-127008713 on Build GRCh38
rs545516720 Chr.10: 127008870-127008870 on Build GRCh38
rs549959502 Chr.10: 127008945-127008945 on Build GRCh38

View Full Product Details