Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127877709-127878206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLRN3
Entrez Gene ID
119467
Gene Name
clarin 3
Gene Aliases
TMEM12, USH3AL1
UniGene
Hs.242014
Gene Chromosome Location
Chr.10: 127877841-127892947 on Build GRCh38
Transcripts
NM_152311.3 XM_011539274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4555668 c.663G>A p.R221R Chr.10: 127878167-127878167 on Build GRCh38
4555769 c.668G>A p.G223E Chr.10: 127878162-127878162 on Build GRCh38
352289 c.680G>C p.*227S Chr.10: 127878150-127878150 on Build GRCh38
6888662 c.648G>A p.M216I Chr.10: 127878182-127878182 on Build GRCh38
372387 c.631G>T p.E211* Chr.10: 127878199-127878199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371588320 Chr.10: 127878108-127878108 on Build GRCh38
rs1036423260 Chr.10: 127877885-127877885 on Build GRCh38
rs1013833738 Chr.10: 127877780-127877780 on Build GRCh38
rs988314965 Chr.10: 127877925-127877925 on Build GRCh38
rs1048801858 Chr.10: 127877942-127877942 on Build GRCh38
rs1030316513 Chr.10: 127877901-127877901 on Build GRCh38
rs986321072 Chr.10: 127877755-127877755 on Build GRCh38
rs59803722 Chr.10: 127877995-127877995 on Build GRCh38
rs574142812 Chr.10: 127877852-127877852 on Build GRCh38
rs371504217 Chr.10: 127877952-127877952 on Build GRCh38
rs979458358 Chr.10: 127877831-127877831 on Build GRCh38
rs953595349 Chr.10: 127877740-127877740 on Build GRCh38
rs939839392 Chr.10: 127878037-127878037 on Build GRCh38
rs146185971 Chr.10: 127877950-127877950 on Build GRCh38
rs534511769 Chr.10: 127878056-127878056 on Build GRCh38
rs184300283 Chr.10: 127878182-127878182 on Build GRCh38
rs748056143 Chr.10: 127878111-127878111 on Build GRCh38
rs141557538 Chr.10: 127877918-127877918 on Build GRCh38
rs953648933 Chr.10: 127877902-127877902 on Build GRCh38
rs926571821 Chr.10: 127877833-127877833 on Build GRCh38
rs754808075 Chr.10: 127877840-127877840 on Build GRCh38
rs546414925 Chr.10: 127878057-127878057 on Build GRCh38
rs1048267415 Chr.10: 127877993-127877993 on Build GRCh38
rs752016614 Chr.10: 127878174-127878174 on Build GRCh38
rs7912303 Chr.10: 127877763-127877763 on Build GRCh38
rs149953463 Chr.10: 127878164-127878164 on Build GRCh38
rs949058411 Chr.10: 127877829-127877829 on Build GRCh38
rs909723468 Chr.10: 127878020-127878020 on Build GRCh38
rs1007817622 Chr.10: 127878151-127878151 on Build GRCh38
rs937965140 Chr.10: 127877990-127877990 on Build GRCh38
rs931485240 Chr.10: 127877932-127877932 on Build GRCh38
rs756500504 Chr.10: 127878179-127878179 on Build GRCh38
rs896436668 Chr.10: 127877768-127877768 on Build GRCh38
rs937914271 Chr.10: 127877834-127877834 on Build GRCh38
rs975563588 Chr.10: 127877907-127877907 on Build GRCh38
rs758283969 Chr.10: 127877969-127877969 on Build GRCh38
rs2296785 Chr.10: 127877905-127877905 on Build GRCh38
rs749338314 Chr.10: 127878181-127878181 on Build GRCh38
rs988167512 Chr.10: 127877888-127877888 on Build GRCh38
rs763299039 Chr.10: 127878144-127878144 on Build GRCh38
rs912800290 Chr.10: 127877900-127877900 on Build GRCh38
rs778738434 Chr.10: 127878105-127878105 on Build GRCh38
rs1032355795 Chr.10: 127878172-127878172 on Build GRCh38
rs114036758 Chr.10: 127877857-127877857 on Build GRCh38
rs182026046 Chr.10: 127877948-127877948 on Build GRCh38
rs567785324 Chr.10: 127878006-127878006 on Build GRCh38
rs56071108 Chr.10: 127877782-127877782 on Build GRCh38
rs1015219537 Chr.10: 127877764-127877764 on Build GRCh38
rs897931403 Chr.10: 127878100-127878100 on Build GRCh38
rs571190289 Chr.10: 127878064-127878064 on Build GRCh38
rs763711278 Chr.10: 127878142-127878143 on Build GRCh38
rs746634779 Chr.10: 127877811-127877811 on Build GRCh38
rs762378081 Chr.10: 127878175-127878175 on Build GRCh38
rs908968538 Chr.10: 127877947-127877947 on Build GRCh38
rs1046311628 Chr.10: 127877832-127877832 on Build GRCh38
rs1032519529 Chr.10: 127877906-127877906 on Build GRCh38
rs956883156 Chr.10: 127877883-127877883 on Build GRCh38
rs936450967 Chr.10: 127877770-127877770 on Build GRCh38
rs117580940 Chr.10: 127878126-127878126 on Build GRCh38
rs750823026 Chr.10: 127878148-127878148 on Build GRCh38
rs139046381 Chr.10: 127878125-127878125 on Build GRCh38
rs1048257895 Chr.10: 127877842-127877842 on Build GRCh38
rs981766919 Chr.10: 127877984-127877984 on Build GRCh38
rs549171621 Chr.10: 127877986-127877986 on Build GRCh38
rs1006728065 Chr.10: 127877860-127877860 on Build GRCh38

View Full Product Details