Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 129843051-129843559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF3
Entrez Gene ID
253738
Gene Name
early B-cell factor 3
Gene Aliases
COE3, EBF-3, O/E-2, OE-2
UniGene
Hs.591374
Gene Chromosome Location
Chr.10: 129835232-129964274 on Build GRCh38
Transcripts
NM_001005463.2 XM_005252667.3 XM_005252668.3 XM_005252669.3 XM_006717739.3 XM_006717740.3 XM_006717741.3 XM_006717742.3 XM_006717743.3 XM_006717744.3 XM_011539574.2 XM_011539575.2 XM_017016027.1 NR_036179.1

Gene Symbol
MIR4297
Entrez Gene ID
100422873
Gene Name
microRNA 4297
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.10: 129843299-129843374 on Build GRCh38
Transcripts
NM_001005463.2 XM_005252667.3 XM_005252668.3 XM_005252669.3 XM_006717739.3 XM_006717740.3 XM_006717741.3 XM_006717742.3 XM_006717743.3 XM_006717744.3 XM_011539574.2 XM_011539575.2 XM_017016027.1 NR_036179.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5685871 c.1134G>A p.V378V Chr.10: 129843170-129843170 on Build GRCh38
5732902 c.1144C>T p.R382W Chr.10: 129843187-129843187 on Build GRCh38
7319036 c.1131G>C p.L377L Chr.10: 129843173-129843173 on Build GRCh38
5580935 c.1174G>A p.G392R Chr.10: 129843157-129843157 on Build GRCh38
5732901 c.1117C>T p.R373W Chr.10: 129843187-129843187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747093637 Chr.10: 129843488-129843488 on Build GRCh38
rs765779838 Chr.10: 129843212-129843212 on Build GRCh38
rs747190003 Chr.10: 129843276-129843276 on Build GRCh38
rs753470853 Chr.10: 129843311-129843311 on Build GRCh38
rs757562720 Chr.10: 129843330-129843330 on Build GRCh38
rs971244179 Chr.10: 129843462-129843462 on Build GRCh38
rs141717894 Chr.10: 129843405-129843405 on Build GRCh38
rs552250926 Chr.10: 129843211-129843211 on Build GRCh38
rs754404775 Chr.10: 129843110-129843110 on Build GRCh38
rs546805685 Chr.10: 129843183-129843183 on Build GRCh38
rs890438139 Chr.10: 129843075-129843075 on Build GRCh38
rs532753259 Chr.10: 129843482-129843482 on Build GRCh38
rs777732440 Chr.10: 129843269-129843269 on Build GRCh38
rs773427214 Chr.10: 129843096-129843096 on Build GRCh38
rs912102247 Chr.10: 129843479-129843479 on Build GRCh38
rs987544250 Chr.10: 129843272-129843272 on Build GRCh38
rs943343454 Chr.10: 129843282-129843282 on Build GRCh38
rs368474714 Chr.10: 129843141-129843141 on Build GRCh38
rs748173058 Chr.10: 129843257-129843257 on Build GRCh38
rs185516749 Chr.10: 129843312-129843312 on Build GRCh38
rs966973952 Chr.10: 129843366-129843366 on Build GRCh38
rs1000426077 Chr.10: 129843461-129843461 on Build GRCh38
rs868224337 Chr.10: 129843202-129843202 on Build GRCh38
rs760190381 Chr.10: 129843189-129843189 on Build GRCh38
rs748310898 Chr.10: 129843170-129843170 on Build GRCh38
rs146580982 Chr.10: 129843187-129843187 on Build GRCh38
rs370927134 Chr.10: 129843197-129843197 on Build GRCh38
rs375143852 Chr.10: 129843253-129843253 on Build GRCh38
rs77585913 Chr.10: 129843391-129843391 on Build GRCh38
rs150173087 Chr.10: 129843458-129843458 on Build GRCh38
rs560892486 Chr.10: 129843179-129843179 on Build GRCh38
rs772255512 Chr.10: 129843261-129843261 on Build GRCh38
rs920575477 Chr.10: 129843499-129843499 on Build GRCh38
rs949407538 Chr.10: 129843217-129843217 on Build GRCh38
rs572911242 Chr.10: 129843076-129843076 on Build GRCh38
rs1012039338 Chr.10: 129843431-129843431 on Build GRCh38
rs1032352144 Chr.10: 129843469-129843469 on Build GRCh38
rs367622835 Chr.10: 129843086-129843086 on Build GRCh38
rs751165401 Chr.10: 129843235-129843235 on Build GRCh38
rs767329347 Chr.10: 129843354-129843354 on Build GRCh38
rs775027465 Chr.10: 129843098-129843099 on Build GRCh38
rs538289418 Chr.10: 129843277-129843277 on Build GRCh38
rs751304900 Chr.10: 129843131-129843131 on Build GRCh38
rs796129106 Chr.10: 129843214-129843214 on Build GRCh38
rs767804328 Chr.10: 129843095-129843095 on Build GRCh38
rs374630902 Chr.10: 129843238-129843238 on Build GRCh38
rs397749697 Chr.10: 129843103-129843104 on Build GRCh38
rs373572583 Chr.10: 129843244-129843244 on Build GRCh38
rs766795814 Chr.10: 129843104-129843104 on Build GRCh38
rs766658789 Chr.10: 129843219-129843220 on Build GRCh38
rs763636962 Chr.10: 129843224-129843224 on Build GRCh38
rs777095766 Chr.10: 129843340-129843340 on Build GRCh38
rs554364540 Chr.10: 129843317-129843317 on Build GRCh38
rs968952835 Chr.10: 129843193-129843193 on Build GRCh38
rs768424110 Chr.10: 129843099-129843100 on Build GRCh38
rs1035197560 Chr.10: 129843147-129843147 on Build GRCh38
rs907788925 Chr.10: 129843528-129843528 on Build GRCh38
rs761507461 Chr.10: 129843340-129843340 on Build GRCh38
rs781092735 Chr.10: 129843239-129843239 on Build GRCh38
rs779087702 Chr.10: 129843166-129843166 on Build GRCh38
rs1027928121 Chr.10: 129843072-129843072 on Build GRCh38
rs763592494 Chr.10: 129843316-129843316 on Build GRCh38
rs988046676 Chr.10: 129843472-129843472 on Build GRCh38
rs555279867 Chr.10: 129843283-129843283 on Build GRCh38
rs767539879 Chr.10: 129843085-129843085 on Build GRCh38
rs764664435 Chr.10: 129843114-129843114 on Build GRCh38
rs568805393 Chr.10: 129843285-129843285 on Build GRCh38
rs925468131 Chr.10: 129843142-129843142 on Build GRCh38
rs750266730 Chr.10: 129843243-129843243 on Build GRCh38
rs574886945 Chr.10: 129843437-129843437 on Build GRCh38
rs956198719 Chr.10: 129843470-129843470 on Build GRCh38
rs114362263 Chr.10: 129843365-129843365 on Build GRCh38
rs773541406 Chr.10: 129843418-129843418 on Build GRCh38
rs780509719 Chr.10: 129843442-129843442 on Build GRCh38
rs201483148 Chr.10: 129843123-129843123 on Build GRCh38
rs190280340 Chr.10: 129843518-129843518 on Build GRCh38
rs762587221 Chr.10: 129843222-129843222 on Build GRCh38
rs560831168 Chr.10: 129843478-129843478 on Build GRCh38
rs773866474 Chr.10: 129843100-129843101 on Build GRCh38
rs770276080 Chr.10: 129843304-129843304 on Build GRCh38
rs886247897 Chr.10: 129843331-129843331 on Build GRCh38
rs35210769 Chr.10: 129843097-129843098 on Build GRCh38
rs993344793 Chr.10: 129843071-129843071 on Build GRCh38
rs558833516 Chr.10: 129843519-129843519 on Build GRCh38
rs529814619 Chr.10: 129843182-129843182 on Build GRCh38
rs1047605754 Chr.10: 129843527-129843527 on Build GRCh38
rs368498164 Chr.10: 129843073-129843073 on Build GRCh38
rs200563752 Chr.10: 129843158-129843158 on Build GRCh38
rs760052658 Chr.10: 129843113-129843113 on Build GRCh38
rs201064631 Chr.10: 129843124-129843124 on Build GRCh38
rs373670730 Chr.10: 129843213-129843213 on Build GRCh38
rs757015366 Chr.10: 129843096-129843097 on Build GRCh38
rs376655042 Chr.10: 129843481-129843481 on Build GRCh38
rs758957159 Chr.10: 129843440-129843440 on Build GRCh38
rs994750859 Chr.10: 129843100-129843100 on Build GRCh38
rs887515054 Chr.10: 129843074-129843074 on Build GRCh38
rs532644240 Chr.10: 129843208-129843208 on Build GRCh38
rs1038880968 Chr.10: 129843500-129843500 on Build GRCh38
rs745731802 Chr.10: 129843146-129843146 on Build GRCh38
rs757133312 Chr.10: 129843134-129843134 on Build GRCh38
rs397732024 Chr.10: 129843104-129843105 on Build GRCh38
rs1017326375 Chr.10: 129843081-129843081 on Build GRCh38

View Full Product Details