Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 130166428-130166940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX3
Entrez Gene ID
10539
Gene Name
glutaredoxin 3
Gene Aliases
GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3
UniGene
Hs.42644
Gene Chromosome Location
Chr.10: 130136375-130182877 on Build GRCh38
Transcripts
NM_001199868.1 NM_001321980.1 NM_006541.4 XM_017015474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3382748 c.525G>A p.Q175Q Chr.10: 130166553-130166553 on Build GRCh38
6128659 c.663A>G p.A221A Chr.10: 130166930-130166930 on Build GRCh38
6551469 c.586A>T p.S196C Chr.10: 130166614-130166614 on Build GRCh38
4438231 c.483C>A p.F161L Chr.10: 130166511-130166511 on Build GRCh38
6128661 c.567G>T p.Q189H Chr.10: 130166595-130166595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745326160 Chr.10: 130166609-130166609 on Build GRCh38
rs776185901 Chr.10: 130166503-130166503 on Build GRCh38
rs1053225409 Chr.10: 130166501-130166501 on Build GRCh38
rs767502993 Chr.10: 130166672-130166672 on Build GRCh38
rs750960659 Chr.10: 130166542-130166542 on Build GRCh38
rs147781921 Chr.10: 130166539-130166539 on Build GRCh38
rs200601247 Chr.10: 130166625-130166625 on Build GRCh38
rs34458027 Chr.10: 130166520-130166520 on Build GRCh38
rs926182314 Chr.10: 130166799-130166799 on Build GRCh38
rs761618892 Chr.10: 130166904-130166904 on Build GRCh38
rs369013678 Chr.10: 130166878-130166878 on Build GRCh38
rs576119657 Chr.10: 130166867-130166867 on Build GRCh38
rs960757315 Chr.10: 130166735-130166735 on Build GRCh38
rs373775553 Chr.10: 130166639-130166639 on Build GRCh38
rs140178704 Chr.10: 130166559-130166559 on Build GRCh38
rs894186714 Chr.10: 130166810-130166810 on Build GRCh38
rs761517084 Chr.10: 130166660-130166660 on Build GRCh38
rs752622541 Chr.10: 130166718-130166718 on Build GRCh38
rs372460784 Chr.10: 130166906-130166906 on Build GRCh38
rs112782379 Chr.10: 130166726-130166726 on Build GRCh38
rs368839826 Chr.10: 130166704-130166704 on Build GRCh38
rs376523898 Chr.10: 130166700-130166700 on Build GRCh38
rs372431410 Chr.10: 130166706-130166706 on Build GRCh38
rs774268279 Chr.10: 130166658-130166658 on Build GRCh38
rs781091036 Chr.10: 130166896-130166897 on Build GRCh38
rs753038200 Chr.10: 130166491-130166492 on Build GRCh38
rs761279925 Chr.10: 130166499-130166499 on Build GRCh38
rs753902179 Chr.10: 130166548-130166548 on Build GRCh38
rs369417078 Chr.10: 130166895-130166895 on Build GRCh38
rs539416184 Chr.10: 130166820-130166820 on Build GRCh38
rs750405705 Chr.10: 130166683-130166683 on Build GRCh38
rs3736900 Chr.10: 130166717-130166717 on Build GRCh38
rs1034056949 Chr.10: 130166806-130166806 on Build GRCh38
rs931296542 Chr.10: 130166613-130166613 on Build GRCh38
rs3736898 Chr.10: 130166909-130166909 on Build GRCh38
rs996521997 Chr.10: 130166586-130166586 on Build GRCh38
rs1041971408 Chr.10: 130166761-130166761 on Build GRCh38
rs772504497 Chr.10: 130166475-130166475 on Build GRCh38
rs779276423 Chr.10: 130166488-130166491 on Build GRCh38
rs748463632 Chr.10: 130166470-130166470 on Build GRCh38
rs991895088 Chr.10: 130166736-130166736 on Build GRCh38
rs980388769 Chr.10: 130166779-130166779 on Build GRCh38
rs771045423 Chr.10: 130166882-130166883 on Build GRCh38
rs969062101 Chr.10: 130166771-130166771 on Build GRCh38
rs549281423 Chr.10: 130166569-130166569 on Build GRCh38
rs1014430115 Chr.10: 130166841-130166841 on Build GRCh38
rs999857678 Chr.10: 130166795-130166795 on Build GRCh38
rs371758988 Chr.10: 130166521-130166521 on Build GRCh38
rs775084748 Chr.10: 130166624-130166624 on Build GRCh38
rs549952259 Chr.10: 130166515-130166515 on Build GRCh38
rs765443500 Chr.10: 130166697-130166697 on Build GRCh38
rs900488522 Chr.10: 130166560-130166560 on Build GRCh38
rs758654353 Chr.10: 130166571-130166571 on Build GRCh38
rs1040794924 Chr.10: 130166526-130166526 on Build GRCh38
rs3736899 Chr.10: 130166762-130166762 on Build GRCh38
rs988979972 Chr.10: 130166861-130166861 on Build GRCh38
rs777129463 Chr.10: 130166591-130166591 on Build GRCh38
rs747151952 Chr.10: 130166874-130166876 on Build GRCh38
rs773886964 Chr.10: 130166902-130166902 on Build GRCh38
rs556149227 Chr.10: 130166847-130166847 on Build GRCh38
rs747334510 Chr.10: 130166707-130166707 on Build GRCh38
rs746406715 Chr.10: 130166875-130166875 on Build GRCh38
rs759188105 Chr.10: 130166511-130166511 on Build GRCh38
rs755065577 Chr.10: 130166692-130166692 on Build GRCh38
rs770329492 Chr.10: 130166500-130166500 on Build GRCh38
rs775115931 Chr.10: 130166882-130166882 on Build GRCh38
rs755686208 Chr.10: 130166473-130166477 on Build GRCh38
rs746088437 Chr.10: 130166740-130166742 on Build GRCh38
rs781629526 Chr.10: 130166873-130166873 on Build GRCh38
rs375832879 Chr.10: 130166498-130166498 on Build GRCh38
rs778253179 Chr.10: 130166573-130166573 on Build GRCh38
rs748888201 Chr.10: 130166891-130166891 on Build GRCh38
rs565905520 Chr.10: 130166587-130166587 on Build GRCh38
rs778249901 Chr.10: 130166869-130166873 on Build GRCh38
rs575518915 Chr.10: 130166802-130166802 on Build GRCh38
rs763783848 Chr.10: 130166627-130166627 on Build GRCh38
rs758944914 Chr.10: 130166492-130166493 on Build GRCh38
rs199617584 Chr.10: 130166604-130166604 on Build GRCh38
rs150329694 Chr.10: 130166619-130166619 on Build GRCh38

View Full Product Details