Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132186740-132187229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL4
Entrez Gene ID
10570
Gene Name
dihydropyrimidinase like 4
Gene Aliases
CRMP3, DRP-4, ULIP4
UniGene
Hs.100058
Gene Chromosome Location
Chr.10: 132184983-132205776 on Build GRCh38
Transcripts
NM_006426.2 XM_005252658.3 XM_011539125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582055 c.8T>C p.F3S Chr.10: 132187071-132187071 on Build GRCh38
2023280 c.27delC p.R11fs*8 Chr.10: 132187090-132187090 on Build GRCh38
6649163 c.32G>A p.R11Q Chr.10: 132187095-132187095 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764833166 Chr.10: 132186773-132186773 on Build GRCh38
rs752311092 Chr.10: 132187027-132187036 on Build GRCh38
rs777949906 Chr.10: 132187108-132187108 on Build GRCh38
rs944586952 Chr.10: 132186790-132186790 on Build GRCh38
rs536395278 Chr.10: 132187058-132187058 on Build GRCh38
rs530905968 Chr.10: 132186975-132186975 on Build GRCh38
rs539903657 Chr.10: 132186774-132186774 on Build GRCh38
rs371429633 Chr.10: 132187059-132187059 on Build GRCh38
rs938042191 Chr.10: 132187143-132187143 on Build GRCh38
rs746361938 Chr.10: 132187053-132187053 on Build GRCh38
rs567913811 Chr.10: 132186851-132186852 on Build GRCh38
rs757485817 Chr.10: 132187111-132187111 on Build GRCh38
rs12772795 Chr.10: 132186812-132186812 on Build GRCh38
rs566685136 Chr.10: 132187116-132187116 on Build GRCh38
rs189978507 Chr.10: 132186799-132186799 on Build GRCh38
rs994581374 Chr.10: 132186973-132186973 on Build GRCh38
rs1049852120 Chr.10: 132187199-132187199 on Build GRCh38
rs917886365 Chr.10: 132187164-132187164 on Build GRCh38
rs776750467 Chr.10: 132186995-132187006 on Build GRCh38
rs911818409 Chr.10: 132186770-132186770 on Build GRCh38
rs753686812 Chr.10: 132187095-132187095 on Build GRCh38
rs560433512 Chr.10: 132186820-132186820 on Build GRCh38
rs753834082 Chr.10: 132187136-132187136 on Build GRCh38
rs765162642 Chr.10: 132186998-132187005 on Build GRCh38
rs71013511 Chr.10: 132186870-132186871 on Build GRCh38
rs533340157 Chr.10: 132186768-132186769 on Build GRCh38
rs1037684378 Chr.10: 132187179-132187179 on Build GRCh38
rs761470716 Chr.10: 132187125-132187125 on Build GRCh38
rs759505531 Chr.10: 132187139-132187139 on Build GRCh38
rs766405880 Chr.10: 132187092-132187092 on Build GRCh38
rs12778190 Chr.10: 132186840-132186840 on Build GRCh38
rs746297877 Chr.10: 132187113-132187113 on Build GRCh38
rs796173063 Chr.10: 132186773-132186774 on Build GRCh38
rs946807372 Chr.10: 132186841-132186841 on Build GRCh38
rs867022088 Chr.10: 132186844-132186844 on Build GRCh38
rs760375152 Chr.10: 132187090-132187090 on Build GRCh38
rs767306362 Chr.10: 132187076-132187076 on Build GRCh38
rs61865793 Chr.10: 132186918-132186918 on Build GRCh38
rs377006117 Chr.10: 132186995-132187010 on Build GRCh38
rs747210665 Chr.10: 132187027-132187027 on Build GRCh38
rs763443523 Chr.10: 132187125-132187126 on Build GRCh38
rs898657539 Chr.10: 132186951-132186951 on Build GRCh38
rs764270730 Chr.10: 132187154-132187154 on Build GRCh38
rs766065700 Chr.10: 132187133-132187133 on Build GRCh38
rs35725115 Chr.10: 132186871-132186871 on Build GRCh38
rs988446014 Chr.10: 132187094-132187094 on Build GRCh38
rs898708464 Chr.10: 132187195-132187196 on Build GRCh38
rs1043819182 Chr.10: 132186888-132186888 on Build GRCh38
rs12356773 Chr.10: 132186997-132186997 on Build GRCh38
rs1001193496 Chr.10: 132186890-132186890 on Build GRCh38
rs758552818 Chr.10: 132187101-132187101 on Build GRCh38
rs778180178 Chr.10: 132187021-132187021 on Build GRCh38
rs867567174 Chr.10: 132186882-132186882 on Build GRCh38
rs547325316 Chr.10: 132186856-132186857 on Build GRCh38
rs1011450000 Chr.10: 132186825-132186825 on Build GRCh38
rs374127056 Chr.10: 132187121-132187121 on Build GRCh38
rs776243647 Chr.10: 132187017-132187017 on Build GRCh38
rs376906063 Chr.10: 132187057-132187057 on Build GRCh38
rs761415785 Chr.10: 132187068-132187068 on Build GRCh38
rs770274679 Chr.10: 132187056-132187056 on Build GRCh38
rs762562258 Chr.10: 132187049-132187050 on Build GRCh38
rs867797684 Chr.10: 132186851-132186851 on Build GRCh38
rs905798310 Chr.10: 132186863-132186863 on Build GRCh38
rs868198844 Chr.10: 132186907-132186907 on Build GRCh38
rs995637667 Chr.10: 132187197-132187197 on Build GRCh38
rs552405884 Chr.10: 132186929-132186929 on Build GRCh38
rs562642197 Chr.10: 132186941-132186941 on Build GRCh38
rs377655697 Chr.10: 132187126-132187126 on Build GRCh38
rs374310332 Chr.10: 132186768-132186768 on Build GRCh38
rs538865163 Chr.10: 132187117-132187117 on Build GRCh38
rs1027360438 Chr.10: 132187025-132187025 on Build GRCh38
rs1025736036 Chr.10: 132187009-132187009 on Build GRCh38
rs200531919 Chr.10: 132187088-132187088 on Build GRCh38
rs12778202 Chr.10: 132186871-132186871 on Build GRCh38
rs558325925 Chr.10: 132187182-132187182 on Build GRCh38
rs772736372 Chr.10: 132187130-132187130 on Build GRCh38
rs781697541 Chr.10: 132187112-132187112 on Build GRCh38
rs765257958 Chr.10: 132187027-132187033 on Build GRCh38
rs12772999 Chr.10: 132186872-132186872 on Build GRCh38
rs940307357 Chr.10: 132187169-132187169 on Build GRCh38
rs868469128 Chr.10: 132187077-132187077 on Build GRCh38
rs1004841811 Chr.10: 132186938-132186939 on Build GRCh38
rs888879174 Chr.10: 132187202-132187202 on Build GRCh38
rs771442689 Chr.10: 132187036-132187036 on Build GRCh38
rs1035775676 Chr.10: 132186896-132186896 on Build GRCh38
rs550713971 Chr.10: 132187001-132187001 on Build GRCh38
rs201598490 Chr.10: 132187038-132187038 on Build GRCh38
rs773785797 Chr.10: 132187122-132187122 on Build GRCh38
rs926696614 Chr.10: 132187140-132187140 on Build GRCh38
rs150208300 Chr.10: 132187132-132187132 on Build GRCh38
rs751859217 Chr.10: 132187109-132187109 on Build GRCh38
rs758403237 Chr.10: 132187017-132187017 on Build GRCh38
rs896740657 Chr.10: 132186928-132186928 on Build GRCh38
rs770795836 Chr.10: 132187014-132187018 on Build GRCh38
rs774018210 Chr.10: 132187062-132187062 on Build GRCh38
rs937991570 Chr.10: 132186923-132186923 on Build GRCh38
rs759599378 Chr.10: 132187019-132187033 on Build GRCh38

View Full Product Details