Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132190370-132190875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL4
Entrez Gene ID
10570
Gene Name
dihydropyrimidinase like 4
Gene Aliases
CRMP3, DRP-4, ULIP4
UniGene
Hs.100058
Gene Chromosome Location
Chr.10: 132184983-132205776 on Build GRCh38
Transcripts
NM_006426.2 XM_005252658.3 XM_011539125.2 XM_017015482.1 XM_017015483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966209 c.128+10C>T p.? Chr.10: 132190845-132190845 on Build GRCh38
7422942 c.83A>G p.D28G Chr.10: 132190790-132190790 on Build GRCh38
1703255 c.77A>G p.N26S Chr.10: 132190784-132190784 on Build GRCh38
7320066 c.46C>T p.R16C Chr.10: 132190753-132190753 on Build GRCh38
4012757 c.82G>A p.D28N Chr.10: 132190789-132190789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775593712 Chr.10: 132190843-132190843 on Build GRCh38
rs535157919 Chr.10: 132190538-132190538 on Build GRCh38
rs770128944 Chr.10: 132190459-132190459 on Build GRCh38
rs11146234 Chr.10: 132190424-132190424 on Build GRCh38
rs12253575 Chr.10: 132190404-132190404 on Build GRCh38
rs750692941 Chr.10: 132190779-132190779 on Build GRCh38
rs958720144 Chr.10: 132190394-132190394 on Build GRCh38
rs751255437 Chr.10: 132190820-132190820 on Build GRCh38
rs780244783 Chr.10: 132190715-132190715 on Build GRCh38
rs376647581 Chr.10: 132190845-132190845 on Build GRCh38
rs940529539 Chr.10: 132190468-132190468 on Build GRCh38
rs769692881 Chr.10: 132190822-132190822 on Build GRCh38
rs557199623 Chr.10: 132190711-132190711 on Build GRCh38
rs978114803 Chr.10: 132190677-132190677 on Build GRCh38
rs138267211 Chr.10: 132190818-132190818 on Build GRCh38
rs906400171 Chr.10: 132190566-132190566 on Build GRCh38
rs1050931548 Chr.10: 132190833-132190833 on Build GRCh38
rs977666144 Chr.10: 132190408-132190408 on Build GRCh38
rs557976404 Chr.10: 132190573-132190573 on Build GRCh38
rs201250019 Chr.10: 132190846-132190846 on Build GRCh38
rs776219190 Chr.10: 132190755-132190755 on Build GRCh38
rs755334082 Chr.10: 132190793-132190793 on Build GRCh38
rs751629149 Chr.10: 132190699-132190699 on Build GRCh38
rs371388070 Chr.10: 132190804-132190804 on Build GRCh38
rs531626377 Chr.10: 132190432-132190432 on Build GRCh38
rs968738481 Chr.10: 132190633-132190633 on Build GRCh38
rs564850167 Chr.10: 132190815-132190815 on Build GRCh38
rs920289076 Chr.10: 132190752-132190752 on Build GRCh38
rs759162414 Chr.10: 132190632-132190632 on Build GRCh38
rs200170880 Chr.10: 132190733-132190733 on Build GRCh38
rs890447163 Chr.10: 132190569-132190569 on Build GRCh38
rs377542379 Chr.10: 132190609-132190609 on Build GRCh38
rs969377762 Chr.10: 132190407-132190407 on Build GRCh38
rs932035694 Chr.10: 132190498-132190498 on Build GRCh38
rs770789966 Chr.10: 132190754-132190754 on Build GRCh38
rs933500572 Chr.10: 132190423-132190423 on Build GRCh38
rs748574576 Chr.10: 132190449-132190449 on Build GRCh38
rs993830060 Chr.10: 132190542-132190542 on Build GRCh38
rs767449073 Chr.10: 132190776-132190776 on Build GRCh38
rs137897999 Chr.10: 132190761-132190761 on Build GRCh38
rs750592185 Chr.10: 132190713-132190713 on Build GRCh38
rs576791001 Chr.10: 132190597-132190597 on Build GRCh38
rs143960105 Chr.10: 132190809-132190809 on Build GRCh38
rs1050473840 Chr.10: 132190522-132190524 on Build GRCh38
rs146177929 Chr.10: 132190753-132190753 on Build GRCh38
rs756227101 Chr.10: 132190787-132190787 on Build GRCh38
rs1001680638 Chr.10: 132190691-132190691 on Build GRCh38
rs779381002 Chr.10: 132190741-132190741 on Build GRCh38
rs771781117 Chr.10: 132190652-132190652 on Build GRCh38
rs910288138 Chr.10: 132190445-132190445 on Build GRCh38
rs778262326 Chr.10: 132190474-132190474 on Build GRCh38
rs372000002 Chr.10: 132190651-132190651 on Build GRCh38
rs149476256 Chr.10: 132190789-132190789 on Build GRCh38
rs765321126 Chr.10: 132190768-132190768 on Build GRCh38
rs748781826 Chr.10: 132190543-132190543 on Build GRCh38
rs188652417 Chr.10: 132190714-132190714 on Build GRCh38
rs1003361758 Chr.10: 132190579-132190579 on Build GRCh38
rs543679856 Chr.10: 132190625-132190625 on Build GRCh38
rs910697494 Chr.10: 132190739-132190739 on Build GRCh38
rs771538881 Chr.10: 132190745-132190745 on Build GRCh38
rs369982944 Chr.10: 132190848-132190848 on Build GRCh38
rs114236678 Chr.10: 132190403-132190403 on Build GRCh38
rs756810988 Chr.10: 132190836-132190837 on Build GRCh38
rs372435685 Chr.10: 132190849-132190849 on Build GRCh38
rs142076855 Chr.10: 132190411-132190411 on Build GRCh38
rs528164531 Chr.10: 132190803-132190803 on Build GRCh38
rs775561065 Chr.10: 132190773-132190773 on Build GRCh38
rs747724573 Chr.10: 132190742-132190742 on Build GRCh38
rs553748098 Chr.10: 132190641-132190641 on Build GRCh38
rs920700475 Chr.10: 132190485-132190485 on Build GRCh38
rs770494609 Chr.10: 132190606-132190606 on Build GRCh38
rs760145755 Chr.10: 132190816-132190816 on Build GRCh38
rs1037348514 Chr.10: 132190471-132190471 on Build GRCh38
rs762036818 Chr.10: 132190775-132190775 on Build GRCh38
rs766780274 Chr.10: 132190788-132190788 on Build GRCh38
rs551335158 Chr.10: 132190509-132190509 on Build GRCh38
rs1052690900 Chr.10: 132190574-132190574 on Build GRCh38
rs959977595 Chr.10: 132190447-132190447 on Build GRCh38
rs145633581 Chr.10: 132190431-132190431 on Build GRCh38
rs745721815 Chr.10: 132190820-132190820 on Build GRCh38
rs749807691 Chr.10: 132190725-132190725 on Build GRCh38

View Full Product Details