Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133262737-133263233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM8
Entrez Gene ID
101
Gene Name
ADAM metallopeptidase domain 8
Gene Aliases
CD156, CD156a, MS2
UniGene
Hs.501574
Gene Chromosome Location
Chr.10: 133262416-133276903 on Build GRCh38
Transcripts
NM_001109.4 NM_001164489.1 NM_001164490.1 XM_011539117.2 XM_017015465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129292 c.2456G>T p.G819V Chr.10: 133263175-133263175 on Build GRCh38
7315470 c.2431C>T p.P811S Chr.10: 133263200-133263200 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs894756670 Chr.10: 133262865-133262865 on Build GRCh38
rs759035466 Chr.10: 133263101-133263101 on Build GRCh38
rs764166381 Chr.10: 133263040-133263040 on Build GRCh38
rs34549109 Chr.10: 133263069-133263070 on Build GRCh38
rs922703620 Chr.10: 133262826-133262826 on Build GRCh38
rs1056009423 Chr.10: 133262864-133262864 on Build GRCh38
rs776766935 Chr.10: 133263151-133263151 on Build GRCh38
rs967740087 Chr.10: 133262905-133262905 on Build GRCh38
rs775251845 Chr.10: 133263107-133263107 on Build GRCh38
rs950236423 Chr.10: 133262907-133262907 on Build GRCh38
rs779149952 Chr.10: 133263154-133263154 on Build GRCh38
rs1018616906 Chr.10: 133262778-133262778 on Build GRCh38
rs935085348 Chr.10: 133262880-133262880 on Build GRCh38
rs1030646607 Chr.10: 133262767-133262767 on Build GRCh38
rs909863804 Chr.10: 133262840-133262840 on Build GRCh38
rs753825484 Chr.10: 133263193-133263193 on Build GRCh38
rs576947417 Chr.10: 133263137-133263137 on Build GRCh38
rs762796698 Chr.10: 133263033-133263033 on Build GRCh38
rs761658180 Chr.10: 133263124-133263124 on Build GRCh38
rs893482922 Chr.10: 133262893-133262893 on Build GRCh38
rs779061298 Chr.10: 133263071-133263071 on Build GRCh38
rs767434175 Chr.10: 133263129-133263129 on Build GRCh38
rs34253049 Chr.10: 133263080-133263080 on Build GRCh38
rs970999215 Chr.10: 133262765-133262765 on Build GRCh38
rs999207304 Chr.10: 133262990-133262990 on Build GRCh38
rs751516230 Chr.10: 133263043-133263043 on Build GRCh38
rs1029935213 Chr.10: 133263085-133263085 on Build GRCh38
rs1053407856 Chr.10: 133262882-133262883 on Build GRCh38
rs764347913 Chr.10: 133263187-133263187 on Build GRCh38
rs760526884 Chr.10: 133263190-133263190 on Build GRCh38
rs762887142 Chr.10: 133263109-133263109 on Build GRCh38
rs79659580 Chr.10: 133263177-133263177 on Build GRCh38
rs539284986 Chr.10: 133262899-133262899 on Build GRCh38
rs532626774 Chr.10: 133262829-133262829 on Build GRCh38
rs989327450 Chr.10: 133262777-133262777 on Build GRCh38
rs754395281 Chr.10: 133263197-133263197 on Build GRCh38
rs1003760936 Chr.10: 133262939-133262940 on Build GRCh38
rs983508225 Chr.10: 133263024-133263024 on Build GRCh38
rs982764398 Chr.10: 133262834-133262834 on Build GRCh38
rs982838006 Chr.10: 133263088-133263088 on Build GRCh38
rs538199355 Chr.10: 133263015-133263015 on Build GRCh38
rs569488390 Chr.10: 133262991-133262991 on Build GRCh38
rs181531266 Chr.10: 133262920-133262920 on Build GRCh38
rs753886877 Chr.10: 133263057-133263057 on Build GRCh38
rs769600887 Chr.10: 133263105-133263105 on Build GRCh38
rs758483166 Chr.10: 133262820-133262820 on Build GRCh38
rs906272945 Chr.10: 133262965-133262965 on Build GRCh38
rs771053269 Chr.10: 133263150-133263151 on Build GRCh38
rs545726760 Chr.10: 133263143-133263143 on Build GRCh38
rs781693749 Chr.10: 133263174-133263174 on Build GRCh38
rs755015233 Chr.10: 133263151-133263151 on Build GRCh38
rs909220505 Chr.10: 133262823-133262823 on Build GRCh38
rs750205020 Chr.10: 133263188-133263188 on Build GRCh38
rs950372424 Chr.10: 133263144-133263144 on Build GRCh38
rs1024010832 Chr.10: 133262902-133262902 on Build GRCh38
rs752560613 Chr.10: 133263025-133263025 on Build GRCh38
rs2230576 Chr.10: 133263092-133263092 on Build GRCh38
rs1012154943 Chr.10: 133262898-133262898 on Build GRCh38
rs766234759 Chr.10: 133263191-133263191 on Build GRCh38
rs924107821 Chr.10: 133263120-133263120 on Build GRCh38
rs748546654 Chr.10: 133263075-133263075 on Build GRCh38
rs957718866 Chr.10: 133262776-133262776 on Build GRCh38
rs556384229 Chr.10: 133263163-133263163 on Build GRCh38
rs776405242 Chr.10: 133263008-133263008 on Build GRCh38
rs778301208 Chr.10: 133263086-133263086 on Build GRCh38
rs397733581 Chr.10: 133263176-133263176 on Build GRCh38
rs745408689 Chr.10: 133263176-133263176 on Build GRCh38
rs1052148802 Chr.10: 133263000-133263000 on Build GRCh38
rs1021966006 Chr.10: 133262822-133262822 on Build GRCh38
rs756271386 Chr.10: 133263044-133263044 on Build GRCh38
rs554340450 Chr.10: 133263100-133263100 on Build GRCh38
rs759477757 Chr.10: 133262804-133262804 on Build GRCh38
rs200356896 Chr.10: 133263172-133263172 on Build GRCh38
rs1051509132 Chr.10: 133262846-133262846 on Build GRCh38
rs755172975 Chr.10: 133263070-133263070 on Build GRCh38
rs753724346 Chr.10: 133262808-133262808 on Build GRCh38
rs775336305 Chr.10: 133263185-133263185 on Build GRCh38
rs763963955 Chr.10: 133263116-133263116 on Build GRCh38
rs561363568 Chr.10: 133263173-133263173 on Build GRCh38
rs2995300 Chr.10: 133263019-133263019 on Build GRCh38
rs950818460 Chr.10: 133262782-133262782 on Build GRCh38
rs770542538 Chr.10: 133263093-133263093 on Build GRCh38
rs58683011 Chr.10: 133263176-133263177 on Build GRCh38
rs779991363 Chr.10: 133263053-133263053 on Build GRCh38
rs546715431 Chr.10: 133263117-133263117 on Build GRCh38
rs983395018 Chr.10: 133262805-133262805 on Build GRCh38
rs576260903 Chr.10: 133263170-133263170 on Build GRCh38
rs941357272 Chr.10: 133262859-133262859 on Build GRCh38

View Full Product Details