Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133270630-133271120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM8
Entrez Gene ID
101
Gene Name
ADAM metallopeptidase domain 8
Gene Aliases
CD156, CD156a, MS2
UniGene
Hs.501574
Gene Chromosome Location
Chr.10: 133262416-133276903 on Build GRCh38
Transcripts
NM_001109.4 NM_001164489.1 NM_001164490.1 XM_011539117.2 XM_017015465.1 XM_017015466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7334674 c.1496C>T p.S499F Chr.10: 133270949-133270949 on Build GRCh38
7354219 c.1498G>T p.G500W Chr.10: 133270947-133270947 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772836901 Chr.10: 133270898-133270899 on Build GRCh38
rs778693757 Chr.10: 133270908-133270908 on Build GRCh38
rs566868936 Chr.10: 133270922-133270922 on Build GRCh38
rs752104438 Chr.10: 133270765-133270765 on Build GRCh38
rs770098946 Chr.10: 133270832-133270832 on Build GRCh38
rs371796178 Chr.10: 133270742-133270742 on Build GRCh38
rs771227437 Chr.10: 133270670-133270670 on Build GRCh38
rs140434374 Chr.10: 133270930-133270930 on Build GRCh38
rs778105545 Chr.10: 133270688-133270688 on Build GRCh38
rs143152645 Chr.10: 133270800-133270800 on Build GRCh38
rs751407554 Chr.10: 133270872-133270872 on Build GRCh38
rs75429524 Chr.10: 133271029-133271029 on Build GRCh38
rs869257800 Chr.10: 133270731-133270732 on Build GRCh38
rs5789159 Chr.10: 133270728-133270729 on Build GRCh38
rs768948174 Chr.10: 133270928-133270928 on Build GRCh38
rs149612742 Chr.10: 133270984-133270984 on Build GRCh38
rs879834763 Chr.10: 133271031-133271031 on Build GRCh38
rs755094141 Chr.10: 133271062-133271062 on Build GRCh38
rs772279957 Chr.10: 133271082-133271082 on Build GRCh38
rs375446008 Chr.10: 133270859-133270859 on Build GRCh38
rs147308914 Chr.10: 133271001-133271001 on Build GRCh38
rs929406529 Chr.10: 133270873-133270873 on Build GRCh38
rs774796093 Chr.10: 133270874-133270874 on Build GRCh38
rs776626216 Chr.10: 133270743-133270743 on Build GRCh38
rs113710821 Chr.10: 133270720-133270720 on Build GRCh38
rs200848428 Chr.10: 133270725-133270725 on Build GRCh38
rs952887512 Chr.10: 133270714-133270714 on Build GRCh38
rs761100437 Chr.10: 133271004-133271004 on Build GRCh38
rs752531473 Chr.10: 133270958-133270958 on Build GRCh38
rs779344892 Chr.10: 133271063-133271063 on Build GRCh38
rs1018869176 Chr.10: 133270964-133270964 on Build GRCh38
rs754558649 Chr.10: 133270905-133270905 on Build GRCh38
rs749851228 Chr.10: 133270780-133270780 on Build GRCh38
rs761811967 Chr.10: 133270749-133270752 on Build GRCh38
rs574610493 Chr.10: 133271008-133271008 on Build GRCh38
rs781054039 Chr.10: 133270695-133270695 on Build GRCh38
rs139328971 Chr.10: 133271045-133271045 on Build GRCh38
rs138556925 Chr.10: 133270978-133270978 on Build GRCh38
rs749989019 Chr.10: 133270887-133270887 on Build GRCh38
rs3008321 Chr.10: 133270818-133270818 on Build GRCh38
rs267602422 Chr.10: 133271007-133271007 on Build GRCh38
rs754018009 Chr.10: 133271061-133271061 on Build GRCh38
rs774249455 Chr.10: 133270723-133270723 on Build GRCh38
rs764921978 Chr.10: 133271010-133271010 on Build GRCh38
rs749482164 Chr.10: 133270926-133270926 on Build GRCh38
rs558464821 Chr.10: 133271020-133271020 on Build GRCh38
rs756350439 Chr.10: 133270942-133270942 on Build GRCh38
rs1027138620 Chr.10: 133270700-133270700 on Build GRCh38
rs144989788 Chr.10: 133270977-133270977 on Build GRCh38
rs547298427 Chr.10: 133270919-133270919 on Build GRCh38
rs747891872 Chr.10: 133270915-133270915 on Build GRCh38
rs765812249 Chr.10: 133270946-133270946 on Build GRCh38
rs878974095 Chr.10: 133270840-133270840 on Build GRCh38
rs562283495 Chr.10: 133270772-133270772 on Build GRCh38
rs748580315 Chr.10: 133270990-133270990 on Build GRCh38
rs201299089 Chr.10: 133270727-133270728 on Build GRCh38
rs140754915 Chr.10: 133270957-133270957 on Build GRCh38
rs755556120 Chr.10: 133270897-133270897 on Build GRCh38
rs778889738 Chr.10: 133270814-133270814 on Build GRCh38
rs146751529 Chr.10: 133270903-133270903 on Build GRCh38
rs994462521 Chr.10: 133270686-133270686 on Build GRCh38
rs866260302 Chr.10: 133270880-133270880 on Build GRCh38
rs2995314 Chr.10: 133271097-133271097 on Build GRCh38
rs759210711 Chr.10: 133270956-133270956 on Build GRCh38
rs375821860 Chr.10: 133270749-133270749 on Build GRCh38
rs910352117 Chr.10: 133270761-133270761 on Build GRCh38
rs564253958 Chr.10: 133270744-133270744 on Build GRCh38
rs965502239 Chr.10: 133270748-133270748 on Build GRCh38
rs369088570 Chr.10: 133270822-133270822 on Build GRCh38
rs768096302 Chr.10: 133270885-133270885 on Build GRCh38
rs200740125 Chr.10: 133270963-133270963 on Build GRCh38
rs753332226 Chr.10: 133270948-133270948 on Build GRCh38
rs998384798 Chr.10: 133270995-133270995 on Build GRCh38
rs539109707 Chr.10: 133271005-133271005 on Build GRCh38
rs373192234 Chr.10: 133270985-133270985 on Build GRCh38
rs763437340 Chr.10: 133270863-133270863 on Build GRCh38
rs766113298 Chr.10: 133270900-133270900 on Build GRCh38
rs369406097 Chr.10: 133271101-133271101 on Build GRCh38
rs752337146 Chr.10: 133271019-133271019 on Build GRCh38
rs749352707 Chr.10: 133271000-133271000 on Build GRCh38
rs554084627 Chr.10: 133271046-133271046 on Build GRCh38
rs779541040 Chr.10: 133270792-133270792 on Build GRCh38
rs137890445 Chr.10: 133270947-133270947 on Build GRCh38
rs33946523 Chr.10: 133270729-133270730 on Build GRCh38
rs751452101 Chr.10: 133270966-133270966 on Build GRCh38
rs760298733 Chr.10: 133270945-133270945 on Build GRCh38
rs149084614 Chr.10: 133270791-133270791 on Build GRCh38
rs757768476 Chr.10: 133270770-133270770 on Build GRCh38
rs756810501 Chr.10: 133270690-133270690 on Build GRCh38
rs141276821 Chr.10: 133270943-133270943 on Build GRCh38
rs1049028483 Chr.10: 133270890-133270890 on Build GRCh38
rs367927115 Chr.10: 133270689-133270689 on Build GRCh38
rs576461106 Chr.10: 133270680-133270680 on Build GRCh38
rs767186548 Chr.10: 133270786-133270787 on Build GRCh38
rs760468817 Chr.10: 133271030-133271031 on Build GRCh38
rs760802790 Chr.10: 133270740-133270740 on Build GRCh38
rs530968981 Chr.10: 133270783-133270783 on Build GRCh38
rs372927486 Chr.10: 133270916-133270916 on Build GRCh38
rs754716395 Chr.10: 133270808-133270808 on Build GRCh38
rs745517326 Chr.10: 133270696-133270696 on Build GRCh38
rs561598864 Chr.10: 133270821-133270821 on Build GRCh38
rs748994860 Chr.10: 133270722-133270722 on Build GRCh38
rs774885826 Chr.10: 133270929-133270929 on Build GRCh38
rs777544211 Chr.10: 133270917-133270917 on Build GRCh38
rs10632315 Chr.10: 133270730-133270731 on Build GRCh38
rs758170777 Chr.10: 133270962-133270962 on Build GRCh38
rs909210915 Chr.10: 133271087-133271087 on Build GRCh38
rs763200010 Chr.10: 133270764-133270764 on Build GRCh38
rs545492678 Chr.10: 133270702-133270702 on Build GRCh38
rs747589598 Chr.10: 133271098-133271098 on Build GRCh38
rs751316677 Chr.10: 133271038-133271038 on Build GRCh38
rs949477572 Chr.10: 133271092-133271092 on Build GRCh38

View Full Product Details