Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133362344-133362889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHS1
Entrez Gene ID
1892
Gene Name
enoyl-CoA hydratase, short chain 1
Gene Aliases
ECHS1D, SCEH
UniGene
Hs.76394
Gene Chromosome Location
Chr.10: 133362483-133373404 on Build GRCh38
Transcripts
NM_004092.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675685 c.861C>A p.F287L Chr.10: 133362880-133362880 on Build GRCh38
4974003 c.872G>T p.*291L Chr.10: 133362869-133362869 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs887901402 Chr.10: 133362568-133362568 on Build GRCh38
rs566614463 Chr.10: 133362540-133362540 on Build GRCh38
rs537538341 Chr.10: 133362607-133362607 on Build GRCh38
rs916232257 Chr.10: 133362507-133362507 on Build GRCh38
rs998196658 Chr.10: 133362603-133362603 on Build GRCh38
rs764304433 Chr.10: 133362832-133362832 on Build GRCh38
rs546897089 Chr.10: 133362526-133362526 on Build GRCh38
rs755635585 Chr.10: 133362855-133362855 on Build GRCh38
rs778240697 Chr.10: 133362478-133362478 on Build GRCh38
rs765943111 Chr.10: 133362863-133362863 on Build GRCh38
rs748413661 Chr.10: 133362595-133362595 on Build GRCh38
rs1047891670 Chr.10: 133362701-133362701 on Build GRCh38
rs999730745 Chr.10: 133362634-133362634 on Build GRCh38
rs753313551 Chr.10: 133362416-133362416 on Build GRCh38
rs538932115 Chr.10: 133362567-133362567 on Build GRCh38
rs1039299470 Chr.10: 133362674-133362674 on Build GRCh38
rs902037398 Chr.10: 133362414-133362414 on Build GRCh38
rs942101221 Chr.10: 133362586-133362586 on Build GRCh38
rs943347201 Chr.10: 133362637-133362637 on Build GRCh38
rs1039094531 Chr.10: 133362767-133362767 on Build GRCh38
rs767247598 Chr.10: 133362840-133362840 on Build GRCh38
rs372081807 Chr.10: 133362828-133362828 on Build GRCh38
rs990573572 Chr.10: 133362720-133362720 on Build GRCh38
rs983943076 Chr.10: 133362747-133362747 on Build GRCh38
rs886596748 Chr.10: 133362715-133362715 on Build GRCh38
rs113149678 Chr.10: 133362716-133362716 on Build GRCh38
rs373661920 Chr.10: 133362732-133362732 on Build GRCh38
rs573873276 Chr.10: 133362726-133362726 on Build GRCh38
rs576889001 Chr.10: 133362739-133362739 on Build GRCh38
rs183588815 Chr.10: 133362733-133362733 on Build GRCh38
rs191814768 Chr.10: 133362443-133362443 on Build GRCh38
rs745319690 Chr.10: 133362700-133362700 on Build GRCh38
rs957799060 Chr.10: 133362718-133362718 on Build GRCh38
rs990186571 Chr.10: 133362631-133362631 on Build GRCh38
rs774760601 Chr.10: 133362838-133362838 on Build GRCh38
rs568879243 Chr.10: 133362602-133362602 on Build GRCh38
rs957768099 Chr.10: 133362465-133362465 on Build GRCh38
rs529952344 Chr.10: 133362509-133362509 on Build GRCh38
rs991443918 Chr.10: 133362538-133362538 on Build GRCh38
rs376401874 Chr.10: 133362814-133362814 on Build GRCh38
rs1029435556 Chr.10: 133362423-133362423 on Build GRCh38
rs756856221 Chr.10: 133362633-133362633 on Build GRCh38
rs971269703 Chr.10: 133362577-133362577 on Build GRCh38
rs1038508500 Chr.10: 133362725-133362725 on Build GRCh38
rs902691272 Chr.10: 133362626-133362626 on Build GRCh38
rs929774536 Chr.10: 133362555-133362555 on Build GRCh38
rs914611110 Chr.10: 133362858-133362858 on Build GRCh38
rs1003887236 Chr.10: 133362447-133362447 on Build GRCh38
rs573418441 Chr.10: 133362746-133362746 on Build GRCh38
rs1016632181 Chr.10: 133362496-133362496 on Build GRCh38
rs188884786 Chr.10: 133362738-133362738 on Build GRCh38
rs778552582 Chr.10: 133362664-133362664 on Build GRCh38
rs1032061270 Chr.10: 133362458-133362458 on Build GRCh38
rs943669027 Chr.10: 133362375-133362375 on Build GRCh38
rs4604 Chr.10: 133362587-133362587 on Build GRCh38
rs755107628 Chr.10: 133362560-133362560 on Build GRCh38
rs750148216 Chr.10: 133362845-133362845 on Build GRCh38
rs1050040 Chr.10: 133362754-133362754 on Build GRCh38
rs867868717 Chr.10: 133362527-133362527 on Build GRCh38
rs41313481 Chr.10: 133362417-133362417 on Build GRCh38
rs762331214 Chr.10: 133362839-133362839 on Build GRCh38
rs1803952 Chr.10: 133362647-133362647 on Build GRCh38
rs923493486 Chr.10: 133362688-133362688 on Build GRCh38
rs116786579 Chr.10: 133362759-133362759 on Build GRCh38
rs990934135 Chr.10: 133362501-133362501 on Build GRCh38
rs15116 Chr.10: 133362756-133362756 on Build GRCh38
rs955870758 Chr.10: 133362783-133362783 on Build GRCh38
rs1050126 Chr.10: 133362548-133362548 on Build GRCh38
rs966846396 Chr.10: 133362436-133362439 on Build GRCh38
rs74164159 Chr.10: 133362391-133362391 on Build GRCh38

View Full Product Details